Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66482335-66482864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP3
Entrez Gene ID
10072
Gene Name
dipeptidyl peptidase 3
Gene Aliases
DPPIII
UniGene
Hs.502914
Gene Chromosome Location
Chr.11: 66480013-66509659 on Build GRCh38
Transcripts
NM_001256670.1 NM_005700.4 NM_130443.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4622257 c.192C>T p.S64S Chr.11: 66482392-66482392 on Build GRCh38
4919883 c.216C>T p.P72P Chr.11: 66482416-66482416 on Build GRCh38
5580904 c.195C>A p.R65R Chr.11: 66482395-66482395 on Build GRCh38
1356222 c.190A>G p.S64G Chr.11: 66482390-66482390 on Build GRCh38
1638937 c.270+8C>A p.? Chr.11: 66482478-66482478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375779987 Chr.11: 66482360-66482360 on Build GRCh38
rs200170886 Chr.11: 66482495-66482495 on Build GRCh38
rs147041520 Chr.11: 66482662-66482662 on Build GRCh38
rs748629398 Chr.11: 66482416-66482416 on Build GRCh38
rs907478885 Chr.11: 66482379-66482379 on Build GRCh38
rs202184719 Chr.11: 66482394-66482394 on Build GRCh38
rs928277220 Chr.11: 66482673-66482673 on Build GRCh38
rs772749096 Chr.11: 66482419-66482419 on Build GRCh38
rs761706330 Chr.11: 66482467-66482467 on Build GRCh38
rs34243583 Chr.11: 66482366-66482366 on Build GRCh38
rs966222654 Chr.11: 66482692-66482692 on Build GRCh38
rs945727286 Chr.11: 66482792-66482792 on Build GRCh38
rs577731291 Chr.11: 66482405-66482405 on Build GRCh38
rs113075304 Chr.11: 66482567-66482567 on Build GRCh38
rs765310681 Chr.11: 66482464-66482464 on Build GRCh38
rs374335679 Chr.11: 66482627-66482627 on Build GRCh38
rs11826683 Chr.11: 66482427-66482427 on Build GRCh38
rs755872748 Chr.11: 66482502-66482502 on Build GRCh38
rs928552853 Chr.11: 66482835-66482835 on Build GRCh38
rs768363395 Chr.11: 66482488-66482488 on Build GRCh38
rs371108340 Chr.11: 66482409-66482409 on Build GRCh38
rs953116455 Chr.11: 66482761-66482763 on Build GRCh38
rs199572961 Chr.11: 66482455-66482455 on Build GRCh38
rs779727702 Chr.11: 66482503-66482503 on Build GRCh38
rs907066049 Chr.11: 66482543-66482543 on Build GRCh38
rs199654699 Chr.11: 66482426-66482426 on Build GRCh38
rs1006404554 Chr.11: 66482817-66482817 on Build GRCh38
rs754622319 Chr.11: 66482509-66482509 on Build GRCh38
rs922067874 Chr.11: 66482748-66482748 on Build GRCh38
rs189834963 Chr.11: 66482634-66482634 on Build GRCh38
rs200572164 Chr.11: 66482471-66482471 on Build GRCh38
rs887995453 Chr.11: 66482791-66482791 on Build GRCh38
rs778459374 Chr.11: 66482511-66482511 on Build GRCh38
rs200361217 Chr.11: 66482402-66482402 on Build GRCh38
rs1056055352 Chr.11: 66482779-66482779 on Build GRCh38
rs147658388 Chr.11: 66482403-66482403 on Build GRCh38
rs1045395773 Chr.11: 66482358-66482358 on Build GRCh38
rs989859442 Chr.11: 66482767-66482767 on Build GRCh38
rs187410072 Chr.11: 66482404-66482404 on Build GRCh38
rs774010092 Chr.11: 66482373-66482373 on Build GRCh38
rs373915905 Chr.11: 66482529-66482529 on Build GRCh38
rs752805050 Chr.11: 66482466-66482466 on Build GRCh38
rs750216926 Chr.11: 66482482-66482482 on Build GRCh38
rs753846896 Chr.11: 66482375-66482375 on Build GRCh38
rs766675703 Chr.11: 66482370-66482370 on Build GRCh38
rs1028069150 Chr.11: 66482604-66482604 on Build GRCh38
rs201569456 Chr.11: 66482420-66482420 on Build GRCh38
rs774093297 Chr.11: 66482680-66482680 on Build GRCh38
rs985334196 Chr.11: 66482460-66482460 on Build GRCh38
rs147041684 Chr.11: 66482401-66482401 on Build GRCh38
rs1045454039 Chr.11: 66482600-66482600 on Build GRCh38
rs201636331 Chr.11: 66482456-66482456 on Build GRCh38
rs757207991 Chr.11: 66482395-66482395 on Build GRCh38
rs1189692 Chr.11: 66482544-66482544 on Build GRCh38
rs767490295 Chr.11: 66482481-66482481 on Build GRCh38
rs150218434 Chr.11: 66482459-66482459 on Build GRCh38
rs371535435 Chr.11: 66482558-66482558 on Build GRCh38
rs536918372 Chr.11: 66482417-66482417 on Build GRCh38
rs945761431 Chr.11: 66482575-66482575 on Build GRCh38
rs749178354 Chr.11: 66482361-66482362 on Build GRCh38
rs952076108 Chr.11: 66482605-66482605 on Build GRCh38
rs181594909 Chr.11: 66482819-66482819 on Build GRCh38
rs548345938 Chr.11: 66482502-66482513 on Build GRCh38
rs1012250866 Chr.11: 66482603-66482603 on Build GRCh38
rs575752647 Chr.11: 66482393-66482393 on Build GRCh38
rs976267406 Chr.11: 66482727-66482727 on Build GRCh38
rs531545268 Chr.11: 66482785-66482785 on Build GRCh38
rs114567543 Chr.11: 66482381-66482381 on Build GRCh38
rs957230225 Chr.11: 66482565-66482565 on Build GRCh38
rs1030270097 Chr.11: 66482433-66482433 on Build GRCh38
rs1006764379 Chr.11: 66482413-66482413 on Build GRCh38
rs768800260 Chr.11: 66482506-66482506 on Build GRCh38
rs937616982 Chr.11: 66482699-66482699 on Build GRCh38
rs201532921 Chr.11: 66482489-66482489 on Build GRCh38
rs990355017 Chr.11: 66482566-66482566 on Build GRCh38
rs200532781 Chr.11: 66482410-66482410 on Build GRCh38

View Full Product Details