Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66523365-66523860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBS1
Entrez Gene ID
582
Gene Name
Bardet-Biedl syndrome 1
Gene Aliases
BBS2L2
UniGene
Hs.502915
Gene Chromosome Location
Chr.11: 66510648-66533613 on Build GRCh38
Transcripts
NM_024649.4 XM_005273874.3

Gene Symbol
ZDHHC24
Entrez Gene ID
254359
Gene Name
zinc finger DHHC-type containing 24
Gene Aliases
-
UniGene
Hs.659933
Gene Chromosome Location
Chr.11: 66520625-66546200 on Build GRCh38
Transcripts
NM_024649.4 XM_005273874.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4458200 c.1186C>T p.R396C Chr.11: 66523847-66523847 on Build GRCh38
1470566 c.917G>A p.S306N Chr.11: 66523542-66523542 on Build GRCh38
4665244 c.934C>T p.H312Y Chr.11: 66523559-66523559 on Build GRCh38
5649873 c.1071G>A p.K357K Chr.11: 66523732-66523732 on Build GRCh38
4405306 c.1160T>A p.L387Q Chr.11: 66523821-66523821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776322401 Chr.11: 66523492-66523492 on Build GRCh38
rs747656068 Chr.11: 66523826-66523826 on Build GRCh38
rs1057516427 Chr.11: 66523480-66523480 on Build GRCh38
rs888475582 Chr.11: 66523524-66523524 on Build GRCh38
rs368824329 Chr.11: 66523696-66523696 on Build GRCh38
rs1037134217 Chr.11: 66523526-66523526 on Build GRCh38
rs765388270 Chr.11: 66523821-66523822 on Build GRCh38
rs764886049 Chr.11: 66523418-66523418 on Build GRCh38
rs751914638 Chr.11: 66523435-66523435 on Build GRCh38
rs759690825 Chr.11: 66523777-66523777 on Build GRCh38
rs942862410 Chr.11: 66523538-66523538 on Build GRCh38
rs753241093 Chr.11: 66523428-66523428 on Build GRCh38
rs755942584 Chr.11: 66523715-66523715 on Build GRCh38
rs755994026 Chr.11: 66523805-66523805 on Build GRCh38
rs374822711 Chr.11: 66523609-66523609 on Build GRCh38
rs56177555 Chr.11: 66523451-66523451 on Build GRCh38
rs199986163 Chr.11: 66523746-66523746 on Build GRCh38
rs768292767 Chr.11: 66523736-66523736 on Build GRCh38
rs777705416 Chr.11: 66523475-66523475 on Build GRCh38
rs772509403 Chr.11: 66523744-66523744 on Build GRCh38
rs773664520 Chr.11: 66523741-66523741 on Build GRCh38
rs571201308 Chr.11: 66523659-66523659 on Build GRCh38
rs372092113 Chr.11: 66523515-66523515 on Build GRCh38
rs756220791 Chr.11: 66523534-66523534 on Build GRCh38
rs778610841 Chr.11: 66523819-66523819 on Build GRCh38
rs754022677 Chr.11: 66523693-66523693 on Build GRCh38
rs771291825 Chr.11: 66523839-66523839 on Build GRCh38
rs747459381 Chr.11: 66523838-66523838 on Build GRCh38
rs780784789 Chr.11: 66523583-66523583 on Build GRCh38
rs971605369 Chr.11: 66523755-66523755 on Build GRCh38
rs776855360 Chr.11: 66523775-66523775 on Build GRCh38
rs866867885 Chr.11: 66523819-66523821 on Build GRCh38
rs745672331 Chr.11: 66523586-66523586 on Build GRCh38
rs896125678 Chr.11: 66523651-66523651 on Build GRCh38
rs905064850 Chr.11: 66523663-66523663 on Build GRCh38
rs766632086 Chr.11: 66523685-66523685 on Build GRCh38
rs201872547 Chr.11: 66523808-66523808 on Build GRCh38
rs768239720 Chr.11: 66523616-66523616 on Build GRCh38
rs762268360 Chr.11: 66523529-66523529 on Build GRCh38
rs779935398 Chr.11: 66523807-66523807 on Build GRCh38
rs201805739 Chr.11: 66523432-66523432 on Build GRCh38
rs759166794 Chr.11: 66523493-66523493 on Build GRCh38
rs113994179 Chr.11: 66523453-66523453 on Build GRCh38
rs762566248 Chr.11: 66523514-66523514 on Build GRCh38
rs529212299 Chr.11: 66523479-66523479 on Build GRCh38
rs745700176 Chr.11: 66523712-66523712 on Build GRCh38
rs748899217 Chr.11: 66523726-66523726 on Build GRCh38
rs1000438556 Chr.11: 66523656-66523656 on Build GRCh38
rs533592610 Chr.11: 66523669-66523669 on Build GRCh38
rs780246167 Chr.11: 66523559-66523559 on Build GRCh38
rs766684863 Chr.11: 66523754-66523754 on Build GRCh38
rs770668729 Chr.11: 66523484-66523484 on Build GRCh38
rs747686856 Chr.11: 66523743-66523743 on Build GRCh38
rs925660059 Chr.11: 66523426-66523426 on Build GRCh38
rs751978471 Chr.11: 66523521-66523521 on Build GRCh38
rs1049027938 Chr.11: 66523814-66523814 on Build GRCh38
rs1032626950 Chr.11: 66523667-66523667 on Build GRCh38
rs755076424 Chr.11: 66523694-66523694 on Build GRCh38
rs1057518418 Chr.11: 66523536-66523538 on Build GRCh38
rs781177816 Chr.11: 66523706-66523706 on Build GRCh38
rs753972808 Chr.11: 66523560-66523560 on Build GRCh38
rs755023569 Chr.11: 66523564-66523564 on Build GRCh38
rs759627328 Chr.11: 66523592-66523594 on Build GRCh38
rs901307899 Chr.11: 66523555-66523555 on Build GRCh38
rs553625102 Chr.11: 66523679-66523679 on Build GRCh38
rs777721856 Chr.11: 66523573-66523573 on Build GRCh38
rs201437917 Chr.11: 66523603-66523603 on Build GRCh38
rs746875134 Chr.11: 66523577-66523577 on Build GRCh38
rs1057517332 Chr.11: 66523454-66523454 on Build GRCh38
rs867676232 Chr.11: 66523415-66523415 on Build GRCh38
rs773617456 Chr.11: 66523834-66523834 on Build GRCh38
rs765494351 Chr.11: 66523778-66523778 on Build GRCh38
rs898584166 Chr.11: 66523677-66523677 on Build GRCh38
rs963070589 Chr.11: 66523437-66523437 on Build GRCh38
rs200350565 Chr.11: 66523705-66523705 on Build GRCh38
rs754741457 Chr.11: 66523815-66523815 on Build GRCh38
rs587777830 Chr.11: 66523476-66523476 on Build GRCh38
rs767395683 Chr.11: 66523793-66523793 on Build GRCh38
rs35209408 Chr.11: 66523792-66523792 on Build GRCh38
rs199692416 Chr.11: 66523831-66523831 on Build GRCh38
rs869025205 Chr.11: 66523784-66523784 on Build GRCh38
rs794727006 Chr.11: 66523512-66523512 on Build GRCh38
rs202036802 Chr.11: 66523429-66523429 on Build GRCh38
rs200758309 Chr.11: 66523748-66523748 on Build GRCh38
rs555503958 Chr.11: 66523786-66523786 on Build GRCh38
rs145094101 Chr.11: 66523512-66523512 on Build GRCh38
rs767925430 Chr.11: 66523678-66523678 on Build GRCh38
rs920296440 Chr.11: 66523816-66523817 on Build GRCh38
rs1057516371 Chr.11: 66523753-66523753 on Build GRCh38
rs778802597 Chr.11: 66523695-66523695 on Build GRCh38
rs191924860 Chr.11: 66523627-66523627 on Build GRCh38
rs746775716 Chr.11: 66523483-66523483 on Build GRCh38
rs544274797 Chr.11: 66523794-66523794 on Build GRCh38
rs761009605 Chr.11: 66523681-66523681 on Build GRCh38
rs557602559 Chr.11: 66523654-66523654 on Build GRCh38
rs774942506 Chr.11: 66523620-66523620 on Build GRCh38
rs547524683 Chr.11: 66523498-66523498 on Build GRCh38
rs1057517143 Chr.11: 66523496-66523496 on Build GRCh38
rs748076552 Chr.11: 66523468-66523468 on Build GRCh38
rs1057516661 Chr.11: 66523723-66523723 on Build GRCh38
rs780169168 Chr.11: 66523465-66523465 on Build GRCh38
rs142243482 Chr.11: 66523753-66523753 on Build GRCh38
rs1004279908 Chr.11: 66523525-66523525 on Build GRCh38
rs775261434 Chr.11: 66523608-66523608 on Build GRCh38
rs750668484 Chr.11: 66523533-66523533 on Build GRCh38
rs769422545 Chr.11: 66523502-66523502 on Build GRCh38

View Full Product Details