Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66592084-66592583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC87
Entrez Gene ID
55231
Gene Name
coiled-coil domain containing 87
Gene Aliases
-
UniGene
Hs.121072
Gene Chromosome Location
Chr.11: 66590169-66593083 on Build GRCh38
Transcripts
NM_018219.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3452136 c.881C>T p.T294I Chr.11: 66592135-66592135 on Build GRCh38
4508145 c.766C>T p.P256S Chr.11: 66592250-66592250 on Build GRCh38
5991240 c.575T>G p.V192G Chr.11: 66592441-66592441 on Build GRCh38
2040523 c.920G>A p.R307Q Chr.11: 66592096-66592096 on Build GRCh38
3452137 c.869C>T p.P290L Chr.11: 66592147-66592147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754357470 Chr.11: 66592379-66592379 on Build GRCh38
rs755538055 Chr.11: 66592384-66592384 on Build GRCh38
rs755683473 Chr.11: 66592369-66592369 on Build GRCh38
rs776905672 Chr.11: 66592421-66592421 on Build GRCh38
rs751470912 Chr.11: 66592330-66592330 on Build GRCh38
rs147889901 Chr.11: 66592328-66592328 on Build GRCh38
rs748593019 Chr.11: 66592388-66592388 on Build GRCh38
rs151064106 Chr.11: 66592544-66592544 on Build GRCh38
rs943750561 Chr.11: 66592186-66592186 on Build GRCh38
rs761694573 Chr.11: 66592455-66592455 on Build GRCh38
rs558926916 Chr.11: 66592333-66592333 on Build GRCh38
rs760459661 Chr.11: 66592548-66592548 on Build GRCh38
rs997425258 Chr.11: 66592339-66592339 on Build GRCh38
rs141535876 Chr.11: 66592414-66592414 on Build GRCh38
rs777937456 Chr.11: 66592505-66592505 on Build GRCh38
rs370432829 Chr.11: 66592532-66592532 on Build GRCh38
rs1110707 Chr.11: 66592550-66592550 on Build GRCh38
rs968953280 Chr.11: 66592371-66592371 on Build GRCh38
rs751266637 Chr.11: 66592214-66592214 on Build GRCh38
rs779360543 Chr.11: 66592387-66592387 on Build GRCh38
rs267603130 Chr.11: 66592472-66592472 on Build GRCh38
rs749079356 Chr.11: 66592356-66592356 on Build GRCh38
rs761589010 Chr.11: 66592535-66592535 on Build GRCh38
rs745995805 Chr.11: 66592529-66592529 on Build GRCh38
rs536620132 Chr.11: 66592269-66592269 on Build GRCh38
rs924333101 Chr.11: 66592140-66592140 on Build GRCh38
rs753840413 Chr.11: 66592517-66592517 on Build GRCh38
rs969880517 Chr.11: 66592540-66592540 on Build GRCh38
rs768961190 Chr.11: 66592178-66592178 on Build GRCh38
rs200965633 Chr.11: 66592257-66592257 on Build GRCh38
rs779419042 Chr.11: 66592493-66592493 on Build GRCh38
rs774589996 Chr.11: 66592180-66592180 on Build GRCh38
rs779474431 Chr.11: 66592372-66592372 on Build GRCh38
rs752631966 Chr.11: 66592208-66592208 on Build GRCh38
rs766520683 Chr.11: 66592426-66592426 on Build GRCh38
rs762029595 Chr.11: 66592188-66592188 on Build GRCh38
rs373078402 Chr.11: 66592312-66592312 on Build GRCh38
rs867712350 Chr.11: 66592160-66592160 on Build GRCh38
rs546601638 Chr.11: 66592422-66592422 on Build GRCh38
rs746618032 Chr.11: 66592137-66592137 on Build GRCh38
rs764584208 Chr.11: 66592559-66592559 on Build GRCh38
rs756722434 Chr.11: 66592139-66592139 on Build GRCh38
rs753725382 Chr.11: 66592106-66592106 on Build GRCh38
rs780708702 Chr.11: 66592229-66592229 on Build GRCh38
rs769013983 Chr.11: 66592262-66592262 on Build GRCh38
rs772275388 Chr.11: 66592391-66592391 on Build GRCh38
rs771146081 Chr.11: 66592420-66592420 on Build GRCh38
rs376045221 Chr.11: 66592485-66592485 on Build GRCh38
rs867624554 Chr.11: 66592294-66592294 on Build GRCh38
rs758790803 Chr.11: 66592503-66592503 on Build GRCh38
rs548250985 Chr.11: 66592247-66592247 on Build GRCh38
rs780562668 Chr.11: 66592251-66592251 on Build GRCh38
rs931269126 Chr.11: 66592213-66592213 on Build GRCh38
rs767508357 Chr.11: 66592193-66592193 on Build GRCh38
rs756976292 Chr.11: 66592350-66592350 on Build GRCh38
rs768476858 Chr.11: 66592444-66592444 on Build GRCh38
rs543801025 Chr.11: 66592469-66592469 on Build GRCh38
rs568214022 Chr.11: 66592423-66592423 on Build GRCh38
rs1021844021 Chr.11: 66592344-66592344 on Build GRCh38
rs144906506 Chr.11: 66592169-66592169 on Build GRCh38
rs183896760 Chr.11: 66592310-66592310 on Build GRCh38
rs767252116 Chr.11: 66592354-66592354 on Build GRCh38
rs200400457 Chr.11: 66592407-66592407 on Build GRCh38
rs574904032 Chr.11: 66592435-66592435 on Build GRCh38
rs141059944 Chr.11: 66592162-66592162 on Build GRCh38
rs760829152 Chr.11: 66592309-66592309 on Build GRCh38
rs773407347 Chr.11: 66592203-66592203 on Build GRCh38
rs915687484 Chr.11: 66592506-66592506 on Build GRCh38
rs150149433 Chr.11: 66592558-66592558 on Build GRCh38
rs777588203 Chr.11: 66592128-66592128 on Build GRCh38
rs753437521 Chr.11: 66592553-66592553 on Build GRCh38
rs780692825 Chr.11: 66592143-66592143 on Build GRCh38
rs760640773 Chr.11: 66592206-66592206 on Build GRCh38
rs758244646 Chr.11: 66592127-66592127 on Build GRCh38
rs1037624256 Chr.11: 66592190-66592190 on Build GRCh38
rs759687487 Chr.11: 66592426-66592426 on Build GRCh38
rs556454265 Chr.11: 66592304-66592304 on Build GRCh38
rs745361299 Chr.11: 66592145-66592145 on Build GRCh38
rs76910333 Chr.11: 66592487-66592487 on Build GRCh38
rs748456915 Chr.11: 66592283-66592283 on Build GRCh38
rs188685970 Chr.11: 66592364-66592364 on Build GRCh38
rs753104546 Chr.11: 66592501-66592501 on Build GRCh38
rs752578641 Chr.11: 66592120-66592120 on Build GRCh38
rs780155949 Chr.11: 66592427-66592427 on Build GRCh38
rs763910776 Chr.11: 66592211-66592211 on Build GRCh38
rs376232930 Chr.11: 66592288-66592288 on Build GRCh38
rs778712229 Chr.11: 66592109-66592109 on Build GRCh38
rs1020248305 Chr.11: 66592394-66592394 on Build GRCh38
rs780284264 Chr.11: 66592243-66592243 on Build GRCh38
rs747383629 Chr.11: 66592508-66592508 on Build GRCh38
rs368118677 Chr.11: 66592117-66592117 on Build GRCh38
rs767177309 Chr.11: 66592459-66592459 on Build GRCh38
rs771914166 Chr.11: 66592539-66592539 on Build GRCh38
rs760772674 Chr.11: 66592302-66592303 on Build GRCh38
rs17853294 Chr.11: 66592365-66592365 on Build GRCh38
rs781536350 Chr.11: 66592523-66592523 on Build GRCh38
rs771342455 Chr.11: 66592515-66592515 on Build GRCh38
rs372682665 Chr.11: 66592429-66592429 on Build GRCh38
rs765267350 Chr.11: 66592207-66592207 on Build GRCh38
rs756995987 Chr.11: 66592218-66592218 on Build GRCh38
rs770403475 Chr.11: 66592154-66592154 on Build GRCh38
rs368879587 Chr.11: 66592108-66592108 on Build GRCh38
rs915734754 Chr.11: 66592171-66592171 on Build GRCh38
rs766003568 Chr.11: 66592474-66592474 on Build GRCh38

View Full Product Details