Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66592485-66592979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC87
Entrez Gene ID
55231
Gene Name
coiled-coil domain containing 87
Gene Aliases
-
UniGene
Hs.121072
Gene Chromosome Location
Chr.11: 66590169-66593083 on Build GRCh38
Transcripts
NM_018219.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7317651 c.474C>T p.L158L Chr.11: 66592542-66592542 on Build GRCh38
6622530 c.78_79insC p.T27fs*2 Chr.11: 66592937-66592938 on Build GRCh38
2040537 c.289G>C p.E97Q Chr.11: 66592727-66592727 on Build GRCh38
74046 c.250A>T p.I84F Chr.11: 66592766-66592766 on Build GRCh38
5988980 c.125G>T p.G42V Chr.11: 66592891-66592891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140452825 Chr.11: 66592628-66592628 on Build GRCh38
rs768490903 Chr.11: 66592897-66592897 on Build GRCh38
rs920409985 Chr.11: 66592901-66592901 on Build GRCh38
rs746704575 Chr.11: 66592867-66592868 on Build GRCh38
rs770725502 Chr.11: 66592689-66592689 on Build GRCh38
rs1014340190 Chr.11: 66592823-66592823 on Build GRCh38
rs151064106 Chr.11: 66592544-66592544 on Build GRCh38
rs754049542 Chr.11: 66592772-66592772 on Build GRCh38
rs757676657 Chr.11: 66592591-66592591 on Build GRCh38
rs760459661 Chr.11: 66592548-66592548 on Build GRCh38
rs749375590 Chr.11: 66592614-66592614 on Build GRCh38
rs760976469 Chr.11: 66592816-66592816 on Build GRCh38
rs911812829 Chr.11: 66592923-66592923 on Build GRCh38
rs777937456 Chr.11: 66592505-66592505 on Build GRCh38
rs370432829 Chr.11: 66592532-66592532 on Build GRCh38
rs150176794 Chr.11: 66592735-66592735 on Build GRCh38
rs999049522 Chr.11: 66592716-66592716 on Build GRCh38
rs1110707 Chr.11: 66592550-66592550 on Build GRCh38
rs868616442 Chr.11: 66592906-66592906 on Build GRCh38
rs780321060 Chr.11: 66592663-66592663 on Build GRCh38
rs746051003 Chr.11: 66592605-66592605 on Build GRCh38
rs755035257 Chr.11: 66592673-66592673 on Build GRCh38
rs755024739 Chr.11: 66592586-66592586 on Build GRCh38
rs761589010 Chr.11: 66592535-66592535 on Build GRCh38
rs200959265 Chr.11: 66592617-66592617 on Build GRCh38
rs774926602 Chr.11: 66592636-66592636 on Build GRCh38
rs763623346 Chr.11: 66592729-66592729 on Build GRCh38
rs756825207 Chr.11: 66592899-66592900 on Build GRCh38
rs745995805 Chr.11: 66592529-66592529 on Build GRCh38
rs765436279 Chr.11: 66592850-66592850 on Build GRCh38
rs753258358 Chr.11: 66592568-66592568 on Build GRCh38
rs550209025 Chr.11: 66592699-66592699 on Build GRCh38
rs761403969 Chr.11: 66592814-66592814 on Build GRCh38
rs759859325 Chr.11: 66592832-66592832 on Build GRCh38
rs781127005 Chr.11: 66592882-66592882 on Build GRCh38
rs546067186 Chr.11: 66592586-66592586 on Build GRCh38
rs746827364 Chr.11: 66592622-66592622 on Build GRCh38
rs768068768 Chr.11: 66592806-66592806 on Build GRCh38
rs753840413 Chr.11: 66592517-66592517 on Build GRCh38
rs969880517 Chr.11: 66592540-66592540 on Build GRCh38
rs756138526 Chr.11: 66592957-66592957 on Build GRCh38
rs530433678 Chr.11: 66592678-66592678 on Build GRCh38
rs756349522 Chr.11: 66592660-66592660 on Build GRCh38
rs759785775 Chr.11: 66592943-66592943 on Build GRCh38
rs138577027 Chr.11: 66592609-66592609 on Build GRCh38
rs761185674 Chr.11: 66592924-66592924 on Build GRCh38
rs752066055 Chr.11: 66592644-66592644 on Build GRCh38
rs368797364 Chr.11: 66592802-66592802 on Build GRCh38
rs764584208 Chr.11: 66592559-66592559 on Build GRCh38
rs745696193 Chr.11: 66592783-66592783 on Build GRCh38
rs750338539 Chr.11: 66592887-66592887 on Build GRCh38
rs757731836 Chr.11: 66592648-66592648 on Build GRCh38
rs750345118 Chr.11: 66592954-66592954 on Build GRCh38
rs769369017 Chr.11: 66592700-66592700 on Build GRCh38
rs763055203 Chr.11: 66592948-66592948 on Build GRCh38
rs1026780711 Chr.11: 66592752-66592752 on Build GRCh38
rs781380374 Chr.11: 66592797-66592803 on Build GRCh38
rs774063829 Chr.11: 66592811-66592811 on Build GRCh38
rs758790803 Chr.11: 66592503-66592503 on Build GRCh38
rs767930731 Chr.11: 66592652-66592652 on Build GRCh38
rs764655265 Chr.11: 66592584-66592584 on Build GRCh38
rs199562889 Chr.11: 66592635-66592635 on Build GRCh38
rs779606742 Chr.11: 66592799-66592799 on Build GRCh38
rs865913084 Chr.11: 66592930-66592930 on Build GRCh38
rs778678850 Chr.11: 66592899-66592899 on Build GRCh38
rs907495341 Chr.11: 66592714-66592714 on Build GRCh38
rs1002857887 Chr.11: 66592715-66592715 on Build GRCh38
rs367570263 Chr.11: 66592740-66592740 on Build GRCh38
rs781073881 Chr.11: 66592782-66592782 on Build GRCh38
rs758883041 Chr.11: 66592579-66592579 on Build GRCh38
rs912465560 Chr.11: 66592810-66592810 on Build GRCh38
rs758279543 Chr.11: 66592793-66592794 on Build GRCh38
rs915687484 Chr.11: 66592506-66592506 on Build GRCh38
rs150149433 Chr.11: 66592558-66592558 on Build GRCh38
rs753437521 Chr.11: 66592553-66592553 on Build GRCh38
rs893310467 Chr.11: 66592650-66592650 on Build GRCh38
rs950560733 Chr.11: 66592904-66592904 on Build GRCh38
rs772748851 Chr.11: 66592815-66592815 on Build GRCh38
rs773858158 Chr.11: 66592912-66592912 on Build GRCh38
rs1022679432 Chr.11: 66592827-66592827 on Build GRCh38
rs780799376 Chr.11: 66592900-66592900 on Build GRCh38
rs878919764 Chr.11: 66592667-66592667 on Build GRCh38
rs920939775 Chr.11: 66592824-66592824 on Build GRCh38
rs1053955601 Chr.11: 66592848-66592848 on Build GRCh38
rs1041915209 Chr.11: 66592870-66592870 on Build GRCh38
rs142353912 Chr.11: 66592825-66592825 on Build GRCh38
rs374478878 Chr.11: 66592595-66592595 on Build GRCh38
rs138087514 Chr.11: 66592820-66592820 on Build GRCh38
rs200905415 Chr.11: 66592944-66592944 on Build GRCh38
rs749023439 Chr.11: 66592892-66592892 on Build GRCh38
rs765489712 Chr.11: 66592947-66592947 on Build GRCh38
rs546096509 Chr.11: 66592627-66592627 on Build GRCh38
rs779845664 Chr.11: 66592890-66592890 on Build GRCh38
rs746882555 Chr.11: 66592683-66592683 on Build GRCh38
rs200871346 Chr.11: 66592900-66592900 on Build GRCh38
rs775189445 Chr.11: 66592703-66592703 on Build GRCh38
rs756268632 Chr.11: 66592608-66592608 on Build GRCh38
rs773177875 Chr.11: 66592621-66592621 on Build GRCh38
rs145549774 Chr.11: 66592657-66592657 on Build GRCh38
rs761734856 Chr.11: 66592905-66592905 on Build GRCh38
rs1003140973 Chr.11: 66592929-66592929 on Build GRCh38
rs758447209 Chr.11: 66592872-66592872 on Build GRCh38
rs762491237 Chr.11: 66592640-66592640 on Build GRCh38
rs765262078 Chr.11: 66592630-66592632 on Build GRCh38
rs757248706 Chr.11: 66592781-66592781 on Build GRCh38
rs575497542 Chr.11: 66592587-66592587 on Build GRCh38
rs747383629 Chr.11: 66592508-66592508 on Build GRCh38
rs143855952 Chr.11: 66592851-66592851 on Build GRCh38
rs946205685 Chr.11: 66592853-66592853 on Build GRCh38
rs775006700 Chr.11: 66592731-66592731 on Build GRCh38
rs764561779 Chr.11: 66592643-66592643 on Build GRCh38
rs755870174 Chr.11: 66592889-66592889 on Build GRCh38
rs752613597 Chr.11: 66592716-66592719 on Build GRCh38
rs71045948 Chr.11: 66592888-66592889 on Build GRCh38
rs771914166 Chr.11: 66592539-66592539 on Build GRCh38
rs951298348 Chr.11: 66592786-66592786 on Build GRCh38
rs761071119 Chr.11: 66592762-66592762 on Build GRCh38
rs772721340 Chr.11: 66592935-66592935 on Build GRCh38
rs781536350 Chr.11: 66592523-66592523 on Build GRCh38
rs771342455 Chr.11: 66592515-66592515 on Build GRCh38
rs769585190 Chr.11: 66592797-66592797 on Build GRCh38
rs755017754 Chr.11: 66592778-66592778 on Build GRCh38
rs564852028 Chr.11: 66592745-66592745 on Build GRCh38
rs764097663 Chr.11: 66592953-66592953 on Build GRCh38
rs777116559 Chr.11: 66592941-66592941 on Build GRCh38
rs1010455946 Chr.11: 66592658-66592658 on Build GRCh38
rs771927580 Chr.11: 66592926-66592926 on Build GRCh38
rs762394264 Chr.11: 66592727-66592727 on Build GRCh38
rs201087681 Chr.11: 66592599-66592599 on Build GRCh38

View Full Product Details