Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67365265-67365772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCF1
Entrez Gene ID
23529
Gene Name
cardiotrophin-like cytokine factor 1
Gene Aliases
BSF-3, BSF3, CISS2, CLC, NNT-1, NNT1, NR6
UniGene
Hs.502977
Gene Chromosome Location
Chr.11: 67364168-67374177 on Build GRCh38
Transcripts
NM_001166212.1 NM_013246.2 NR_024469.1

Gene Symbol
LOC100130987
Entrez Gene ID
100130987
Gene Name
uncharacterized LOC100130987
Gene Aliases
-
UniGene
Hs.664585
Gene Chromosome Location
Chr.11: 67317839-67391687 on Build GRCh38
Transcripts
NM_001166212.1 NM_013246.2 NR_024469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5777345 c.313C>T p.Q105* Chr.11: 67365501-67365501 on Build GRCh38
1475807 c.322G>A p.E108K Chr.11: 67365492-67365492 on Build GRCh38
5734470 c.377C>T p.T126I Chr.11: 67365437-67365437 on Build GRCh38
1605108 c.364C>G p.R122G Chr.11: 67365450-67365450 on Build GRCh38
1605107 c.523C>A p.H175N Chr.11: 67365291-67365291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758521384 Chr.11: 67365664-67365664 on Build GRCh38
rs376016004 Chr.11: 67365327-67365327 on Build GRCh38
rs778794209 Chr.11: 67365543-67365543 on Build GRCh38
rs747180318 Chr.11: 67365674-67365674 on Build GRCh38
rs78755659 Chr.11: 67365333-67365333 on Build GRCh38
rs563402891 Chr.11: 67365521-67365521 on Build GRCh38
rs368762582 Chr.11: 67365462-67365462 on Build GRCh38
rs528341431 Chr.11: 67365603-67365603 on Build GRCh38
rs373702399 Chr.11: 67365673-67365673 on Build GRCh38
rs781002061 Chr.11: 67365530-67365530 on Build GRCh38
rs544450000 Chr.11: 67365615-67365615 on Build GRCh38
rs770150295 Chr.11: 67365681-67365693 on Build GRCh38
rs935059903 Chr.11: 67365719-67365719 on Build GRCh38
rs762129649 Chr.11: 67365367-67365367 on Build GRCh38
rs778526790 Chr.11: 67365420-67365420 on Build GRCh38
rs34057094 Chr.11: 67365422-67365422 on Build GRCh38
rs752250486 Chr.11: 67365421-67365421 on Build GRCh38
rs562640101 Chr.11: 67365655-67365655 on Build GRCh38
rs754365534 Chr.11: 67365463-67365463 on Build GRCh38
rs567374288 Chr.11: 67365582-67365582 on Build GRCh38
rs373018736 Chr.11: 67365366-67365366 on Build GRCh38
rs144403804 Chr.11: 67365330-67365330 on Build GRCh38
rs754424165 Chr.11: 67365410-67365410 on Build GRCh38
rs773547381 Chr.11: 67365566-67365566 on Build GRCh38
rs151050272 Chr.11: 67365449-67365449 on Build GRCh38
rs370638359 Chr.11: 67365662-67365662 on Build GRCh38
rs375238966 Chr.11: 67365381-67365381 on Build GRCh38
rs923612988 Chr.11: 67365689-67365689 on Build GRCh38
rs371858892 Chr.11: 67365452-67365452 on Build GRCh38
rs140841317 Chr.11: 67365456-67365456 on Build GRCh38
rs979639960 Chr.11: 67365555-67365555 on Build GRCh38
rs147960206 Chr.11: 67365370-67365370 on Build GRCh38
rs765776881 Chr.11: 67365588-67365588 on Build GRCh38
rs772337610 Chr.11: 67365565-67365565 on Build GRCh38
rs1014885737 Chr.11: 67365404-67365404 on Build GRCh38
rs746809332 Chr.11: 67365425-67365425 on Build GRCh38
rs761340220 Chr.11: 67365292-67365292 on Build GRCh38
rs764602656 Chr.11: 67365492-67365492 on Build GRCh38
rs766990125 Chr.11: 67365380-67365380 on Build GRCh38
rs746249138 Chr.11: 67365677-67365679 on Build GRCh38
rs777397541 Chr.11: 67365517-67365517 on Build GRCh38
rs140991132 Chr.11: 67365332-67365332 on Build GRCh38
rs994226554 Chr.11: 67365730-67365730 on Build GRCh38
rs149603003 Chr.11: 67365314-67365314 on Build GRCh38
rs1001719096 Chr.11: 67365630-67365630 on Build GRCh38
rs760120872 Chr.11: 67365465-67365465 on Build GRCh38
rs898598975 Chr.11: 67365717-67365717 on Build GRCh38
rs748257910 Chr.11: 67365544-67365544 on Build GRCh38
rs573454925 Chr.11: 67365331-67365331 on Build GRCh38
rs142875118 Chr.11: 67365426-67365426 on Build GRCh38
rs571607608 Chr.11: 67365735-67365735 on Build GRCh38
rs761887384 Chr.11: 67365450-67365450 on Build GRCh38
rs768834730 Chr.11: 67365360-67365360 on Build GRCh38
rs761109106 Chr.11: 67365461-67365461 on Build GRCh38
rs983554417 Chr.11: 67365515-67365515 on Build GRCh38
rs756930235 Chr.11: 67365609-67365609 on Build GRCh38
rs750832975 Chr.11: 67365371-67365371 on Build GRCh38
rs546587564 Chr.11: 67365727-67365727 on Build GRCh38
rs918569099 Chr.11: 67365509-67365509 on Build GRCh38
rs767245313 Chr.11: 67365616-67365616 on Build GRCh38
rs145660153 Chr.11: 67365538-67365538 on Build GRCh38
rs1043394931 Chr.11: 67365322-67365322 on Build GRCh38
rs145294376 Chr.11: 67365581-67365581 on Build GRCh38
rs140383361 Chr.11: 67365573-67365573 on Build GRCh38
rs763640141 Chr.11: 67365604-67365604 on Build GRCh38
rs104894198 Chr.11: 67365493-67365493 on Build GRCh38
rs768583993 Chr.11: 67365444-67365444 on Build GRCh38
rs761200783 Chr.11: 67365375-67365375 on Build GRCh38
rs200464690 Chr.11: 67365644-67365644 on Build GRCh38
rs377140946 Chr.11: 67365646-67365646 on Build GRCh38
rs559258400 Chr.11: 67365350-67365350 on Build GRCh38
rs775926530 Chr.11: 67365589-67365589 on Build GRCh38
rs150131356 Chr.11: 67365523-67365523 on Build GRCh38
rs140020543 Chr.11: 67365423-67365423 on Build GRCh38
rs1024456074 Chr.11: 67365414-67365414 on Build GRCh38
rs745851012 Chr.11: 67365427-67365427 on Build GRCh38
rs757920303 Chr.11: 67365499-67365499 on Build GRCh38
rs942538789 Chr.11: 67365540-67365540 on Build GRCh38
rs754604226 Chr.11: 67365305-67365305 on Build GRCh38
rs747274369 Chr.11: 67365572-67365572 on Build GRCh38
rs946398553 Chr.11: 67365301-67365301 on Build GRCh38
rs367649677 Chr.11: 67365535-67365535 on Build GRCh38
rs369389500 Chr.11: 67365358-67365358 on Build GRCh38
rs754455483 Chr.11: 67365385-67365385 on Build GRCh38
rs1056155201 Chr.11: 67365748-67365748 on Build GRCh38
rs747883491 Chr.11: 67365319-67365319 on Build GRCh38

View Full Product Details