Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 76495959-76496481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMSY
Entrez Gene ID
56946
Gene Name
EMSY, BRCA2 interacting transcriptional repressor
Gene Aliases
C11orf30, GL002
UniGene
Hs.352588
Gene Chromosome Location
Chr.11: 76444928-76552901 on Build GRCh38
Transcripts
NM_001300942.1 NM_001300943.1 NM_001300944.1 NM_020193.4 XM_005274112.4 XM_006718637.3 XM_006718638.3 XM_006718639.3 XM_011545173.2 XM_011545174.2 XM_011545175.2 XM_011545177.2 XM_011545178.2 XM_011545179.2 XM_017018021.1 XM_017018022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
213880 c.1316C>G p.P439R Chr.11: 76496422-76496422 on Build GRCh38
69872 c.1243G>C p.V415L Chr.11: 76496349-76496349 on Build GRCh38
6583784 c.1291C>G p.H431D Chr.11: 76496397-76496397 on Build GRCh38
6587331 c.1281T>A p.S427S Chr.11: 76496387-76496387 on Build GRCh38
318985 c.1229A>G p.Q410R Chr.11: 76496335-76496335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61754147 Chr.11: 76496262-76496262 on Build GRCh38
rs944471263 Chr.11: 76496108-76496108 on Build GRCh38
rs767567909 Chr.11: 76496330-76496330 on Build GRCh38
rs985984124 Chr.11: 76496102-76496102 on Build GRCh38
rs768801916 Chr.11: 76496388-76496388 on Build GRCh38
rs760577895 Chr.11: 76496415-76496415 on Build GRCh38
rs386755151 Chr.11: 76496062-76496076 on Build GRCh38
rs920488918 Chr.11: 76496095-76496095 on Build GRCh38
rs554291684 Chr.11: 76496052-76496052 on Build GRCh38
rs891875160 Chr.11: 76496126-76496126 on Build GRCh38
rs936208509 Chr.11: 76496389-76496389 on Build GRCh38
rs774758872 Chr.11: 76496198-76496198 on Build GRCh38
rs372110730 Chr.11: 76496403-76496403 on Build GRCh38
rs902851188 Chr.11: 76496145-76496145 on Build GRCh38
rs926082669 Chr.11: 76496050-76496050 on Build GRCh38
rs764809048 Chr.11: 76496431-76496431 on Build GRCh38
rs149291840 Chr.11: 76496398-76496398 on Build GRCh38
rs758931507 Chr.11: 76496422-76496422 on Build GRCh38
rs188547925 Chr.11: 76496321-76496321 on Build GRCh38
rs1053172630 Chr.11: 76496120-76496120 on Build GRCh38
rs760208526 Chr.11: 76496316-76496318 on Build GRCh38
rs367574485 Chr.11: 76496325-76496325 on Build GRCh38
rs574452899 Chr.11: 76496060-76496060 on Build GRCh38
rs767290112 Chr.11: 76496409-76496409 on Build GRCh38
rs143568764 Chr.11: 76496421-76496421 on Build GRCh38
rs1017445873 Chr.11: 76496161-76496161 on Build GRCh38
rs879050577 Chr.11: 76496202-76496202 on Build GRCh38
rs769936852 Chr.11: 76496211-76496211 on Build GRCh38
rs756104983 Chr.11: 76496307-76496307 on Build GRCh38
rs775606975 Chr.11: 76496385-76496385 on Build GRCh38
rs994669008 Chr.11: 76496179-76496179 on Build GRCh38
rs185214824 Chr.11: 76496153-76496153 on Build GRCh38
rs1012435812 Chr.11: 76496203-76496203 on Build GRCh38
rs763229879 Chr.11: 76496219-76496219 on Build GRCh38
rs113875958 Chr.11: 76496209-76496209 on Build GRCh38
rs776298786 Chr.11: 76496040-76496040 on Build GRCh38
rs369442765 Chr.11: 76496062-76496062 on Build GRCh38
rs750290621 Chr.11: 76496303-76496303 on Build GRCh38
rs948962290 Chr.11: 76496030-76496030 on Build GRCh38
rs750356838 Chr.11: 76496196-76496196 on Build GRCh38
rs368860952 Chr.11: 76496447-76496447 on Build GRCh38
rs764261455 Chr.11: 76496248-76496248 on Build GRCh38
rs962323266 Chr.11: 76496057-76496057 on Build GRCh38
rs917532942 Chr.11: 76496017-76496017 on Build GRCh38
rs761544856 Chr.11: 76496263-76496263 on Build GRCh38
rs757629476 Chr.11: 76496356-76496356 on Build GRCh38
rs771446578 Chr.11: 76496180-76496180 on Build GRCh38
rs74680029 Chr.11: 76496155-76496155 on Build GRCh38
rs911722952 Chr.11: 76496106-76496106 on Build GRCh38
rs375127434 Chr.11: 76496387-76496387 on Build GRCh38
rs201002831 Chr.11: 76496375-76496375 on Build GRCh38
rs563055892 Chr.11: 76496194-76496194 on Build GRCh38
rs963504948 Chr.11: 76496171-76496171 on Build GRCh38
rs775490904 Chr.11: 76496216-76496216 on Build GRCh38
rs767326969 Chr.11: 76496264-76496264 on Build GRCh38
rs781700827 Chr.11: 76496372-76496372 on Build GRCh38
rs193022338 Chr.11: 76496151-76496151 on Build GRCh38
rs962089538 Chr.11: 76496350-76496350 on Build GRCh38
rs188375960 Chr.11: 76496142-76496142 on Build GRCh38
rs768366372 Chr.11: 76496031-76496031 on Build GRCh38
rs773061957 Chr.11: 76496410-76496410 on Build GRCh38
rs766280069 Chr.11: 76496417-76496417 on Build GRCh38
rs531713649 Chr.11: 76496229-76496229 on Build GRCh38
rs778381474 Chr.11: 76496343-76496343 on Build GRCh38
rs548633654 Chr.11: 76496230-76496230 on Build GRCh38
rs970810285 Chr.11: 76496028-76496028 on Build GRCh38
rs138783195 Chr.11: 76496381-76496381 on Build GRCh38
rs931817393 Chr.11: 76496096-76496096 on Build GRCh38
rs144872506 Chr.11: 76496351-76496351 on Build GRCh38
rs761631355 Chr.11: 76496041-76496041 on Build GRCh38

View Full Product Details