Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14437731-14438284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF7IP
Entrez Gene ID
55729
Gene Name
activating transcription factor 7 interacting protein
Gene Aliases
AM, ATF-IP, MCAF, MCAF1, p621
UniGene
Hs.504856
Gene Chromosome Location
Chr.12: 14365632-14502935 on Build GRCh38
Transcripts
NM_001286514.1 NM_001286515.1 NM_018179.4 NM_181352.1 XM_005253424.3 XM_006719108.3 XM_006719109.3 XM_011520754.2 XM_011520755.2 XM_011520756.2 XM_017019638.1 XM_017019639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2002841 c.1827G>A p.A609A Chr.12: 14438165-14438165 on Build GRCh38
6382209 c.1828C>A p.Q610K Chr.12: 14438169-14438169 on Build GRCh38
5006527 c.1870C>T p.R624* Chr.12: 14438211-14438211 on Build GRCh38
6505253 c.1823G>A p.C608Y Chr.12: 14438161-14438161 on Build GRCh38
2002844 c.1915C>T p.L639F Chr.12: 14438253-14438253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111474484 Chr.12: 14437978-14437978 on Build GRCh38
rs10734876 Chr.12: 14438047-14438047 on Build GRCh38
rs569389255 Chr.12: 14437829-14437829 on Build GRCh38
rs896685081 Chr.12: 14438009-14438009 on Build GRCh38
rs972487959 Chr.12: 14437782-14437782 on Build GRCh38
rs1001819933 Chr.12: 14437861-14437861 on Build GRCh38
rs1002346233 Chr.12: 14437871-14437871 on Build GRCh38
rs761269893 Chr.12: 14438088-14438088 on Build GRCh38
rs368961957 Chr.12: 14438084-14438084 on Build GRCh38
rs373856781 Chr.12: 14438024-14438024 on Build GRCh38
rs992648585 Chr.12: 14437907-14437907 on Build GRCh38
rs767448478 Chr.12: 14438102-14438102 on Build GRCh38
rs994037707 Chr.12: 14438021-14438021 on Build GRCh38
rs534654020 Chr.12: 14437897-14437897 on Build GRCh38
rs955244789 Chr.12: 14438060-14438060 on Build GRCh38
rs946408491 Chr.12: 14437980-14437980 on Build GRCh38
rs911460810 Chr.12: 14438001-14438001 on Build GRCh38
rs747534018 Chr.12: 14438190-14438190 on Build GRCh38
rs922524865 Chr.12: 14437825-14437825 on Build GRCh38
rs952087291 Chr.12: 14437916-14437916 on Build GRCh38
rs539200385 Chr.12: 14437814-14437814 on Build GRCh38
rs760402413 Chr.12: 14438120-14438120 on Build GRCh38
rs896635196 Chr.12: 14437909-14437909 on Build GRCh38
rs1042733719 Chr.12: 14438070-14438070 on Build GRCh38
rs76525918 Chr.12: 14438061-14438061 on Build GRCh38
rs575017520 Chr.12: 14437988-14437988 on Build GRCh38
rs955383055 Chr.12: 14437969-14437969 on Build GRCh38
rs138145099 Chr.12: 14438157-14438157 on Build GRCh38
rs532661084 Chr.12: 14438209-14438209 on Build GRCh38
rs577793143 Chr.12: 14437921-14437921 on Build GRCh38
rs960965821 Chr.12: 14437780-14437780 on Build GRCh38
rs918030737 Chr.12: 14437931-14437931 on Build GRCh38
rs1041170552 Chr.12: 14438046-14438046 on Build GRCh38
rs765421547 Chr.12: 14438162-14438162 on Build GRCh38
rs1002296920 Chr.12: 14437763-14437763 on Build GRCh38
rs1035238212 Chr.12: 14437779-14437779 on Build GRCh38
rs370459735 Chr.12: 14437870-14437870 on Build GRCh38
rs984925978 Chr.12: 14437791-14437791 on Build GRCh38
rs571793721 Chr.12: 14437806-14437806 on Build GRCh38
rs1006186718 Chr.12: 14437920-14437920 on Build GRCh38
rs913608088 Chr.12: 14437846-14437846 on Build GRCh38
rs937669150 Chr.12: 14438257-14438257 on Build GRCh38
rs1043350819 Chr.12: 14437989-14437989 on Build GRCh38
rs937791774 Chr.12: 14437994-14437994 on Build GRCh38
rs922483404 Chr.12: 14437945-14437945 on Build GRCh38
rs112640443 Chr.12: 14437880-14437880 on Build GRCh38
rs193097616 Chr.12: 14437926-14437926 on Build GRCh38
rs1010365489 Chr.12: 14438052-14438053 on Build GRCh38
rs573148380 Chr.12: 14437869-14437869 on Build GRCh38
rs61754406 Chr.12: 14438165-14438165 on Build GRCh38
rs201415676 Chr.12: 14438130-14438130 on Build GRCh38
rs373169074 Chr.12: 14438171-14438171 on Build GRCh38
rs988111538 Chr.12: 14437970-14437970 on Build GRCh38
rs544790448 Chr.12: 14437977-14437977 on Build GRCh38
rs547233281 Chr.12: 14437803-14437803 on Build GRCh38
rs1050463953 Chr.12: 14437996-14437996 on Build GRCh38
rs1006237310 Chr.12: 14438035-14438035 on Build GRCh38
rs555003545 Chr.12: 14437850-14437850 on Build GRCh38
rs1021678442 Chr.12: 14437805-14437805 on Build GRCh38
rs961431751 Chr.12: 14437942-14437942 on Build GRCh38
rs536714256 Chr.12: 14437835-14437835 on Build GRCh38
rs377610222 Chr.12: 14438240-14438240 on Build GRCh38
rs766164869 Chr.12: 14438127-14438127 on Build GRCh38
rs959898030 Chr.12: 14437899-14437899 on Build GRCh38
rs1017645441 Chr.12: 14438039-14438039 on Build GRCh38
rs1056124994 Chr.12: 14438002-14438002 on Build GRCh38
rs963872506 Chr.12: 14437815-14437815 on Build GRCh38
rs528086064 Chr.12: 14438048-14438048 on Build GRCh38
rs565557284 Chr.12: 14438145-14438145 on Build GRCh38
rs189256507 Chr.12: 14437910-14437910 on Build GRCh38
rs952137478 Chr.12: 14437786-14437786 on Build GRCh38
rs551216061 Chr.12: 14438250-14438250 on Build GRCh38
rs910605126 Chr.12: 14437812-14437812 on Build GRCh38
rs762700810 Chr.12: 14438080-14438080 on Build GRCh38
rs111354886 Chr.12: 14437929-14437929 on Build GRCh38
rs752794706 Chr.12: 14438164-14438164 on Build GRCh38
rs931326976 Chr.12: 14437976-14437976 on Build GRCh38
rs888003538 Chr.12: 14438032-14438032 on Build GRCh38
rs555713169 Chr.12: 14437783-14437783 on Build GRCh38
rs1026974391 Chr.12: 14437913-14437913 on Build GRCh38
rs964687518 Chr.12: 14438043-14438043 on Build GRCh38
rs547004592 Chr.12: 14438052-14438052 on Build GRCh38
rs369993625 Chr.12: 14438083-14438083 on Build GRCh38
rs926388969 Chr.12: 14437992-14437992 on Build GRCh38
rs551129696 Chr.12: 14437823-14437823 on Build GRCh38
rs572575234 Chr.12: 14437881-14437881 on Build GRCh38

View Full Product Details