Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 72021031-72021624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHODH
Entrez Gene ID
1723
Gene Name
dihydroorotate dehydrogenase (quinone)
Gene Aliases
DHOdehase, POADS, URA1
UniGene
Hs.654427
Gene Chromosome Location
Chr.16: 72008744-72027659 on Build GRCh38
Transcripts
NM_001361.4 XM_005255827.3 XM_005255829.3 XM_017022990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6643284 c.606delC p.L204fs*7 Chr.16: 72021212-72021212 on Build GRCh38
1379667 c.595C>T p.R199C Chr.16: 72021201-72021201 on Build GRCh38
3511942 c.643C>T p.P215S Chr.16: 72021249-72021249 on Build GRCh38
3511943 c.674G>A p.G225E Chr.16: 72021280-72021280 on Build GRCh38
6030751 c.651T>C p.T217T Chr.16: 72021257-72021257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369556950 Chr.16: 72021261-72021261 on Build GRCh38
rs374029102 Chr.16: 72021096-72021096 on Build GRCh38
rs573177564 Chr.16: 72021212-72021212 on Build GRCh38
rs779876255 Chr.16: 72021325-72021325 on Build GRCh38
rs747045289 Chr.16: 72021271-72021271 on Build GRCh38
rs754445166 Chr.16: 72021151-72021151 on Build GRCh38
rs764101696 Chr.16: 72021171-72021171 on Build GRCh38
rs750079059 Chr.16: 72021316-72021316 on Build GRCh38
rs1040178398 Chr.16: 72021385-72021385 on Build GRCh38
rs998696002 Chr.16: 72021394-72021394 on Build GRCh38
rs568481099 Chr.16: 72021549-72021549 on Build GRCh38
rs545513947 Chr.16: 72021302-72021302 on Build GRCh38
rs559542296 Chr.16: 72021055-72021055 on Build GRCh38
rs776620021 Chr.16: 72021093-72021093 on Build GRCh38
rs757910086 Chr.16: 72021210-72021210 on Build GRCh38
rs533490624 Chr.16: 72021074-72021074 on Build GRCh38
rs753231997 Chr.16: 72021209-72021209 on Build GRCh38
rs563533148 Chr.16: 72021335-72021335 on Build GRCh38
rs765734052 Chr.16: 72021169-72021169 on Build GRCh38
rs745617898 Chr.16: 72021183-72021183 on Build GRCh38
rs761246691 Chr.16: 72021298-72021298 on Build GRCh38
rs755461287 Chr.16: 72021159-72021159 on Build GRCh38
rs771282792 Chr.16: 72021283-72021283 on Build GRCh38
rs567514273 Chr.16: 72021111-72021111 on Build GRCh38
rs905556929 Chr.16: 72021254-72021254 on Build GRCh38
rs887418298 Chr.16: 72021541-72021541 on Build GRCh38
rs1036057845 Chr.16: 72021589-72021589 on Build GRCh38
rs778020343 Chr.16: 72021269-72021269 on Build GRCh38
rs771117354 Chr.16: 72021341-72021341 on Build GRCh38
rs528598713 Chr.16: 72021272-72021272 on Build GRCh38
rs56387295 Chr.16: 72021437-72021437 on Build GRCh38
rs571843761 Chr.16: 72021497-72021497 on Build GRCh38
rs754997733 Chr.16: 72021331-72021331 on Build GRCh38
rs773510526 Chr.16: 72021131-72021131 on Build GRCh38
rs149123373 Chr.16: 72021176-72021176 on Build GRCh38
rs777166937 Chr.16: 72021202-72021202 on Build GRCh38
rs772180035 Chr.16: 72021197-72021197 on Build GRCh38
rs777478366 Chr.16: 72021188-72021188 on Build GRCh38
rs763498232 Chr.16: 72021295-72021295 on Build GRCh38
rs377062884 Chr.16: 72021078-72021078 on Build GRCh38
rs201970636 Chr.16: 72021189-72021189 on Build GRCh38
rs774434640 Chr.16: 72021484-72021484 on Build GRCh38
rs746068212 Chr.16: 72021346-72021346 on Build GRCh38
rs779915805 Chr.16: 72021260-72021260 on Build GRCh38
rs538331493 Chr.16: 72021559-72021559 on Build GRCh38
rs200890010 Chr.16: 72021149-72021149 on Build GRCh38
rs1049322761 Chr.16: 72021267-72021267 on Build GRCh38
rs375041156 Chr.16: 72021294-72021294 on Build GRCh38
rs376933922 Chr.16: 72021337-72021337 on Build GRCh38
rs1045984044 Chr.16: 72021240-72021240 on Build GRCh38
rs779945012 Chr.16: 72021500-72021500 on Build GRCh38
rs192172987 Chr.16: 72021342-72021342 on Build GRCh38
rs988868914 Chr.16: 72021392-72021392 on Build GRCh38
rs373069655 Chr.16: 72021075-72021075 on Build GRCh38
rs1001703809 Chr.16: 72021098-72021098 on Build GRCh38
rs201202896 Chr.16: 72021288-72021288 on Build GRCh38
rs372858879 Chr.16: 72021203-72021203 on Build GRCh38
rs930191691 Chr.16: 72021338-72021338 on Build GRCh38
rs770695096 Chr.16: 72021092-72021092 on Build GRCh38
rs371527785 Chr.16: 72021178-72021178 on Build GRCh38
rs368919851 Chr.16: 72021345-72021345 on Build GRCh38
rs758014545 Chr.16: 72021079-72021079 on Build GRCh38
rs546842803 Chr.16: 72021479-72021479 on Build GRCh38
rs753525797 Chr.16: 72021330-72021330 on Build GRCh38
rs951090523 Chr.16: 72021568-72021568 on Build GRCh38
rs554943001 Chr.16: 72021204-72021204 on Build GRCh38
rs1046036497 Chr.16: 72021476-72021476 on Build GRCh38
rs746657681 Chr.16: 72021177-72021177 on Build GRCh38
rs755880891 Chr.16: 72021248-72021248 on Build GRCh38
rs954515669 Chr.16: 72021152-72021152 on Build GRCh38
rs1053297105 Chr.16: 72021211-72021212 on Build GRCh38
rs1023906139 Chr.16: 72021496-72021496 on Build GRCh38
rs983299629 Chr.16: 72021147-72021147 on Build GRCh38
rs776932166 Chr.16: 72021287-72021287 on Build GRCh38
rs374360785 Chr.16: 72021088-72021088 on Build GRCh38
rs913413537 Chr.16: 72021423-72021423 on Build GRCh38
rs565105946 Chr.16: 72021268-72021268 on Build GRCh38
rs1030965375 Chr.16: 72021447-72021447 on Build GRCh38
rs578023727 Chr.16: 72021296-72021296 on Build GRCh38
rs377694783 Chr.16: 72021363-72021363 on Build GRCh38
rs761180542 Chr.16: 72021140-72021140 on Build GRCh38
rs770187150 Chr.16: 72021291-72021291 on Build GRCh38
rs748033418 Chr.16: 72021112-72021112 on Build GRCh38
rs1048727648 Chr.16: 72021536-72021536 on Build GRCh38
rs182981376 Chr.16: 72021434-72021434 on Build GRCh38
rs748193279 Chr.16: 72021190-72021190 on Build GRCh38
rs975061893 Chr.16: 72021143-72021143 on Build GRCh38
rs1862752 Chr.16: 72021085-72021085 on Build GRCh38
rs892046171 Chr.16: 72021087-72021087 on Build GRCh38
rs754391821 Chr.16: 72021073-72021073 on Build GRCh38
rs750190054 Chr.16: 72021230-72021230 on Build GRCh38
rs780785810 Chr.16: 72021222-72021222 on Build GRCh38
rs765958693 Chr.16: 72021068-72021068 on Build GRCh38
rs267606769 Chr.16: 72021201-72021201 on Build GRCh38
rs899886753 Chr.16: 72021390-72021390 on Build GRCh38
rs774320307 Chr.16: 72021118-72021118 on Build GRCh38
rs372694919 Chr.16: 72021297-72021297 on Build GRCh38
rs540401962 Chr.16: 72021242-72021242 on Build GRCh38
rs148523165 Chr.16: 72021179-72021179 on Build GRCh38
rs375309794 Chr.16: 72021221-72021221 on Build GRCh38
rs1018657894 Chr.16: 72021129-72021129 on Build GRCh38
rs1018399335 Chr.16: 72021213-72021213 on Build GRCh38
rs1030091045 Chr.16: 72021244-72021244 on Build GRCh38
rs559262199 Chr.16: 72021395-72021395 on Build GRCh38
rs546416162 Chr.16: 72021174-72021174 on Build GRCh38
rs752149904 Chr.16: 72021173-72021173 on Build GRCh38
rs747094553 Chr.16: 72021340-72021340 on Build GRCh38
rs267606767 Chr.16: 72021211-72021211 on Build GRCh38
rs548972726 Chr.16: 72021419-72021419 on Build GRCh38
rs536547425 Chr.16: 72021200-72021200 on Build GRCh38
rs201244734 Chr.16: 72021125-72021125 on Build GRCh38
rs201023112 Chr.16: 72021113-72021113 on Build GRCh38
rs200598938 Chr.16: 72021198-72021198 on Build GRCh38
rs982977806 Chr.16: 72021599-72021599 on Build GRCh38
rs969448364 Chr.16: 72021305-72021305 on Build GRCh38
rs763689021 Chr.16: 72021325-72021325 on Build GRCh38
rs969124332 Chr.16: 72021548-72021548 on Build GRCh38
rs755930158 Chr.16: 72021317-72021317 on Build GRCh38

View Full Product Details