Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89191989-89192531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH15
Entrez Gene ID
1013
Gene Name
cadherin 15
Gene Aliases
CDH14, CDH3, CDHM, MCAD, MRD3
UniGene
Hs.148090
Gene Chromosome Location
Chr.16: 89171755-89195492 on Build GRCh38
Transcripts
NM_004933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6380211 c.1742G>A p.R581H Chr.16: 89192331-89192331 on Build GRCh38
1735546 c.1769C>T p.P590L Chr.16: 89192358-89192358 on Build GRCh38
4129510 c.1750A>C p.K584Q Chr.16: 89192339-89192339 on Build GRCh38
6374753 c.1838G>A p.S613N Chr.16: 89192427-89192427 on Build GRCh38
6624698 c.1759G>A p.V587I Chr.16: 89192348-89192348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755077990 Chr.16: 89192285-89192285 on Build GRCh38
rs925923888 Chr.16: 89192134-89192134 on Build GRCh38
rs994511879 Chr.16: 89192274-89192274 on Build GRCh38
rs759475457 Chr.16: 89192429-89192429 on Build GRCh38
rs1045996091 Chr.16: 89192363-89192363 on Build GRCh38
rs760219012 Chr.16: 89192219-89192219 on Build GRCh38
rs762629906 Chr.16: 89192421-89192421 on Build GRCh38
rs187121295 Chr.16: 89192188-89192188 on Build GRCh38
rs1015045120 Chr.16: 89192027-89192027 on Build GRCh38
rs1011100089 Chr.16: 89192097-89192097 on Build GRCh38
rs374508070 Chr.16: 89192028-89192028 on Build GRCh38
rs756336374 Chr.16: 89192377-89192377 on Build GRCh38
rs953204418 Chr.16: 89192062-89192062 on Build GRCh38
rs774035764 Chr.16: 89192424-89192424 on Build GRCh38
rs1009199666 Chr.16: 89192420-89192420 on Build GRCh38
rs778153562 Chr.16: 89192378-89192378 on Build GRCh38
rs754263255 Chr.16: 89192291-89192291 on Build GRCh38
rs767583230 Chr.16: 89192362-89192362 on Build GRCh38
rs973292581 Chr.16: 89192032-89192032 on Build GRCh38
rs926049647 Chr.16: 89192336-89192336 on Build GRCh38
rs769928074 Chr.16: 89192313-89192313 on Build GRCh38
rs757941155 Chr.16: 89192442-89192445 on Build GRCh38
rs758568874 Chr.16: 89192505-89192505 on Build GRCh38
rs1041685743 Chr.16: 89192106-89192106 on Build GRCh38
rs773757986 Chr.16: 89192163-89192163 on Build GRCh38
rs1029097997 Chr.16: 89192414-89192414 on Build GRCh38
rs1040029831 Chr.16: 89192048-89192048 on Build GRCh38
rs976036463 Chr.16: 89192236-89192236 on Build GRCh38
rs991493951 Chr.16: 89192494-89192494 on Build GRCh38
rs895377367 Chr.16: 89192458-89192458 on Build GRCh38
rs369195898 Chr.16: 89192438-89192438 on Build GRCh38
rs774994552 Chr.16: 89192330-89192330 on Build GRCh38
rs921602077 Chr.16: 89192051-89192051 on Build GRCh38
rs765863927 Chr.16: 89192470-89192477 on Build GRCh38
rs950379575 Chr.16: 89192361-89192361 on Build GRCh38
rs776142745 Chr.16: 89192399-89192399 on Build GRCh38
rs75791347 Chr.16: 89192339-89192339 on Build GRCh38
rs935997767 Chr.16: 89192337-89192337 on Build GRCh38
rs939600678 Chr.16: 89192324-89192324 on Build GRCh38
rs754091466 Chr.16: 89192453-89192453 on Build GRCh38
rs540619908 Chr.16: 89192359-89192359 on Build GRCh38
rs902742819 Chr.16: 89192128-89192128 on Build GRCh38
rs576550717 Chr.16: 89192199-89192199 on Build GRCh38
rs555622852 Chr.16: 89192237-89192237 on Build GRCh38
rs771507650 Chr.16: 89192329-89192329 on Build GRCh38
rs150055194 Chr.16: 89192311-89192311 on Build GRCh38
rs764309530 Chr.16: 89192451-89192451 on Build GRCh38
rs757666255 Chr.16: 89192381-89192381 on Build GRCh38
rs867279497 Chr.16: 89192411-89192411 on Build GRCh38
rs776028872 Chr.16: 89192333-89192333 on Build GRCh38
rs375071693 Chr.16: 89192380-89192380 on Build GRCh38
rs994500514 Chr.16: 89192410-89192410 on Build GRCh38
rs983258883 Chr.16: 89192195-89192195 on Build GRCh38
rs368419611 Chr.16: 89192225-89192225 on Build GRCh38
rs749838702 Chr.16: 89192379-89192380 on Build GRCh38
rs948318008 Chr.16: 89192467-89192467 on Build GRCh38
rs951952748 Chr.16: 89192189-89192189 on Build GRCh38
rs556790462 Chr.16: 89192024-89192025 on Build GRCh38
rs893955184 Chr.16: 89192090-89192091 on Build GRCh38
rs779298552 Chr.16: 89192478-89192478 on Build GRCh38
rs965913983 Chr.16: 89192200-89192200 on Build GRCh38
rs780416394 Chr.16: 89192490-89192490 on Build GRCh38
rs767359125 Chr.16: 89192432-89192432 on Build GRCh38
rs751220504 Chr.16: 89192469-89192470 on Build GRCh38
rs1019221384 Chr.16: 89192320-89192320 on Build GRCh38
rs779984001 Chr.16: 89192269-89192269 on Build GRCh38
rs752749129 Chr.16: 89192373-89192373 on Build GRCh38
rs967195437 Chr.16: 89192322-89192322 on Build GRCh38
rs889023685 Chr.16: 89192417-89192417 on Build GRCh38
rs751480108 Chr.16: 89192348-89192348 on Build GRCh38
rs763503829 Chr.16: 89192167-89192167 on Build GRCh38
rs931721754 Chr.16: 89192067-89192068 on Build GRCh38
rs766136455 Chr.16: 89192423-89192423 on Build GRCh38
rs762727801 Chr.16: 89192342-89192342 on Build GRCh38
rs756248561 Chr.16: 89192440-89192440 on Build GRCh38
rs751505289 Chr.16: 89192278-89192278 on Build GRCh38
rs983354606 Chr.16: 89192022-89192022 on Build GRCh38
rs372310283 Chr.16: 89192247-89192248 on Build GRCh38
rs921847062 Chr.16: 89192049-89192049 on Build GRCh38
rs907815990 Chr.16: 89192025-89192025 on Build GRCh38
rs753225747 Chr.16: 89192135-89192135 on Build GRCh38
rs898949121 Chr.16: 89192405-89192405 on Build GRCh38
rs530716380 Chr.16: 89192508-89192508 on Build GRCh38
rs747627528 Chr.16: 89192408-89192408 on Build GRCh38
rs920419218 Chr.16: 89192013-89192013 on Build GRCh38
rs766224650 Chr.16: 89192347-89192347 on Build GRCh38
rs1033056976 Chr.16: 89192477-89192477 on Build GRCh38
rs757576638 Chr.16: 89192454-89192454 on Build GRCh38
rs750253626 Chr.16: 89192255-89192255 on Build GRCh38
rs758886403 Chr.16: 89192487-89192487 on Build GRCh38
rs997328557 Chr.16: 89192499-89192499 on Build GRCh38
rs539880182 Chr.16: 89192153-89192153 on Build GRCh38
rs781167124 Chr.16: 89192293-89192293 on Build GRCh38
rs563257885 Chr.16: 89192482-89192482 on Build GRCh38
rs752699952 Chr.16: 89192504-89192504 on Build GRCh38
rs760002983 Chr.16: 89192331-89192331 on Build GRCh38
rs1054246200 Chr.16: 89192068-89192068 on Build GRCh38
rs1014628664 Chr.16: 89192015-89192015 on Build GRCh38
rs1042554029 Chr.16: 89192104-89192104 on Build GRCh38
rs944623663 Chr.16: 89192030-89192031 on Build GRCh38
rs1055940925 Chr.16: 89192137-89192137 on Build GRCh38
rs1014423845 Chr.16: 89192170-89192170 on Build GRCh38
rs749673851 Chr.16: 89192317-89192317 on Build GRCh38
rs895124764 Chr.16: 89192332-89192332 on Build GRCh38
rs772842256 Chr.16: 89192416-89192416 on Build GRCh38
rs987743867 Chr.16: 89192085-89192085 on Build GRCh38
rs765178809 Chr.16: 89192323-89192324 on Build GRCh38
rs746338669 Chr.16: 89192392-89192392 on Build GRCh38
rs577570693 Chr.16: 89192395-89192395 on Build GRCh38
rs879802247 Chr.16: 89192254-89192254 on Build GRCh38
rs911976735 Chr.16: 89192087-89192087 on Build GRCh38
rs566167227 Chr.16: 89192091-89192091 on Build GRCh38
rs1028901306 Chr.16: 89192297-89192297 on Build GRCh38
rs761489996 Chr.16: 89192335-89192335 on Build GRCh38
rs763579526 Chr.16: 89192221-89192221 on Build GRCh38
rs779470688 Chr.16: 89192385-89192385 on Build GRCh38
rs758268913 Chr.16: 89192265-89192265 on Build GRCh38
rs778241322 Chr.16: 89192314-89192314 on Build GRCh38
rs901660625 Chr.16: 89192498-89192498 on Build GRCh38

View Full Product Details