Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7601234-7601635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXR2
Entrez Gene ID
9513
Gene Name
FMR1 autosomal homolog 2
Gene Aliases
FMR1L2, FXR2P
UniGene
Hs.52788
Gene Chromosome Location
Chr.17: 7591230-7614897 on Build GRCh38
Transcripts
NM_004860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7099163 c.577C>A p.L193M Chr.17: 7601492-7601492 on Build GRCh38
2744081 c.562G>T p.V188L Chr.17: 7601507-7601507 on Build GRCh38
707085 c.544-1G>A p.? Chr.17: 7601526-7601526 on Build GRCh38
7165881 c.553G>C p.E185Q Chr.17: 7601516-7601516 on Build GRCh38
1285084 c.629C>G p.S210C Chr.17: 7601440-7601440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990177866 Chr.17: 7601299-7601299 on Build GRCh38
rs780843176 Chr.17: 7601567-7601567 on Build GRCh38
rs936520100 Chr.17: 7601587-7601587 on Build GRCh38
rs779483826 Chr.17: 7601373-7601373 on Build GRCh38
rs112934240 Chr.17: 7601318-7601318 on Build GRCh38
rs763396729 Chr.17: 7601533-7601533 on Build GRCh38
rs527566566 Chr.17: 7601330-7601330 on Build GRCh38
rs982899521 Chr.17: 7601297-7601297 on Build GRCh38
rs751041315 Chr.17: 7601558-7601558 on Build GRCh38
rs745836629 Chr.17: 7601561-7601561 on Build GRCh38
rs17643401 Chr.17: 7601540-7601540 on Build GRCh38
rs773638873 Chr.17: 7601328-7601328 on Build GRCh38
rs142277039 Chr.17: 7601578-7601578 on Build GRCh38
rs775462548 Chr.17: 7601481-7601481 on Build GRCh38
rs781164450 Chr.17: 7601331-7601331 on Build GRCh38
rs773109243 Chr.17: 7601454-7601454 on Build GRCh38
rs758141971 Chr.17: 7601360-7601360 on Build GRCh38
rs746736302 Chr.17: 7601390-7601390 on Build GRCh38
rs1052251419 Chr.17: 7601590-7601590 on Build GRCh38
rs1015822153 Chr.17: 7601306-7601306 on Build GRCh38
rs750780667 Chr.17: 7601513-7601513 on Build GRCh38
rs868617231 Chr.17: 7601437-7601437 on Build GRCh38
rs769775559 Chr.17: 7601453-7601453 on Build GRCh38
rs77622986 Chr.17: 7601335-7601335 on Build GRCh38
rs747786710 Chr.17: 7601450-7601450 on Build GRCh38
rs762019264 Chr.17: 7601508-7601508 on Build GRCh38
rs766712760 Chr.17: 7601579-7601579 on Build GRCh38
rs1027044635 Chr.17: 7601261-7601261 on Build GRCh38
rs758909715 Chr.17: 7601372-7601372 on Build GRCh38
rs764314853 Chr.17: 7601494-7601494 on Build GRCh38
rs749231640 Chr.17: 7601479-7601479 on Build GRCh38
rs559943678 Chr.17: 7601279-7601279 on Build GRCh38
rs754570284 Chr.17: 7601560-7601560 on Build GRCh38
rs572373898 Chr.17: 7601275-7601275 on Build GRCh38
rs760742942 Chr.17: 7601489-7601489 on Build GRCh38
rs765633029 Chr.17: 7601511-7601511 on Build GRCh38
rs750998388 Chr.17: 7601365-7601365 on Build GRCh38
rs754587605 Chr.17: 7601443-7601443 on Build GRCh38
rs952894829 Chr.17: 7601280-7601280 on Build GRCh38
rs960277502 Chr.17: 7601314-7601314 on Build GRCh38
rs1022955122 Chr.17: 7601311-7601311 on Build GRCh38
rs746481794 Chr.17: 7601381-7601381 on Build GRCh38
rs980947854 Chr.17: 7601367-7601367 on Build GRCh38

View Full Product Details