Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24609573-24610122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT1
Entrez Gene ID
2678
Gene Name
gamma-glutamyltransferase 1
Gene Aliases
CD224, D22S672, D22S732, GGT, GGT 1, GTG
UniGene
Hs.595809
Gene Chromosome Location
Chr.22: 24583750-24629005 on Build GRCh38
Transcripts
NM_001288833.1 NM_013421.2 NM_013430.2

SNP Detail

SNP ID Chromosome Location
rs888629500 Chr.22: 24610022-24610022 on Build GRCh38
rs201001225 Chr.22: 24609985-24609987 on Build GRCh38
rs184944368 Chr.22: 24610099-24610099 on Build GRCh38
rs532159259 Chr.22: 24609862-24609862 on Build GRCh38
rs4049906 Chr.22: 24610052-24610052 on Build GRCh38
rs149660004 Chr.22: 24609986-24609986 on Build GRCh38
rs62231214 Chr.22: 24609751-24609751 on Build GRCh38
rs577146173 Chr.22: 24609663-24609663 on Build GRCh38
rs952812353 Chr.22: 24609644-24609644 on Build GRCh38
rs934364332 Chr.22: 24609856-24609856 on Build GRCh38
rs1004303388 Chr.22: 24609666-24609666 on Build GRCh38
rs552820947 Chr.22: 24609826-24609826 on Build GRCh38
rs563906307 Chr.22: 24609779-24609779 on Build GRCh38
rs944406220 Chr.22: 24609915-24609915 on Build GRCh38
rs997040409 Chr.22: 24609623-24609623 on Build GRCh38
rs3876105 Chr.22: 24610102-24610102 on Build GRCh38
rs559437164 Chr.22: 24609756-24609756 on Build GRCh38
rs540997847 Chr.22: 24609667-24609667 on Build GRCh38
rs4049905 Chr.22: 24609833-24609833 on Build GRCh38
rs200256865 Chr.22: 24609783-24609783 on Build GRCh38
rs4049904 Chr.22: 24609759-24609759 on Build GRCh38
rs1027058227 Chr.22: 24609634-24609634 on Build GRCh38
rs374980970 Chr.22: 24609825-24609825 on Build GRCh38
rs564758221 Chr.22: 24609847-24609847 on Build GRCh38
rs1041299523 Chr.22: 24609921-24609921 on Build GRCh38
rs4049903 Chr.22: 24609628-24609628 on Build GRCh38
rs201174998 Chr.22: 24609920-24609920 on Build GRCh38
rs576303472 Chr.22: 24609592-24609592 on Build GRCh38
rs543691747 Chr.22: 24609606-24609606 on Build GRCh38
rs777996130 Chr.22: 24609943-24609943 on Build GRCh38
rs911516984 Chr.22: 24609892-24609892 on Build GRCh38
rs74187609 Chr.22: 24610064-24610064 on Build GRCh38
rs747161438 Chr.22: 24609985-24609985 on Build GRCh38
rs978601630 Chr.22: 24609828-24609828 on Build GRCh38
rs146905762 Chr.22: 24609989-24609989 on Build GRCh38
rs4049901 Chr.22: 24609609-24609609 on Build GRCh38
rs574553867 Chr.22: 24609758-24609758 on Build GRCh38
rs374899466 Chr.22: 24610006-24610006 on Build GRCh38
rs1004676703 Chr.22: 24610072-24610072 on Build GRCh38
rs4049902 Chr.22: 24609626-24609626 on Build GRCh38
rs558715201 Chr.22: 24609648-24609648 on Build GRCh38

View Full Product Details