Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29720157-29720702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP7
Entrez Gene ID
164633
Gene Name
calcium binding protein 7
Gene Aliases
CALN2
UniGene
Hs.643608
Gene Chromosome Location
Chr.22: 29720090-29731833 on Build GRCh38
Transcripts
NM_182527.2 XM_006724161.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7358847 c.109G>A p.E37K Chr.22: 29720533-29720533 on Build GRCh38
6617240 c.93G>A p.P31P Chr.22: 29720517-29720517 on Build GRCh38
4618185 c.99C>T p.D33D Chr.22: 29720523-29720523 on Build GRCh38
6394950 c.86A>G p.D29G Chr.22: 29720510-29720510 on Build GRCh38
7358846 c.91C>T p.P31S Chr.22: 29720515-29720515 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201946698 Chr.22: 29720478-29720478 on Build GRCh38
rs750222734 Chr.22: 29720402-29720402 on Build GRCh38
rs750380371 Chr.22: 29720510-29720510 on Build GRCh38
rs1055834576 Chr.22: 29720337-29720337 on Build GRCh38
rs937103124 Chr.22: 29720211-29720211 on Build GRCh38
rs867232139 Chr.22: 29720544-29720544 on Build GRCh38
rs918728455 Chr.22: 29720284-29720284 on Build GRCh38
rs1032331265 Chr.22: 29720262-29720262 on Build GRCh38
rs757339940 Chr.22: 29720556-29720556 on Build GRCh38
rs771410800 Chr.22: 29720444-29720444 on Build GRCh38
rs965811014 Chr.22: 29720278-29720278 on Build GRCh38
rs776053914 Chr.22: 29720574-29720574 on Build GRCh38
rs909587609 Chr.22: 29720300-29720300 on Build GRCh38
rs779771469 Chr.22: 29720412-29720412 on Build GRCh38
rs191509105 Chr.22: 29720267-29720267 on Build GRCh38
rs1035956926 Chr.22: 29720406-29720406 on Build GRCh38
rs1055549849 Chr.22: 29720233-29720233 on Build GRCh38
rs763788690 Chr.22: 29720483-29720483 on Build GRCh38
rs762708772 Chr.22: 29720481-29720481 on Build GRCh38
rs545117020 Chr.22: 29720411-29720411 on Build GRCh38
rs1041574107 Chr.22: 29720361-29720361 on Build GRCh38
rs998659825 Chr.22: 29720471-29720471 on Build GRCh38
rs375700349 Chr.22: 29720416-29720416 on Build GRCh38
rs936993510 Chr.22: 29720314-29720314 on Build GRCh38
rs909530260 Chr.22: 29720186-29720186 on Build GRCh38
rs182614501 Chr.22: 29720634-29720634 on Build GRCh38
rs757199083 Chr.22: 29720398-29720398 on Build GRCh38
rs527598162 Chr.22: 29720546-29720546 on Build GRCh38
rs753576755 Chr.22: 29720536-29720536 on Build GRCh38
rs889047310 Chr.22: 29720642-29720642 on Build GRCh38
rs976961858 Chr.22: 29720282-29720282 on Build GRCh38
rs951633148 Chr.22: 29720286-29720286 on Build GRCh38
rs747101835 Chr.22: 29720548-29720548 on Build GRCh38
rs758609343 Chr.22: 29720543-29720543 on Build GRCh38
rs770305864 Chr.22: 29720470-29720470 on Build GRCh38
rs770359018 Chr.22: 29720572-29720572 on Build GRCh38
rs542794390 Chr.22: 29720559-29720559 on Build GRCh38
rs1018364969 Chr.22: 29720275-29720275 on Build GRCh38
rs781183915 Chr.22: 29720401-29720401 on Build GRCh38
rs560383681 Chr.22: 29720463-29720463 on Build GRCh38
rs1048110035 Chr.22: 29720185-29720185 on Build GRCh38
rs1054300261 Chr.22: 29720390-29720390 on Build GRCh38
rs1041911733 Chr.22: 29720242-29720242 on Build GRCh38
rs1006971106 Chr.22: 29720272-29720272 on Build GRCh38
rs775957525 Chr.22: 29720473-29720473 on Build GRCh38
rs917017438 Chr.22: 29720349-29720349 on Build GRCh38
rs531400391 Chr.22: 29720577-29720577 on Build GRCh38
rs1039731794 Chr.22: 29720651-29720651 on Build GRCh38
rs1000049546 Chr.22: 29720259-29720259 on Build GRCh38
rs777732341 Chr.22: 29720418-29720418 on Build GRCh38
rs901157433 Chr.22: 29720677-29720677 on Build GRCh38
rs924484750 Chr.22: 29720609-29720615 on Build GRCh38
rs760029087 Chr.22: 29720466-29720466 on Build GRCh38
rs1006856463 Chr.22: 29720405-29720405 on Build GRCh38
rs1000100090 Chr.22: 29720372-29720372 on Build GRCh38
rs756002466 Chr.22: 29720514-29720514 on Build GRCh38
rs903000827 Chr.22: 29720367-29720367 on Build GRCh38
rs949813942 Chr.22: 29720351-29720351 on Build GRCh38
rs766162897 Chr.22: 29720515-29720515 on Build GRCh38
rs916881783 Chr.22: 29720596-29720596 on Build GRCh38
rs773799139 Chr.22: 29720492-29720492 on Build GRCh38
rs935841306 Chr.22: 29720633-29720633 on Build GRCh38
rs561088496 Chr.22: 29720571-29720571 on Build GRCh38
rs977565520 Chr.22: 29720608-29720609 on Build GRCh38
rs878980934 Chr.22: 29720289-29720289 on Build GRCh38
rs747101623 Chr.22: 29720433-29720433 on Build GRCh38
rs949856866 Chr.22: 29720598-29720598 on Build GRCh38
rs767401876 Chr.22: 29720502-29720502 on Build GRCh38
rs903117710 Chr.22: 29720257-29720257 on Build GRCh38
rs1025841451 Chr.22: 29720475-29720475 on Build GRCh38
rs761353372 Chr.22: 29720495-29720495 on Build GRCh38
rs768232084 Chr.22: 29720582-29720582 on Build GRCh38

View Full Product Details