Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42618199-42618792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5R3
Entrez Gene ID
1727
Gene Name
cytochrome b5 reductase 3
Gene Aliases
B5R, DIA1
UniGene
Hs.561064
Gene Chromosome Location
Chr.22: 42617840-42649399 on Build GRCh38
Transcripts
NM_000398.6 NM_001129819.2 NM_001171660.1 NM_001171661.1 NM_007326.4

SNP Detail

SNP ID Chromosome Location
rs943375944 Chr.22: 42618559-42618559 on Build GRCh38
rs886569063 Chr.22: 42618616-42618616 on Build GRCh38
rs535394976 Chr.22: 42618330-42618330 on Build GRCh38
rs560619235 Chr.22: 42618356-42618356 on Build GRCh38
rs28602842 Chr.22: 42618725-42618725 on Build GRCh38
rs957741128 Chr.22: 42618417-42618417 on Build GRCh38
rs8190475 Chr.22: 42618345-42618345 on Build GRCh38
rs887215517 Chr.22: 42618393-42618393 on Build GRCh38
rs913710080 Chr.22: 42618464-42618464 on Build GRCh38
rs373606585 Chr.22: 42618724-42618724 on Build GRCh38
rs796374432 Chr.22: 42618725-42618725 on Build GRCh38
rs575963416 Chr.22: 42618342-42618342 on Build GRCh38
rs987299179 Chr.22: 42618732-42618732 on Build GRCh38
rs565502422 Chr.22: 42618412-42618412 on Build GRCh38
rs549952607 Chr.22: 42618382-42618382 on Build GRCh38
rs5758811 Chr.22: 42618709-42618709 on Build GRCh38
rs36011098 Chr.22: 42618708-42618708 on Build GRCh38
rs367972905 Chr.22: 42618290-42618290 on Build GRCh38
rs991772630 Chr.22: 42618606-42618606 on Build GRCh38
rs546246715 Chr.22: 42618366-42618366 on Build GRCh38
rs959085447 Chr.22: 42618605-42618605 on Build GRCh38
rs1051645515 Chr.22: 42618303-42618303 on Build GRCh38
rs550023724 Chr.22: 42618310-42618310 on Build GRCh38
rs543028934 Chr.22: 42618344-42618344 on Build GRCh38
rs564447393 Chr.22: 42618372-42618372 on Build GRCh38
rs980510279 Chr.22: 42618256-42618256 on Build GRCh38
rs191214824 Chr.22: 42618300-42618300 on Build GRCh38
rs980882217 Chr.22: 42618400-42618400 on Build GRCh38
rs920141454 Chr.22: 42618617-42618617 on Build GRCh38
rs796271901 Chr.22: 42618724-42618725 on Build GRCh38
rs539276370 Chr.22: 42618414-42618414 on Build GRCh38
rs573680712 Chr.22: 42618727-42618727 on Build GRCh38
rs1040935077 Chr.22: 42618398-42618398 on Build GRCh38
rs796931131 Chr.22: 42618723-42618723 on Build GRCh38
rs6002817 Chr.22: 42618461-42618461 on Build GRCh38
rs531863245 Chr.22: 42618388-42618388 on Build GRCh38
rs911837577 Chr.22: 42618502-42618502 on Build GRCh38
rs558086551 Chr.22: 42618707-42618708 on Build GRCh38
rs527782100 Chr.22: 42618275-42618275 on Build GRCh38
rs958050842 Chr.22: 42618426-42618426 on Build GRCh38
rs547293392 Chr.22: 42618410-42618410 on Build GRCh38
rs1047854352 Chr.22: 42618607-42618607 on Build GRCh38
rs560705049 Chr.22: 42618263-42618263 on Build GRCh38
rs138986663 Chr.22: 42618697-42618697 on Build GRCh38
rs936512203 Chr.22: 42618262-42618262 on Build GRCh38
rs1051591023 Chr.22: 42618493-42618493 on Build GRCh38
rs537187379 Chr.22: 42618681-42618681 on Build GRCh38
rs981598106 Chr.22: 42618347-42618347 on Build GRCh38
rs796588621 Chr.22: 42618556-42618556 on Build GRCh38
rs1001859773 Chr.22: 42618644-42618644 on Build GRCh38
rs899227080 Chr.22: 42618572-42618572 on Build GRCh38
rs567994910 Chr.22: 42618312-42618312 on Build GRCh38
rs572684803 Chr.22: 42618357-42618357 on Build GRCh38
rs539473847 Chr.22: 42618334-42618334 on Build GRCh38
rs6002816 Chr.22: 42618351-42618351 on Build GRCh38
rs1047134585 Chr.22: 42618638-42618638 on Build GRCh38
rs1024554843 Chr.22: 42618631-42618631 on Build GRCh38
rs1014484670 Chr.22: 42618390-42618390 on Build GRCh38
rs6002819 Chr.22: 42618767-42618767 on Build GRCh38
rs950877916 Chr.22: 42618386-42618386 on Build GRCh38
rs1026505489 Chr.22: 42618382-42618382 on Build GRCh38
rs1041110897 Chr.22: 42618754-42618754 on Build GRCh38
rs1032014779 Chr.22: 42618311-42618311 on Build GRCh38
rs975730798 Chr.22: 42618309-42618309 on Build GRCh38
rs377035556 Chr.22: 42618411-42618411 on Build GRCh38
rs8190476 Chr.22: 42618333-42618333 on Build GRCh38
rs908538998 Chr.22: 42618642-42618642 on Build GRCh38
rs542588073 Chr.22: 42618353-42618353 on Build GRCh38
rs572213557 Chr.22: 42618296-42618296 on Build GRCh38
rs996778563 Chr.22: 42618409-42618409 on Build GRCh38
rs553542057 Chr.22: 42618331-42618331 on Build GRCh38
rs1032651675 Chr.22: 42618416-42618416 on Build GRCh38
rs199517896 Chr.22: 42618364-42618364 on Build GRCh38
rs33966940 Chr.22: 42618536-42618537 on Build GRCh38
rs1009675375 Chr.22: 42618445-42618445 on Build GRCh38
rs989157133 Chr.22: 42618708-42618714 on Build GRCh38
rs1020662829 Chr.22: 42618721-42618721 on Build GRCh38
rs57796194 Chr.22: 42618555-42618556 on Build GRCh38
rs8190474 Chr.22: 42618392-42618392 on Build GRCh38
rs957741067 Chr.22: 42618702-42618702 on Build GRCh38
rs981819168 Chr.22: 42618467-42618467 on Build GRCh38
rs569634247 Chr.22: 42618620-42618620 on Build GRCh38
rs542423980 Chr.22: 42618260-42618260 on Build GRCh38
rs945219076 Chr.22: 42618480-42618480 on Build GRCh38
rs557582318 Chr.22: 42618341-42618341 on Build GRCh38
rs561788002 Chr.22: 42618394-42618394 on Build GRCh38
rs76461531 Chr.22: 42618721-42618723 on Build GRCh38
rs994898564 Chr.22: 42618582-42618582 on Build GRCh38
rs997056517 Chr.22: 42618768-42618768 on Build GRCh38
rs7289184 Chr.22: 42618723-42618723 on Build GRCh38
rs995610286 Chr.22: 42618259-42618259 on Build GRCh38
rs372052721 Chr.22: 42618468-42618468 on Build GRCh38
rs1039472406 Chr.22: 42618564-42618564 on Build GRCh38
rs8190473 Chr.22: 42618406-42618406 on Build GRCh38
rs898480199 Chr.22: 42618236-42618236 on Build GRCh38
rs1020558205 Chr.22: 42618456-42618456 on Build GRCh38
rs7289559 Chr.22: 42618724-42618724 on Build GRCh38
rs531625941 Chr.22: 42618308-42618308 on Build GRCh38
rs34450690 Chr.22: 42618537-42618538 on Build GRCh38
rs995001304 Chr.22: 42618367-42618367 on Build GRCh38
rs1056413889 Chr.22: 42618365-42618365 on Build GRCh38
rs899279814 Chr.22: 42618354-42618354 on Build GRCh38
rs547448233 Chr.22: 42618319-42618320 on Build GRCh38
rs919939508 Chr.22: 42618370-42618370 on Build GRCh38
rs928812615 Chr.22: 42618476-42618476 on Build GRCh38
rs886957745 Chr.22: 42618266-42618266 on Build GRCh38

View Full Product Details