Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46051330-46051734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR34
Entrez Gene ID
55267
Gene Name
proline rich 34
Gene Aliases
C22orf26
UniGene
Hs.567529
Gene Chromosome Location
Chr.22: 46050459-46054144 on Build GRCh38
Transcripts
NM_018280.2

SNP Detail

SNP ID Chromosome Location
rs986909498 Chr.22: 46051592-46051592 on Build GRCh38
rs1047157906 Chr.22: 46051707-46051707 on Build GRCh38
rs573294148 Chr.22: 46051505-46051505 on Build GRCh38
rs80214913 Chr.22: 46051609-46051609 on Build GRCh38
rs936931841 Chr.22: 46051644-46051644 on Build GRCh38
rs558991225 Chr.22: 46051599-46051599 on Build GRCh38
rs190649542 Chr.22: 46051417-46051417 on Build GRCh38
rs756543030 Chr.22: 46051467-46051467 on Build GRCh38
rs954089086 Chr.22: 46051577-46051577 on Build GRCh38
rs898483387 Chr.22: 46051496-46051496 on Build GRCh38
rs532051361 Chr.22: 46051352-46051352 on Build GRCh38
rs142593685 Chr.22: 46051443-46051443 on Build GRCh38
rs368180090 Chr.22: 46051435-46051435 on Build GRCh38
rs978665985 Chr.22: 46051617-46051617 on Build GRCh38
rs369424200 Chr.22: 46051359-46051359 on Build GRCh38
rs1054055628 Chr.22: 46051416-46051416 on Build GRCh38
rs957256435 Chr.22: 46051596-46051596 on Build GRCh38
rs1047550629 Chr.22: 46051500-46051500 on Build GRCh38
rs907143732 Chr.22: 46051554-46051554 on Build GRCh38
rs925523229 Chr.22: 46051632-46051632 on Build GRCh38
rs1039729965 Chr.22: 46051445-46051445 on Build GRCh38
rs989749549 Chr.22: 46051655-46051655 on Build GRCh38
rs1001847395 Chr.22: 46051560-46051560 on Build GRCh38
rs548132546 Chr.22: 46051422-46051422 on Build GRCh38
rs769056274 Chr.22: 46051544-46051544 on Build GRCh38
rs749864293 Chr.22: 46051499-46051499 on Build GRCh38
rs912785679 Chr.22: 46051425-46051425 on Build GRCh38
rs780279459 Chr.22: 46051485-46051485 on Build GRCh38
rs919941319 Chr.22: 46051683-46051683 on Build GRCh38
rs957171240 Chr.22: 46051377-46051377 on Build GRCh38
rs912711671 Chr.22: 46051633-46051633 on Build GRCh38
rs150983976 Chr.22: 46051698-46051698 on Build GRCh38
rs1004691773 Chr.22: 46051511-46051511 on Build GRCh38
rs995473746 Chr.22: 46051532-46051532 on Build GRCh38
rs942721675 Chr.22: 46051434-46051434 on Build GRCh38
rs539099323 Chr.22: 46051521-46051521 on Build GRCh38
rs187685178 Chr.22: 46051454-46051454 on Build GRCh38
rs376245970 Chr.22: 46051413-46051413 on Build GRCh38
rs114143527 Chr.22: 46051392-46051392 on Build GRCh38
rs182941967 Chr.22: 46051423-46051423 on Build GRCh38
rs746070631 Chr.22: 46051429-46051429 on Build GRCh38
rs1034051310 Chr.22: 46051595-46051595 on Build GRCh38
rs1004529057 Chr.22: 46051484-46051484 on Build GRCh38
rs926934861 Chr.22: 46051361-46051361 on Build GRCh38
rs915600308 Chr.22: 46051661-46051661 on Build GRCh38
rs533709746 Chr.22: 46051634-46051634 on Build GRCh38

View Full Product Details