Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115827739-115828293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG12
Entrez Gene ID
9140
Gene Name
autophagy related 12
Gene Aliases
APG12, APG12L, FBR93, HAPG12
UniGene
Hs.264482
Gene Chromosome Location
Chr.5: 115828196-115841851 on Build GRCh38
Transcripts
NM_001277783.1 NM_004707.3 NR_033362.1 NR_033363.1 NR_073603.1

SNP Detail

SNP ID Chromosome Location
rs61582977 Chr.5: 115828013-115828015 on Build GRCh38
rs977733697 Chr.5: 115827771-115827771 on Build GRCh38
rs7726813 Chr.5: 115827779-115827779 on Build GRCh38
rs767651116 Chr.5: 115827796-115827796 on Build GRCh38
rs912612247 Chr.5: 115828109-115828109 on Build GRCh38
rs1022805940 Chr.5: 115827819-115827819 on Build GRCh38
rs931322664 Chr.5: 115828211-115828211 on Build GRCh38
rs188867427 Chr.5: 115828043-115828043 on Build GRCh38
rs902272277 Chr.5: 115828015-115828015 on Build GRCh38
rs921472717 Chr.5: 115827799-115827799 on Build GRCh38
rs902266001 Chr.5: 115828244-115828244 on Build GRCh38
rs749864282 Chr.5: 115827960-115827960 on Build GRCh38
rs1021759453 Chr.5: 115828082-115828082 on Build GRCh38
rs948800415 Chr.5: 115828252-115828252 on Build GRCh38
rs1032005019 Chr.5: 115828022-115828022 on Build GRCh38
rs957686166 Chr.5: 115827781-115827781 on Build GRCh38
rs955846352 Chr.5: 115827907-115827907 on Build GRCh38
rs907165274 Chr.5: 115828042-115828042 on Build GRCh38
rs909342947 Chr.5: 115828225-115828225 on Build GRCh38
rs76678915 Chr.5: 115827899-115827899 on Build GRCh38
rs948520053 Chr.5: 115828025-115828025 on Build GRCh38
rs910768419 Chr.5: 115827967-115827967 on Build GRCh38
rs954210201 Chr.5: 115828050-115828050 on Build GRCh38
rs1013848573 Chr.5: 115828006-115828006 on Build GRCh38
rs932711715 Chr.5: 115827810-115827810 on Build GRCh38
rs747034637 Chr.5: 115828251-115828251 on Build GRCh38
rs919282161 Chr.5: 115828185-115828185 on Build GRCh38
rs562925021 Chr.5: 115827775-115827775 on Build GRCh38
rs976142396 Chr.5: 115827977-115827977 on Build GRCh38
rs144740896 Chr.5: 115828069-115828069 on Build GRCh38
rs772234199 Chr.5: 115828124-115828124 on Build GRCh38
rs1040857227 Chr.5: 115827760-115827760 on Build GRCh38
rs986337455 Chr.5: 115828123-115828123 on Build GRCh38
rs1002535820 Chr.5: 115827871-115827871 on Build GRCh38
rs768834948 Chr.5: 115828259-115828259 on Build GRCh38
rs757774404 Chr.5: 115827964-115827964 on Build GRCh38
rs191710146 Chr.5: 115828122-115828122 on Build GRCh38
rs532179242 Chr.5: 115827834-115827834 on Build GRCh38
rs931273198 Chr.5: 115827945-115827945 on Build GRCh38
rs990288601 Chr.5: 115827915-115827915 on Build GRCh38
rs559607343 Chr.5: 115827929-115827929 on Build GRCh38
rs944938327 Chr.5: 115827974-115827974 on Build GRCh38
rs966311846 Chr.5: 115828140-115828140 on Build GRCh38
rs184798276 Chr.5: 115828001-115828001 on Build GRCh38
rs986860490 Chr.5: 115827825-115827825 on Build GRCh38
rs1055020845 Chr.5: 115828212-115828212 on Build GRCh38
rs750456896 Chr.5: 115827919-115827919 on Build GRCh38
rs60831199 Chr.5: 115828044-115828044 on Build GRCh38
rs986389759 Chr.5: 115827965-115827965 on Build GRCh38
rs553346510 Chr.5: 115828203-115828203 on Build GRCh38
rs916491011 Chr.5: 115828216-115828219 on Build GRCh38
rs891433188 Chr.5: 115828063-115828063 on Build GRCh38
rs573503312 Chr.5: 115828246-115828246 on Build GRCh38
rs548424036 Chr.5: 115827953-115827953 on Build GRCh38
rs559508291 Chr.5: 115828223-115828223 on Build GRCh38
rs897870921 Chr.5: 115827769-115827769 on Build GRCh38
rs1049587684 Chr.5: 115828016-115828016 on Build GRCh38
rs1055346522 Chr.5: 115827980-115827980 on Build GRCh38
rs1044209290 Chr.5: 115828040-115828040 on Build GRCh38
rs919237109 Chr.5: 115827994-115827994 on Build GRCh38
rs912664844 Chr.5: 115827851-115827851 on Build GRCh38
rs887623180 Chr.5: 115828021-115828021 on Build GRCh38
rs755687944 Chr.5: 115827776-115827776 on Build GRCh38
rs976720730 Chr.5: 115828147-115828147 on Build GRCh38
rs192518448 Chr.5: 115827948-115827948 on Build GRCh38

View Full Product Details