Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 118837466-118838022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTWD2
Entrez Gene ID
285605
Gene Name
DTW domain containing 2
Gene Aliases
-
UniGene
Hs.655891
Gene Chromosome Location
Chr.5: 118836074-118989938 on Build GRCh38
Transcripts
NM_001308081.1 NM_173666.3 XM_011543338.2 XM_011543340.2

SNP Detail

SNP ID Chromosome Location
rs73234502 Chr.5: 118837957-118837957 on Build GRCh38
rs935901653 Chr.5: 118837672-118837672 on Build GRCh38
rs537752569 Chr.5: 118837502-118837502 on Build GRCh38
rs921753988 Chr.5: 118837977-118837977 on Build GRCh38
rs571160118 Chr.5: 118837671-118837671 on Build GRCh38
rs1054414143 Chr.5: 118837673-118837673 on Build GRCh38
rs200760864 Chr.5: 118837746-118837746 on Build GRCh38
rs752530068 Chr.5: 118837493-118837493 on Build GRCh38
rs944521124 Chr.5: 118837589-118837589 on Build GRCh38
rs532491290 Chr.5: 118837692-118837692 on Build GRCh38
rs974627012 Chr.5: 118837960-118837960 on Build GRCh38
rs911679643 Chr.5: 118837905-118837905 on Build GRCh38
rs540081767 Chr.5: 118837688-118837688 on Build GRCh38
rs554192776 Chr.5: 118837730-118837730 on Build GRCh38
rs1027343315 Chr.5: 118837836-118837836 on Build GRCh38
rs891767650 Chr.5: 118837702-118837702 on Build GRCh38
rs1010293079 Chr.5: 118837724-118837724 on Build GRCh38
rs950325331 Chr.5: 118837866-118837866 on Build GRCh38
rs935932368 Chr.5: 118837986-118837986 on Build GRCh38
rs557642756 Chr.5: 118837658-118837658 on Build GRCh38
rs911716858 Chr.5: 118837571-118837571 on Build GRCh38
rs747351967 Chr.5: 118837958-118837958 on Build GRCh38
rs370868659 Chr.5: 118837663-118837663 on Build GRCh38
rs572923437 Chr.5: 118837779-118837779 on Build GRCh38
rs765528055 Chr.5: 118837752-118837752 on Build GRCh38
rs553277002 Chr.5: 118837710-118837710 on Build GRCh38
rs1039271469 Chr.5: 118837638-118837638 on Build GRCh38
rs983614747 Chr.5: 118837884-118837884 on Build GRCh38
rs180990651 Chr.5: 118837492-118837492 on Build GRCh38
rs907370230 Chr.5: 118837750-118837750 on Build GRCh38
rs574846020 Chr.5: 118837874-118837874 on Build GRCh38
rs758417768 Chr.5: 118837846-118837846 on Build GRCh38
rs541532371 Chr.5: 118837798-118837798 on Build GRCh38
rs555357011 Chr.5: 118837829-118837829 on Build GRCh38
rs758583981 Chr.5: 118837849-118837849 on Build GRCh38
rs543801490 Chr.5: 118837895-118837895 on Build GRCh38
rs974295115 Chr.5: 118837830-118837830 on Build GRCh38
rs983198184 Chr.5: 118837569-118837569 on Build GRCh38

View Full Product Details