Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161624325-161624871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGR3B
Entrez Gene ID
2215
Gene Name
Fc fragment of IgG receptor IIIb
Gene Aliases
CD16, CD16b, FCG3, FCGR3, FCR-10, FCRIII, FCRIIIb
UniGene
Hs.372679
Gene Chromosome Location
Chr.1: 161623196-161631963 on Build GRCh38
Transcripts
NM_000570.4 NM_001244753.1 NM_001271035.1 NM_001271036.1 NM_001271037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7430361 c.608C>T p.S203F Chr.1: 161624609-161624609 on Build GRCh38
7430360 c.700T>C p.*234R Chr.1: 161624517-161624517 on Build GRCh38
5641082 c.607T>C p.S203P Chr.1: 161624610-161624610 on Build GRCh38
255602 c.582G>T p.L194F Chr.1: 161624635-161624635 on Build GRCh38
898579 c.616G>A p.G206R Chr.1: 161624601-161624601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746873464 Chr.1: 161624528-161624528 on Build GRCh38
rs1016051507 Chr.1: 161624452-161624452 on Build GRCh38
rs201601910 Chr.1: 161624518-161624518 on Build GRCh38
rs751873543 Chr.1: 161624671-161624671 on Build GRCh38
rs1038748354 Chr.1: 161624385-161624385 on Build GRCh38
rs781246596 Chr.1: 161624621-161624621 on Build GRCh38
rs764580654 Chr.1: 161624590-161624590 on Build GRCh38
rs560051377 Chr.1: 161624624-161624624 on Build GRCh38
rs545265964 Chr.1: 161624839-161624839 on Build GRCh38
rs978928295 Chr.1: 161624618-161624618 on Build GRCh38
rs769754931 Chr.1: 161624544-161624544 on Build GRCh38
rs534433689 Chr.1: 161624411-161624411 on Build GRCh38
rs895820578 Chr.1: 161624781-161624781 on Build GRCh38
rs142480337 Chr.1: 161624768-161624768 on Build GRCh38
rs1049022758 Chr.1: 161624424-161624424 on Build GRCh38
rs376757069 Chr.1: 161624468-161624468 on Build GRCh38
rs61803009 Chr.1: 161624456-161624456 on Build GRCh38
rs1023372722 Chr.1: 161624799-161624799 on Build GRCh38
rs768731670 Chr.1: 161624530-161624530 on Build GRCh38
rs562888488 Chr.1: 161624678-161624678 on Build GRCh38
rs779163663 Chr.1: 161624512-161624512 on Build GRCh38
rs900335560 Chr.1: 161624386-161624386 on Build GRCh38
rs371366380 Chr.1: 161624477-161624477 on Build GRCh38
rs568756065 Chr.1: 161624350-161624350 on Build GRCh38
rs752644223 Chr.1: 161624679-161624679 on Build GRCh38
rs547672432 Chr.1: 161624421-161624421 on Build GRCh38
rs895780402 Chr.1: 161624478-161624478 on Build GRCh38
rs761203323 Chr.1: 161624494-161624494 on Build GRCh38
rs781340215 Chr.1: 161624533-161624533 on Build GRCh38
rs540459343 Chr.1: 161624568-161624568 on Build GRCh38
rs370248850 Chr.1: 161624661-161624661 on Build GRCh38
rs774057129 Chr.1: 161624642-161624642 on Build GRCh38
rs762537225 Chr.1: 161624558-161624558 on Build GRCh38
rs765859000 Chr.1: 161624488-161624488 on Build GRCh38
rs765566147 Chr.1: 161624655-161624655 on Build GRCh38
rs61803010 Chr.1: 161624735-161624735 on Build GRCh38
rs773739723 Chr.1: 161624571-161624571 on Build GRCh38
rs762142186 Chr.1: 161624600-161624600 on Build GRCh38
rs769561727 Chr.1: 161624480-161624480 on Build GRCh38
rs574639231 Chr.1: 161624550-161624550 on Build GRCh38
rs879196167 Chr.1: 161624380-161624380 on Build GRCh38
rs138362091 Chr.1: 161624634-161624634 on Build GRCh38
rs780024332 Chr.1: 161624520-161624520 on Build GRCh38
rs755089839 Chr.1: 161624675-161624675 on Build GRCh38
rs267598136 Chr.1: 161624567-161624567 on Build GRCh38
rs759104285 Chr.1: 161624587-161624587 on Build GRCh38
rs71485860 Chr.1: 161624735-161624736 on Build GRCh38
rs148717049 Chr.1: 161624762-161624762 on Build GRCh38
rs762336406 Chr.1: 161624654-161624654 on Build GRCh38
rs754188119 Chr.1: 161624498-161624498 on Build GRCh38
rs754380403 Chr.1: 161624591-161624591 on Build GRCh38
rs757684137 Chr.1: 161624511-161624511 on Build GRCh38
rs971897293 Chr.1: 161624523-161624523 on Build GRCh38
rs762495381 Chr.1: 161624485-161624485 on Build GRCh38
rs766509789 Chr.1: 161624670-161624670 on Build GRCh38
rs777070197 Chr.1: 161624653-161624653 on Build GRCh38
rs749031518 Chr.1: 161624691-161624691 on Build GRCh38
rs558150467 Chr.1: 161624467-161624467 on Build GRCh38
rs766283545 Chr.1: 161624662-161624663 on Build GRCh38
rs775127796 Chr.1: 161624776-161624776 on Build GRCh38
rs745348868 Chr.1: 161624645-161624645 on Build GRCh38
rs750615825 Chr.1: 161624606-161624606 on Build GRCh38
rs762858040 Chr.1: 161624663-161624663 on Build GRCh38
rs61803008 Chr.1: 161624436-161624436 on Build GRCh38
rs766420385 Chr.1: 161624614-161624614 on Build GRCh38
rs1033121089 Chr.1: 161624623-161624623 on Build GRCh38
rs537431910 Chr.1: 161624484-161624484 on Build GRCh38
rs1048949300 Chr.1: 161624699-161624699 on Build GRCh38
rs1000234421 Chr.1: 161624830-161624830 on Build GRCh38
rs538742500 Chr.1: 161624779-161624779 on Build GRCh38
rs61803011 Chr.1: 161624736-161624736 on Build GRCh38
rs886387398 Chr.1: 161624425-161624425 on Build GRCh38
rs905979605 Chr.1: 161624800-161624800 on Build GRCh38
rs753199357 Chr.1: 161624491-161624491 on Build GRCh38
rs756009884 Chr.1: 161624682-161624682 on Build GRCh38
rs772778024 Chr.1: 161624645-161624647 on Build GRCh38
rs771618565 Chr.1: 161624652-161624652 on Build GRCh38
rs374752953 Chr.1: 161624517-161624517 on Build GRCh38
rs989069112 Chr.1: 161624647-161624647 on Build GRCh38
rs61803012 Chr.1: 161624771-161624771 on Build GRCh38
rs545955956 Chr.1: 161624662-161624662 on Build GRCh38
rs747954960 Chr.1: 161624537-161624537 on Build GRCh38
rs1014225593 Chr.1: 161624795-161624795 on Build GRCh38
rs755891248 Chr.1: 161624627-161624627 on Build GRCh38
rs777460504 Chr.1: 161624633-161624633 on Build GRCh38
rs765539832 Chr.1: 161624604-161624604 on Build GRCh38
rs373797758 Chr.1: 161624689-161624689 on Build GRCh38
rs1004786602 Chr.1: 161624432-161624432 on Build GRCh38
rs758550229 Chr.1: 161624609-161624609 on Build GRCh38

View Full Product Details