Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173806269-173806855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPL
Entrez Gene ID
91687
Gene Name
centromere protein L
Gene Aliases
C1orf155, CENP-L, dJ383J4.3
UniGene
Hs.531856
Gene Chromosome Location
Chr.1: 173799550-173824639 on Build GRCh38
Transcripts
NM_001127181.2 NM_001171182.1 NM_033319.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6019616 c.506C>G p.S169C Chr.1: 173806491-173806491 on Build GRCh38
5785881 c.486C>T p.L162L Chr.1: 173806511-173806511 on Build GRCh38
5735739 c.427A>C p.I143L Chr.1: 173806570-173806570 on Build GRCh38
5718338 c.506C>T p.S169F Chr.1: 173806491-173806491 on Build GRCh38
3396132 c.428T>C p.I143T Chr.1: 173806569-173806569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962326281 Chr.1: 173806578-173806578 on Build GRCh38
rs771899900 Chr.1: 173806559-173806559 on Build GRCh38
rs1014029368 Chr.1: 173806524-173806524 on Build GRCh38
rs748887766 Chr.1: 173806640-173806640 on Build GRCh38
rs554849443 Chr.1: 173806322-173806322 on Build GRCh38
rs547630143 Chr.1: 173806620-173806620 on Build GRCh38
rs994029974 Chr.1: 173806407-173806407 on Build GRCh38
rs187541745 Chr.1: 173806648-173806648 on Build GRCh38
rs1005939482 Chr.1: 173806773-173806773 on Build GRCh38
rs958719672 Chr.1: 173806521-173806521 on Build GRCh38
rs906835238 Chr.1: 173806439-173806439 on Build GRCh38
rs116533910 Chr.1: 173806328-173806328 on Build GRCh38
rs976710846 Chr.1: 173806579-173806579 on Build GRCh38
rs942686224 Chr.1: 173806368-173806368 on Build GRCh38
rs1014413674 Chr.1: 173806829-173806829 on Build GRCh38
rs936992652 Chr.1: 173806759-173806759 on Build GRCh38
rs560897092 Chr.1: 173806695-173806695 on Build GRCh38
rs935824622 Chr.1: 173806356-173806356 on Build GRCh38
rs769153969 Chr.1: 173806427-173806427 on Build GRCh38
rs768470081 Chr.1: 173806651-173806651 on Build GRCh38
rs79947184 Chr.1: 173806351-173806351 on Build GRCh38
rs567453990 Chr.1: 173806769-173806769 on Build GRCh38
rs928346604 Chr.1: 173806753-173806753 on Build GRCh38
rs1005350443 Chr.1: 173806508-173806508 on Build GRCh38
rs557728480 Chr.1: 173806406-173806406 on Build GRCh38
rs1046039890 Chr.1: 173806751-173806751 on Build GRCh38
rs924192640 Chr.1: 173806622-173806622 on Build GRCh38
rs545022441 Chr.1: 173806511-173806511 on Build GRCh38
rs1034084526 Chr.1: 173806453-173806453 on Build GRCh38
rs1054334551 Chr.1: 173806388-173806388 on Build GRCh38
rs1041161278 Chr.1: 173806668-173806668 on Build GRCh38
rs1041460385 Chr.1: 173806377-173806377 on Build GRCh38
rs888378034 Chr.1: 173806737-173806737 on Build GRCh38
rs746194624 Chr.1: 173806675-173806675 on Build GRCh38
rs1030349855 Chr.1: 173806590-173806590 on Build GRCh38
rs571824413 Chr.1: 173806430-173806430 on Build GRCh38
rs1025557763 Chr.1: 173806474-173806474 on Build GRCh38
rs541152182 Chr.1: 173806611-173806611 on Build GRCh38
rs986596905 Chr.1: 173806350-173806350 on Build GRCh38
rs112601819 Chr.1: 173806770-173806770 on Build GRCh38
rs778782546 Chr.1: 173806426-173806426 on Build GRCh38
rs953484625 Chr.1: 173806607-173806607 on Build GRCh38
rs200190200 Chr.1: 173806697-173806698 on Build GRCh38
rs1046932569 Chr.1: 173806735-173806735 on Build GRCh38
rs945915590 Chr.1: 173806411-173806411 on Build GRCh38
rs879451491 Chr.1: 173806682-173806682 on Build GRCh38
rs887977781 Chr.1: 173806485-173806485 on Build GRCh38
rs536153116 Chr.1: 173806610-173806610 on Build GRCh38
rs958157413 Chr.1: 173806631-173806631 on Build GRCh38
rs888243652 Chr.1: 173806481-173806481 on Build GRCh38
rs185035344 Chr.1: 173806580-173806580 on Build GRCh38
rs894338698 Chr.1: 173806805-173806805 on Build GRCh38
rs1001312942 Chr.1: 173806443-173806443 on Build GRCh38
rs572058929 Chr.1: 173806596-173806596 on Build GRCh38
rs989660108 Chr.1: 173806633-173806633 on Build GRCh38
rs891712243 Chr.1: 173806394-173806394 on Build GRCh38
rs1048033883 Chr.1: 173806482-173806482 on Build GRCh38
rs922584297 Chr.1: 173806681-173806681 on Build GRCh38

View Full Product Details