Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196788014-196788574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFHR3
Entrez Gene ID
10878
Gene Name
complement factor H related 3
Gene Aliases
CFHL3, DOWN16, FHR-3, FHR3, HLF4
UniGene
Hs.709217
Gene Chromosome Location
Chr.1: 196774800-196794073 on Build GRCh38
Transcripts
NM_001166624.1 NM_021023.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1560361 c.590G>C p.W197S Chr.1: 196788375-196788375 on Build GRCh38
4552163 c.550G>A p.G184R Chr.1: 196788335-196788335 on Build GRCh38
3480833 c.563G>A p.G188E Chr.1: 196788348-196788348 on Build GRCh38
3480834 c.576T>C p.C192C Chr.1: 196788361-196788361 on Build GRCh38
678417 c.548A>G p.D183G Chr.1: 196788333-196788333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2454307 Chr.1: 196788180-196788180 on Build GRCh38
rs400642 Chr.1: 196788128-196788128 on Build GRCh38
rs950045254 Chr.1: 196788548-196788548 on Build GRCh38
rs374828613 Chr.1: 196788479-196788479 on Build GRCh38
rs776666479 Chr.1: 196788269-196788269 on Build GRCh38
rs769643369 Chr.1: 196788415-196788415 on Build GRCh38
rs529192695 Chr.1: 196788125-196788125 on Build GRCh38
rs749563406 Chr.1: 196788316-196788316 on Build GRCh38
rs774842869 Chr.1: 196788347-196788347 on Build GRCh38
rs758859916 Chr.1: 196788197-196788197 on Build GRCh38
rs780229985 Chr.1: 196788397-196788397 on Build GRCh38
rs750761913 Chr.1: 196788440-196788440 on Build GRCh38
rs561309568 Chr.1: 196788116-196788116 on Build GRCh38
rs550827961 Chr.1: 196788495-196788495 on Build GRCh38
rs2986139 Chr.1: 196788488-196788488 on Build GRCh38
rs267598270 Chr.1: 196788245-196788245 on Build GRCh38
rs762478917 Chr.1: 196788349-196788349 on Build GRCh38
rs746316559 Chr.1: 196788313-196788313 on Build GRCh38
rs138077074 Chr.1: 196788386-196788386 on Build GRCh38
rs759117982 Chr.1: 196788270-196788270 on Build GRCh38
rs5002712 Chr.1: 196788380-196788380 on Build GRCh38
rs745852616 Chr.1: 196788413-196788413 on Build GRCh38
rs200996170 Chr.1: 196788231-196788231 on Build GRCh38
rs760466080 Chr.1: 196788358-196788358 on Build GRCh38
rs186597151 Chr.1: 196788168-196788168 on Build GRCh38
rs76873424 Chr.1: 196788307-196788307 on Build GRCh38
rs772850682 Chr.1: 196788254-196788254 on Build GRCh38
rs556553587 Chr.1: 196788315-196788315 on Build GRCh38
rs752465342 Chr.1: 196788280-196788280 on Build GRCh38
rs993902985 Chr.1: 196788142-196788142 on Build GRCh38
rs373850719 Chr.1: 196788424-196788424 on Build GRCh38
rs768814707 Chr.1: 196788398-196788401 on Build GRCh38
rs572534647 Chr.1: 196788040-196788040 on Build GRCh38
rs138839071 Chr.1: 196788400-196788400 on Build GRCh38
rs576457161 Chr.1: 196788520-196788520 on Build GRCh38
rs555110024 Chr.1: 196788497-196788497 on Build GRCh38
rs776539681 Chr.1: 196788408-196788408 on Build GRCh38
rs762375718 Chr.1: 196788418-196788418 on Build GRCh38
rs543019140 Chr.1: 196788046-196788046 on Build GRCh38
rs147012038 Chr.1: 196788385-196788385 on Build GRCh38
rs5002710 Chr.1: 196788297-196788297 on Build GRCh38
rs771969113 Chr.1: 196788250-196788250 on Build GRCh38
rs138865637 Chr.1: 196788439-196788439 on Build GRCh38
rs537454242 Chr.1: 196788308-196788308 on Build GRCh38
rs754485848 Chr.1: 196788179-196788179 on Build GRCh38
rs745980647 Chr.1: 196788237-196788237 on Build GRCh38
rs180855280 Chr.1: 196788143-196788143 on Build GRCh38
rs548560485 Chr.1: 196788249-196788249 on Build GRCh38
rs913240333 Chr.1: 196788523-196788523 on Build GRCh38
rs370867050 Chr.1: 196788341-196788341 on Build GRCh38
rs752219812 Chr.1: 196788193-196788193 on Build GRCh38
rs578209193 Chr.1: 196788327-196788327 on Build GRCh38
rs770921062 Chr.1: 196788407-196788407 on Build GRCh38
rs201601675 Chr.1: 196788236-196788236 on Build GRCh38
rs756581256 Chr.1: 196788391-196788391 on Build GRCh38
rs533688197 Chr.1: 196788198-196788198 on Build GRCh38
rs748318693 Chr.1: 196788405-196788405 on Build GRCh38
rs768156112 Chr.1: 196788419-196788419 on Build GRCh38
rs774048937 Chr.1: 196788422-196788422 on Build GRCh38
rs747595353 Chr.1: 196788216-196788216 on Build GRCh38
rs368661691 Chr.1: 196788382-196788382 on Build GRCh38
rs757959248 Chr.1: 196788290-196788290 on Build GRCh38
rs963937303 Chr.1: 196788322-196788322 on Build GRCh38
rs551588188 Chr.1: 196788157-196788157 on Build GRCh38
rs749347442 Chr.1: 196788399-196788399 on Build GRCh38
rs764830712 Chr.1: 196788271-196788271 on Build GRCh38
rs375050298 Chr.1: 196788367-196788367 on Build GRCh38
rs766438449 Chr.1: 196788263-196788263 on Build GRCh38
rs372082762 Chr.1: 196788417-196788417 on Build GRCh38
rs2454308 Chr.1: 196788171-196788171 on Build GRCh38
rs377378334 Chr.1: 196788334-196788334 on Build GRCh38
rs768736986 Chr.1: 196788325-196788325 on Build GRCh38
rs751894448 Chr.1: 196788303-196788303 on Build GRCh38
rs572882916 Chr.1: 196788129-196788129 on Build GRCh38
rs201998349 Chr.1: 196788345-196788345 on Build GRCh38
rs400344 Chr.1: 196788262-196788262 on Build GRCh38
rs751159096 Chr.1: 196788371-196788371 on Build GRCh38
rs201711660 Chr.1: 196788447-196788450 on Build GRCh38
rs746828567 Chr.1: 196788330-196788330 on Build GRCh38

View Full Product Details