Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94781462-94782041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C19
Entrez Gene ID
1557
Gene Name
cytochrome P450 family 2 subfamily C member 19
Gene Aliases
CPCJ, CYP2C, CYPIIC17, CYPIIC19, P450C2C, P450IIC19
UniGene
Hs.282409
Gene Chromosome Location
Chr.10: 94762681-94853205 on Build GRCh38
Transcripts
NM_000769.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6639194 c.782G>A p.R261Q Chr.10: 94781960-94781960 on Build GRCh38
5915922 c.727G>A p.D243N Chr.10: 94781905-94781905 on Build GRCh38
3441954 c.658C>T p.P220S Chr.10: 94781836-94781836 on Build GRCh38
1970806 c.762G>A p.S254S Chr.10: 94781940-94781940 on Build GRCh38
6518129 c.754C>A p.Q252K Chr.10: 94781932-94781932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148247410 Chr.10: 94781931-94781931 on Build GRCh38
rs267602635 Chr.10: 94781844-94781844 on Build GRCh38
rs947604018 Chr.10: 94781728-94781728 on Build GRCh38
rs770993888 Chr.10: 94781946-94781946 on Build GRCh38
rs750070968 Chr.10: 94781969-94781969 on Build GRCh38
rs756556587 Chr.10: 94781982-94781982 on Build GRCh38
rs577255883 Chr.10: 94781962-94781962 on Build GRCh38
rs758728151 Chr.10: 94781648-94781648 on Build GRCh38
rs755993846 Chr.10: 94781825-94781825 on Build GRCh38
rs1041987233 Chr.10: 94781754-94781754 on Build GRCh38
rs914529821 Chr.10: 94781950-94781950 on Build GRCh38
rs574462231 Chr.10: 94781960-94781960 on Build GRCh38
rs559538785 Chr.10: 94781568-94781568 on Build GRCh38
rs958357742 Chr.10: 94781507-94781507 on Build GRCh38
rs775544863 Chr.10: 94781899-94781899 on Build GRCh38
rs1012622596 Chr.10: 94781548-94781548 on Build GRCh38
rs768974528 Chr.10: 94781849-94781849 on Build GRCh38
rs768672419 Chr.10: 94782012-94782012 on Build GRCh38
rs550670906 Chr.10: 94781699-94781699 on Build GRCh38
rs778258371 Chr.10: 94781991-94781991 on Build GRCh38
rs199556059 Chr.10: 94782001-94782001 on Build GRCh38
rs772481099 Chr.10: 94782009-94782009 on Build GRCh38
rs767505657 Chr.10: 94781817-94781817 on Build GRCh38
rs375781227 Chr.10: 94781944-94781944 on Build GRCh38
rs892619604 Chr.10: 94781536-94781538 on Build GRCh38
rs770126483 Chr.10: 94781871-94781871 on Build GRCh38
rs59162501 Chr.10: 94781806-94781806 on Build GRCh38
rs780223701 Chr.10: 94782010-94782010 on Build GRCh38
rs528370827 Chr.10: 94781592-94781592 on Build GRCh38
rs879130837 Chr.10: 94781858-94781859 on Build GRCh38
rs753510310 Chr.10: 94781834-94781834 on Build GRCh38
rs749678783 Chr.10: 94781942-94781942 on Build GRCh38
rs760734417 Chr.10: 94781904-94781904 on Build GRCh38
rs57752480 Chr.10: 94781781-94781781 on Build GRCh38
rs6413438 Chr.10: 94781858-94781858 on Build GRCh38
rs532287106 Chr.10: 94781534-94781534 on Build GRCh38
rs773907495 Chr.10: 94781538-94781538 on Build GRCh38
rs764751186 Chr.10: 94781964-94781964 on Build GRCh38
rs200362177 Chr.10: 94781854-94781854 on Build GRCh38
rs866704734 Chr.10: 94781926-94781926 on Build GRCh38
rs759379748 Chr.10: 94781812-94781812 on Build GRCh38
rs964119913 Chr.10: 94781925-94781925 on Build GRCh38
rs751729858 Chr.10: 94781809-94781809 on Build GRCh38
rs140267616 Chr.10: 94781976-94781976 on Build GRCh38
rs954545346 Chr.10: 94781877-94781877 on Build GRCh38
rs368040759 Chr.10: 94781669-94781669 on Build GRCh38
rs749564114 Chr.10: 94781992-94781992 on Build GRCh38
rs976623069 Chr.10: 94781642-94781642 on Build GRCh38
rs7088784 Chr.10: 94781616-94781616 on Build GRCh38
rs72558186 Chr.10: 94781999-94781999 on Build GRCh38
rs765716857 Chr.10: 94781557-94781557 on Build GRCh38
rs972023566 Chr.10: 94781603-94781603 on Build GRCh38
rs761341117 Chr.10: 94781547-94781547 on Build GRCh38
rs894663229 Chr.10: 94781998-94781998 on Build GRCh38
rs767307799 Chr.10: 94781891-94781891 on Build GRCh38
rs963088247 Chr.10: 94781846-94781846 on Build GRCh38
rs745394523 Chr.10: 94781841-94781841 on Build GRCh38
rs548530129 Chr.10: 94781610-94781610 on Build GRCh38
rs781265742 Chr.10: 94781914-94781914 on Build GRCh38
rs556994963 Chr.10: 94781956-94781956 on Build GRCh38
rs867067400 Chr.10: 94781836-94781836 on Build GRCh38
rs566311971 Chr.10: 94781838-94781838 on Build GRCh38
rs986809473 Chr.10: 94781628-94781628 on Build GRCh38
rs763972230 Chr.10: 94781906-94781906 on Build GRCh38
rs1003795838 Chr.10: 94781505-94781505 on Build GRCh38
rs774654016 Chr.10: 94781947-94781947 on Build GRCh38
rs759690171 Chr.10: 94781890-94781890 on Build GRCh38
rs17879677 Chr.10: 94781774-94781774 on Build GRCh38
rs530533457 Chr.10: 94781652-94781652 on Build GRCh38
rs942697360 Chr.10: 94781684-94781684 on Build GRCh38
rs774238831 Chr.10: 94781889-94781889 on Build GRCh38
rs772159414 Chr.10: 94781955-94781955 on Build GRCh38
rs983944038 Chr.10: 94781721-94781721 on Build GRCh38
rs75669415 Chr.10: 94781560-94781560 on Build GRCh38
rs4244285 Chr.10: 94781859-94781859 on Build GRCh38
rs576823729 Chr.10: 94781826-94781826 on Build GRCh38
rs574458036 Chr.10: 94781916-94781916 on Build GRCh38
rs903311100 Chr.10: 94781780-94781780 on Build GRCh38
rs773448686 Chr.10: 94781789-94781789 on Build GRCh38
rs769966695 Chr.10: 94781788-94781788 on Build GRCh38
rs761561376 Chr.10: 94781529-94781531 on Build GRCh38
rs753605541 Chr.10: 94781920-94781920 on Build GRCh38
rs757951462 Chr.10: 94781927-94781927 on Build GRCh38
rs1040138889 Chr.10: 94781697-94781697 on Build GRCh38
rs201983413 Chr.10: 94781794-94781794 on Build GRCh38
rs377674118 Chr.10: 94781979-94781979 on Build GRCh38
rs748628122 Chr.10: 94781932-94781932 on Build GRCh38
rs779184961 Chr.10: 94782006-94782006 on Build GRCh38
rs771243716 Chr.10: 94781886-94781886 on Build GRCh38
rs778044050 Chr.10: 94781785-94781785 on Build GRCh38
rs764137538 Chr.10: 94781959-94781959 on Build GRCh38
rs963646219 Chr.10: 94781571-94781571 on Build GRCh38
rs747088384 Chr.10: 94782011-94782011 on Build GRCh38
rs752498998 Chr.10: 94781820-94781820 on Build GRCh38
rs72558185 Chr.10: 94781899-94781900 on Build GRCh38
rs914811909 Chr.10: 94781716-94781716 on Build GRCh38
rs776655506 Chr.10: 94782015-94782015 on Build GRCh38
rs866297301 Chr.10: 94781855-94781855 on Build GRCh38
rs749507932 Chr.10: 94781885-94781885 on Build GRCh38
rs756681183 Chr.10: 94781940-94781940 on Build GRCh38
rs537050749 Chr.10: 94781951-94781951 on Build GRCh38

View Full Product Details