Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95620962-95621320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH18A1
Entrez Gene ID
5832
Gene Name
aldehyde dehydrogenase 18 family member A1
Gene Aliases
ADCL3, ARCL3A, GSAS, P5CS, PYCS, SPG9A, SPG9B
UniGene
Hs.500645
Gene Chromosome Location
Chr.10: 95605929-95656810 on Build GRCh38
Transcripts
NM_001017423.1 NM_001323412.1 NM_001323413.1 NM_001323414.1 NM_001323415.1 NM_001323416.1 NM_001323417.1 NM_001323418.1 NM_001323419.1 NM_002860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7304258 c.1394A>G p.E465G Chr.10: 95621104-95621104 on Build GRCh38
5650429 c.1429C>T p.L477L Chr.10: 95621069-95621069 on Build GRCh38
6598162 c.1370G>A p.R457H Chr.10: 95621128-95621128 on Build GRCh38
1603900 c.1365G>A p.L455L Chr.10: 95621133-95621133 on Build GRCh38
6210616 c.1274G>A p.R425H Chr.10: 95621224-95621224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768836597 Chr.10: 95621203-95621203 on Build GRCh38
rs373239891 Chr.10: 95621081-95621081 on Build GRCh38
rs539760223 Chr.10: 95621161-95621161 on Build GRCh38
rs748944695 Chr.10: 95620990-95620990 on Build GRCh38
rs776404338 Chr.10: 95621208-95621208 on Build GRCh38
rs762742204 Chr.10: 95621225-95621225 on Build GRCh38
rs886047511 Chr.10: 95621113-95621113 on Build GRCh38
rs922059612 Chr.10: 95621034-95621034 on Build GRCh38
rs772575442 Chr.10: 95621270-95621270 on Build GRCh38
rs758587954 Chr.10: 95621012-95621013 on Build GRCh38
rs760865106 Chr.10: 95621131-95621131 on Build GRCh38
rs369979669 Chr.10: 95621130-95621130 on Build GRCh38
rs763868475 Chr.10: 95621132-95621132 on Build GRCh38
rs748381335 Chr.10: 95621033-95621033 on Build GRCh38
rs750182421 Chr.10: 95621223-95621223 on Build GRCh38
rs775855406 Chr.10: 95621187-95621187 on Build GRCh38
rs144816455 Chr.10: 95621190-95621190 on Build GRCh38
rs767770922 Chr.10: 95621042-95621042 on Build GRCh38
rs756806946 Chr.10: 95620991-95620991 on Build GRCh38
rs563762852 Chr.10: 95621274-95621274 on Build GRCh38
rs1031928854 Chr.10: 95621259-95621259 on Build GRCh38
rs1055045758 Chr.10: 95621157-95621157 on Build GRCh38
rs771386671 Chr.10: 95621122-95621122 on Build GRCh38
rs777121189 Chr.10: 95621003-95621003 on Build GRCh38
rs949183256 Chr.10: 95621017-95621017 on Build GRCh38
rs570730665 Chr.10: 95621128-95621128 on Build GRCh38
rs141292408 Chr.10: 95621183-95621183 on Build GRCh38
rs748211958 Chr.10: 95621290-95621290 on Build GRCh38
rs780407926 Chr.10: 95621277-95621277 on Build GRCh38
rs768600105 Chr.10: 95621281-95621281 on Build GRCh38
rs779315840 Chr.10: 95621267-95621267 on Build GRCh38
rs745305919 Chr.10: 95620998-95620998 on Build GRCh38
rs749559862 Chr.10: 95621121-95621121 on Build GRCh38
rs866572447 Chr.10: 95621209-95621209 on Build GRCh38
rs937571172 Chr.10: 95621053-95621053 on Build GRCh38
rs745604657 Chr.10: 95621096-95621096 on Build GRCh38
rs777749881 Chr.10: 95620989-95620989 on Build GRCh38
rs778655924 Chr.10: 95620994-95620994 on Build GRCh38
rs774430779 Chr.10: 95621127-95621127 on Build GRCh38
rs537092003 Chr.10: 95621178-95621178 on Build GRCh38
rs370606517 Chr.10: 95621289-95621289 on Build GRCh38
rs773774976 Chr.10: 95621120-95621120 on Build GRCh38
rs759518976 Chr.10: 95621041-95621041 on Build GRCh38
rs754482244 Chr.10: 95621240-95621240 on Build GRCh38
rs780793038 Chr.10: 95621244-95621244 on Build GRCh38
rs756168752 Chr.10: 95621050-95621050 on Build GRCh38
rs373404824 Chr.10: 95621168-95621168 on Build GRCh38
rs774648094 Chr.10: 95621035-95621035 on Build GRCh38
rs773147707 Chr.10: 95621293-95621293 on Build GRCh38
rs145289559 Chr.10: 95621177-95621177 on Build GRCh38
rs757733068 Chr.10: 95621250-95621250 on Build GRCh38
rs142712849 Chr.10: 95621234-95621234 on Build GRCh38
rs111344269 Chr.10: 95621147-95621147 on Build GRCh38
rs887933055 Chr.10: 95621123-95621123 on Build GRCh38
rs150429857 Chr.10: 95621117-95621117 on Build GRCh38
rs765113723 Chr.10: 95621216-95621216 on Build GRCh38
rs117709404 Chr.10: 95621169-95621169 on Build GRCh38
rs747278089 Chr.10: 95621278-95621278 on Build GRCh38
rs201069261 Chr.10: 95621245-95621245 on Build GRCh38
rs568501656 Chr.10: 95621044-95621044 on Build GRCh38
rs757876226 Chr.10: 95621105-95621105 on Build GRCh38
rs957992629 Chr.10: 95621202-95621202 on Build GRCh38
rs765958522 Chr.10: 95621228-95621228 on Build GRCh38
rs537411575 Chr.10: 95621115-95621115 on Build GRCh38
rs150526956 Chr.10: 95621184-95621184 on Build GRCh38
rs753532076 Chr.10: 95621055-95621055 on Build GRCh38
rs762724008 Chr.10: 95621297-95621297 on Build GRCh38
rs772376087 Chr.10: 95621186-95621186 on Build GRCh38
rs753823851 Chr.10: 95621135-95621135 on Build GRCh38
rs769705702 Chr.10: 95621291-95621291 on Build GRCh38
rs778369085 Chr.10: 95621087-95621087 on Build GRCh38
rs548657482 Chr.10: 95621008-95621008 on Build GRCh38
rs181340553 Chr.10: 95620999-95620999 on Build GRCh38
rs371225175 Chr.10: 95621015-95621015 on Build GRCh38
rs746165671 Chr.10: 95621269-95621269 on Build GRCh38
rs770247123 Chr.10: 95621118-95621118 on Build GRCh38
rs771965130 Chr.10: 95621163-95621163 on Build GRCh38
rs765122458 Chr.10: 95621140-95621140 on Build GRCh38
rs374652686 Chr.10: 95621016-95621016 on Build GRCh38
rs913610798 Chr.10: 95621013-95621013 on Build GRCh38
rs764145947 Chr.10: 95621051-95621051 on Build GRCh38

View Full Product Details