Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58708408-58708958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYAT
Entrez Gene ID
10249
Gene Name
glycine-N-acyltransferase
Gene Aliases
ACGNAT, CAT, GAT
UniGene
Hs.145384
Gene Chromosome Location
Chr.11: 58708757-58731974 on Build GRCh38
Transcripts
NM_201648.2 XM_017017087.1

SNP Detail

SNP ID Chromosome Location
rs1026098957 Chr.11: 58708452-58708452 on Build GRCh38
rs528787282 Chr.11: 58708882-58708882 on Build GRCh38
rs1002455061 Chr.11: 58708539-58708539 on Build GRCh38
rs925292482 Chr.11: 58708846-58708846 on Build GRCh38
rs1000916051 Chr.11: 58708908-58708908 on Build GRCh38
rs996651923 Chr.11: 58708927-58708927 on Build GRCh38
rs1033935346 Chr.11: 58708556-58708556 on Build GRCh38
rs115909531 Chr.11: 58708739-58708739 on Build GRCh38
rs1005983716 Chr.11: 58708720-58708720 on Build GRCh38
rs1013405647 Chr.11: 58708740-58708740 on Build GRCh38
rs908232631 Chr.11: 58708730-58708730 on Build GRCh38
rs182586510 Chr.11: 58708713-58708713 on Build GRCh38
rs993146643 Chr.11: 58708450-58708450 on Build GRCh38
rs1032847608 Chr.11: 58708918-58708918 on Build GRCh38
rs944219280 Chr.11: 58708609-58708609 on Build GRCh38
rs957170238 Chr.11: 58708920-58708920 on Build GRCh38
rs925261643 Chr.11: 58708567-58708567 on Build GRCh38
rs1013871608 Chr.11: 58708863-58708863 on Build GRCh38
rs143895466 Chr.11: 58708911-58708911 on Build GRCh38
rs552755486 Chr.11: 58708716-58708716 on Build GRCh38
rs977937880 Chr.11: 58708812-58708812 on Build GRCh38
rs897393643 Chr.11: 58708684-58708684 on Build GRCh38
rs752730329 Chr.11: 58708435-58708435 on Build GRCh38
rs190872329 Chr.11: 58708565-58708565 on Build GRCh38
rs574545110 Chr.11: 58708780-58708780 on Build GRCh38
rs763767270 Chr.11: 58708757-58708757 on Build GRCh38
rs1041719466 Chr.11: 58708640-58708640 on Build GRCh38
rs561678700 Chr.11: 58708538-58708538 on Build GRCh38
rs979955078 Chr.11: 58708643-58708643 on Build GRCh38
rs1055357299 Chr.11: 58708794-58708794 on Build GRCh38
rs917821939 Chr.11: 58708534-58708534 on Build GRCh38
rs889988080 Chr.11: 58708596-58708596 on Build GRCh38
rs550814657 Chr.11: 58708464-58708464 on Build GRCh38
rs756876305 Chr.11: 58708906-58708906 on Build GRCh38
rs975265275 Chr.11: 58708696-58708696 on Build GRCh38
rs540204843 Chr.11: 58708507-58708507 on Build GRCh38
rs1049764662 Chr.11: 58708586-58708586 on Build GRCh38
rs985989588 Chr.11: 58708892-58708892 on Build GRCh38
rs546004022 Chr.11: 58708458-58708458 on Build GRCh38
rs958022275 Chr.11: 58708864-58708864 on Build GRCh38
rs79722829 Chr.11: 58708712-58708712 on Build GRCh38
rs959227084 Chr.11: 58708519-58708519 on Build GRCh38
rs1041232216 Chr.11: 58708914-58708914 on Build GRCh38
rs936758162 Chr.11: 58708572-58708572 on Build GRCh38
rs570732000 Chr.11: 58708649-58708649 on Build GRCh38
rs945398074 Chr.11: 58708558-58708558 on Build GRCh38
rs1033431805 Chr.11: 58708735-58708735 on Build GRCh38
rs986464638 Chr.11: 58708717-58708717 on Build GRCh38
rs933663637 Chr.11: 58708441-58708441 on Build GRCh38
rs938168463 Chr.11: 58708774-58708774 on Build GRCh38
rs751246298 Chr.11: 58708884-58708884 on Build GRCh38
rs992525074 Chr.11: 58708523-58708523 on Build GRCh38
rs573107774 Chr.11: 58708495-58708495 on Build GRCh38
rs1013607868 Chr.11: 58708582-58708582 on Build GRCh38
rs544321416 Chr.11: 58708642-58708642 on Build GRCh38
rs966802131 Chr.11: 58708594-58708594 on Build GRCh38
rs530397512 Chr.11: 58708513-58708513 on Build GRCh38
rs1025199712 Chr.11: 58708754-58708754 on Build GRCh38
rs911293847 Chr.11: 58708898-58708898 on Build GRCh38
rs568102282 Chr.11: 58708923-58708923 on Build GRCh38

View Full Product Details