Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 125076404-125076904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AACS
Entrez Gene ID
65985
Gene Name
acetoacetyl-CoA synthetase
Gene Aliases
ACSF1, SUR-5
UniGene
Hs.656073
Gene Chromosome Location
Chr.12: 125065367-125143333 on Build GRCh38
Transcripts
NM_001319839.1 NM_001319840.1 NM_023928.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1181132 c.251C>A p.S84* Chr.12: 125076504-125076504 on Build GRCh38
4643667 c.316C>A p.L106M Chr.12: 125076569-125076569 on Build GRCh38
4621499 c.292C>T p.R98W Chr.12: 125076545-125076545 on Build GRCh38
936805 c.301T>C p.Y101H Chr.12: 125076554-125076554 on Build GRCh38
4827804 c.352A>G p.I118V Chr.12: 125076605-125076605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200646490 Chr.12: 125076504-125076504 on Build GRCh38
rs759470993 Chr.12: 125076792-125076792 on Build GRCh38
rs200709074 Chr.12: 125076512-125076512 on Build GRCh38
rs747008072 Chr.12: 125076499-125076499 on Build GRCh38
rs966202611 Chr.12: 125076827-125076827 on Build GRCh38
rs188762368 Chr.12: 125076720-125076720 on Build GRCh38
rs368032917 Chr.12: 125076514-125076514 on Build GRCh38
rs376488677 Chr.12: 125076545-125076545 on Build GRCh38
rs752957982 Chr.12: 125076485-125076485 on Build GRCh38
rs986407802 Chr.12: 125076578-125076578 on Build GRCh38
rs760037809 Chr.12: 125076539-125076539 on Build GRCh38
rs764177799 Chr.12: 125076556-125076556 on Build GRCh38
rs574232116 Chr.12: 125076474-125076474 on Build GRCh38
rs751807542 Chr.12: 125076566-125076566 on Build GRCh38
rs183422923 Chr.12: 125076682-125076682 on Build GRCh38
rs539845663 Chr.12: 125076768-125076768 on Build GRCh38
rs546461862 Chr.12: 125076455-125076455 on Build GRCh38
rs192532596 Chr.12: 125076476-125076476 on Build GRCh38
rs138181368 Chr.12: 125076505-125076505 on Build GRCh38
rs751714367 Chr.12: 125076634-125076634 on Build GRCh38
rs777886245 Chr.12: 125076490-125076490 on Build GRCh38
rs539389583 Chr.12: 125076646-125076646 on Build GRCh38
rs752483354 Chr.12: 125076555-125076555 on Build GRCh38
rs966484916 Chr.12: 125076632-125076632 on Build GRCh38
rs775525364 Chr.12: 125076624-125076624 on Build GRCh38
rs769070014 Chr.12: 125076599-125076599 on Build GRCh38
rs201757882 Chr.12: 125076656-125076656 on Build GRCh38
rs776412393 Chr.12: 125076748-125076748 on Build GRCh38
rs759611953 Chr.12: 125076548-125076548 on Build GRCh38
rs759523888 Chr.12: 125076614-125076614 on Build GRCh38
rs775876225 Chr.12: 125076515-125076515 on Build GRCh38
rs924844047 Chr.12: 125076851-125076851 on Build GRCh38
rs769651393 Chr.12: 125076866-125076866 on Build GRCh38
rs780637787 Chr.12: 125076576-125076576 on Build GRCh38
rs772683047 Chr.12: 125076602-125076602 on Build GRCh38
rs549236885 Chr.12: 125076527-125076527 on Build GRCh38
rs773932854 Chr.12: 125076451-125076451 on Build GRCh38
rs564583584 Chr.12: 125076820-125076820 on Build GRCh38
rs1012824930 Chr.12: 125076467-125076467 on Build GRCh38
rs199815098 Chr.12: 125076604-125076604 on Build GRCh38
rs145554555 Chr.12: 125076749-125076749 on Build GRCh38
rs753830971 Chr.12: 125076468-125076468 on Build GRCh38
rs965658577 Chr.12: 125076670-125076670 on Build GRCh38
rs1004458517 Chr.12: 125076698-125076698 on Build GRCh38
rs77474358 Chr.12: 125076572-125076572 on Build GRCh38
rs762074712 Chr.12: 125076455-125076456 on Build GRCh38
rs1043571032 Chr.12: 125076872-125076872 on Build GRCh38
rs749580143 Chr.12: 125076579-125076579 on Build GRCh38
rs376860308 Chr.12: 125076445-125076445 on Build GRCh38
rs1022498015 Chr.12: 125076626-125076626 on Build GRCh38
rs772650530 Chr.12: 125076456-125076456 on Build GRCh38
rs374742376 Chr.12: 125076454-125076454 on Build GRCh38
rs939315343 Chr.12: 125076790-125076790 on Build GRCh38
rs746735185 Chr.12: 125076603-125076603 on Build GRCh38
rs578251529 Chr.12: 125076627-125076627 on Build GRCh38
rs747818132 Chr.12: 125076517-125076517 on Build GRCh38
rs200054535 Chr.12: 125076582-125076582 on Build GRCh38
rs572464685 Chr.12: 125076763-125076763 on Build GRCh38
rs1019090228 Chr.12: 125076880-125076880 on Build GRCh38
rs746503499 Chr.12: 125076662-125076662 on Build GRCh38
rs756221181 Chr.12: 125076653-125076653 on Build GRCh38
rs1037493988 Chr.12: 125076760-125076760 on Build GRCh38
rs1041870742 Chr.12: 125076439-125076439 on Build GRCh38
rs112317455 Chr.12: 125076739-125076739 on Build GRCh38
rs763961232 Chr.12: 125076630-125076630 on Build GRCh38
rs369332014 Chr.12: 125076573-125076573 on Build GRCh38
rs777141546 Chr.12: 125076443-125076443 on Build GRCh38
rs753959391 Chr.12: 125076454-125076455 on Build GRCh38
rs770538695 Chr.12: 125076694-125076694 on Build GRCh38
rs372247720 Chr.12: 125076526-125076526 on Build GRCh38
rs760257829 Chr.12: 125076463-125076463 on Build GRCh38
rs867522923 Chr.12: 125076695-125076695 on Build GRCh38
rs187205088 Chr.12: 125076464-125076464 on Build GRCh38
rs750458119 Chr.12: 125076652-125076652 on Build GRCh38
rs762065965 Chr.12: 125076645-125076645 on Build GRCh38
rs977985795 Chr.12: 125076848-125076848 on Build GRCh38
rs999299941 Chr.12: 125076879-125076879 on Build GRCh38
rs769804330 Chr.12: 125076621-125076621 on Build GRCh38
rs774854307 Chr.12: 125076518-125076518 on Build GRCh38
rs761441827 Chr.12: 125076453-125076453 on Build GRCh38
rs1012773824 Chr.12: 125076692-125076692 on Build GRCh38
rs199580421 Chr.12: 125076446-125076446 on Build GRCh38
rs894169606 Chr.12: 125076680-125076680 on Build GRCh38
rs953371415 Chr.12: 125076781-125076781 on Build GRCh38
rs149587566 Chr.12: 125076542-125076542 on Build GRCh38
rs765091999 Chr.12: 125076554-125076554 on Build GRCh38
rs758588639 Chr.12: 125076487-125076487 on Build GRCh38
rs954245982 Chr.12: 125076730-125076730 on Build GRCh38
rs11549081 Chr.12: 125076605-125076605 on Build GRCh38

View Full Product Details