Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 128899148-128899547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLT1D1
Entrez Gene ID
144423
Gene Name
glycosyltransferase 1 domain containing 1
Gene Aliases
-
UniGene
Hs.655668
Gene Chromosome Location
Chr.12: 128853323-128984964 on Build GRCh38
Transcripts
NM_144669.2 NR_133646.1 XM_005253553.4 XM_011537956.2 XM_011537957.2 XM_017018855.1 XM_017018856.1 XM_017018857.1 XM_017018858.1 XM_017018859.1 XM_017018860.1 XM_017018861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3457977 c.355G>A p.E119K Chr.12: 128899267-128899267 on Build GRCh38
7299660 c.333C>T p.V111V Chr.12: 128899245-128899245 on Build GRCh38
7299938 c.366A>G p.Q122Q Chr.12: 128899278-128899278 on Build GRCh38
7299661 c.333C>T p.V111V Chr.12: 128899245-128899245 on Build GRCh38
7299939 c.366A>G p.Q122Q Chr.12: 128899278-128899278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001142346 Chr.12: 128899401-128899401 on Build GRCh38
rs751259408 Chr.12: 128899301-128899301 on Build GRCh38
rs903778215 Chr.12: 128899181-128899181 on Build GRCh38
rs1052376995 Chr.12: 128899515-128899515 on Build GRCh38
rs71449371 Chr.12: 128899178-128899178 on Build GRCh38
rs1033949490 Chr.12: 128899401-128899401 on Build GRCh38
rs975679769 Chr.12: 128899509-128899509 on Build GRCh38
rs768702559 Chr.12: 128899246-128899246 on Build GRCh38
rs1056610310 Chr.12: 128899396-128899396 on Build GRCh38
rs561284251 Chr.12: 128899469-128899469 on Build GRCh38
rs1047592843 Chr.12: 128899341-128899341 on Build GRCh38
rs151212606 Chr.12: 128899304-128899306 on Build GRCh38
rs756896584 Chr.12: 128899308-128899308 on Build GRCh38
rs369236670 Chr.12: 128899308-128899310 on Build GRCh38
rs1018460033 Chr.12: 128899336-128899336 on Build GRCh38
rs770083960 Chr.12: 128899387-128899387 on Build GRCh38
rs145677630 Chr.12: 128899243-128899243 on Build GRCh38
rs575407792 Chr.12: 128899350-128899350 on Build GRCh38
rs771277092 Chr.12: 128899208-128899208 on Build GRCh38
rs777054537 Chr.12: 128899240-128899240 on Build GRCh38
rs775289063 Chr.12: 128899240-128899240 on Build GRCh38
rs933993547 Chr.12: 128899517-128899518 on Build GRCh38
rs755869742 Chr.12: 128899235-128899235 on Build GRCh38
rs776013604 Chr.12: 128899463-128899463 on Build GRCh38
rs540319711 Chr.12: 128899470-128899470 on Build GRCh38
rs772298902 Chr.12: 128899257-128899257 on Build GRCh38
rs374783690 Chr.12: 128899174-128899174 on Build GRCh38
rs749508368 Chr.12: 128899241-128899241 on Build GRCh38
rs199593627 Chr.12: 128899303-128899304 on Build GRCh38
rs916662853 Chr.12: 128899400-128899400 on Build GRCh38
rs372745310 Chr.12: 128899236-128899236 on Build GRCh38
rs974153991 Chr.12: 128899349-128899349 on Build GRCh38
rs774637504 Chr.12: 128899252-128899252 on Build GRCh38
rs764576955 Chr.12: 128899330-128899332 on Build GRCh38
rs963802257 Chr.12: 128899342-128899342 on Build GRCh38
rs756098564 Chr.12: 128899337-128899337 on Build GRCh38
rs147734790 Chr.12: 128899280-128899280 on Build GRCh38
rs983702552 Chr.12: 128899381-128899381 on Build GRCh38
rs565041836 Chr.12: 128899306-128899306 on Build GRCh38
rs141096725 Chr.12: 128899281-128899281 on Build GRCh38
rs951048242 Chr.12: 128899353-128899353 on Build GRCh38
rs756768487 Chr.12: 128899223-128899223 on Build GRCh38
rs761305417 Chr.12: 128899265-128899265 on Build GRCh38
rs201205835 Chr.12: 128899261-128899261 on Build GRCh38
rs745830583 Chr.12: 128899234-128899234 on Build GRCh38
rs781071478 Chr.12: 128899230-128899230 on Build GRCh38
rs748136056 Chr.12: 128899253-128899253 on Build GRCh38
rs147860736 Chr.12: 128899473-128899473 on Build GRCh38
rs35458497 Chr.12: 128899423-128899424 on Build GRCh38
rs939544860 Chr.12: 128899391-128899391 on Build GRCh38
rs188637203 Chr.12: 128899178-128899178 on Build GRCh38
rs113191096 Chr.12: 128899388-128899388 on Build GRCh38
rs1033245566 Chr.12: 128899479-128899479 on Build GRCh38
rs12306088 Chr.12: 128899226-128899226 on Build GRCh38
rs750352168 Chr.12: 128899323-128899323 on Build GRCh38
rs948126674 Chr.12: 128899408-128899408 on Build GRCh38
rs763647472 Chr.12: 128899212-128899212 on Build GRCh38
rs767051067 Chr.12: 128899322-128899322 on Build GRCh38
rs999485115 Chr.12: 128899218-128899218 on Build GRCh38
rs895492277 Chr.12: 128899453-128899453 on Build GRCh38
rs546343845 Chr.12: 128899300-128899300 on Build GRCh38
rs750979621 Chr.12: 128899215-128899215 on Build GRCh38
rs192957423 Chr.12: 128899179-128899179 on Build GRCh38

View Full Product Details