Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42273164-42273672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GANC
Entrez Gene ID
2595
Gene Name
glucosidase alpha, neutral C
Gene Aliases
-
UniGene
Hs.730806
Gene Chromosome Location
Chr.15: 42273658-42353666 on Build GRCh38
Transcripts
NM_001301409.1 NM_001110503.2 NM_001286487.1 NM_015497.4 XM_005254287.1 XM_011521443.2 XM_011521444.2

Gene Symbol
TMEM87A
Entrez Gene ID
25963
Gene Name
transmembrane protein 87A
Gene Aliases
-
UniGene
Hs.511138
Gene Chromosome Location
Chr.15: 42210452-42273584 on Build GRCh38
Transcripts
NM_001301409.1 NM_001110503.2 NM_001286487.1 NM_015497.4 XM_005254287.1 XM_011521443.2 XM_011521444.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7144431 c.117G>T p.R39R Chr.15: 42273282-42273282 on Build GRCh38
6241926 c.46C>A p.L16I Chr.15: 42273353-42273353 on Build GRCh38
5362957 c.35T>A p.V12D Chr.15: 42273364-42273364 on Build GRCh38
7226246 c.9G>T p.A3A Chr.15: 42273390-42273390 on Build GRCh38
5363155 c.69A>G p.P23P Chr.15: 42273330-42273330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543349213 Chr.15: 42273220-42273220 on Build GRCh38
rs372089086 Chr.15: 42273221-42273221 on Build GRCh38
rs770495201 Chr.15: 42273439-42273439 on Build GRCh38
rs755510826 Chr.15: 42273210-42273210 on Build GRCh38
rs200378857 Chr.15: 42273408-42273408 on Build GRCh38
rs771153563 Chr.15: 42273346-42273346 on Build GRCh38
rs750851583 Chr.15: 42273235-42273235 on Build GRCh38
rs753354375 Chr.15: 42273233-42273233 on Build GRCh38
rs756291514 Chr.15: 42273404-42273404 on Build GRCh38
rs1026020507 Chr.15: 42273228-42273228 on Build GRCh38
rs897823254 Chr.15: 42273364-42273364 on Build GRCh38
rs926755000 Chr.15: 42273514-42273518 on Build GRCh38
rs909784562 Chr.15: 42273643-42273643 on Build GRCh38
rs375802819 Chr.15: 42273326-42273326 on Build GRCh38
rs775527588 Chr.15: 42273266-42273266 on Build GRCh38
rs377381365 Chr.15: 42273215-42273215 on Build GRCh38
rs776482461 Chr.15: 42273359-42273359 on Build GRCh38
rs114353354 Chr.15: 42273192-42273192 on Build GRCh38
rs763472215 Chr.15: 42273229-42273229 on Build GRCh38
rs528432447 Chr.15: 42273557-42273557 on Build GRCh38
rs758707562 Chr.15: 42273236-42273236 on Build GRCh38
rs941031091 Chr.15: 42273470-42273470 on Build GRCh38
rs879609337 Chr.15: 42273189-42273189 on Build GRCh38
rs374934765 Chr.15: 42273234-42273234 on Build GRCh38
rs766697929 Chr.15: 42273301-42273301 on Build GRCh38
rs766104790 Chr.15: 42273237-42273241 on Build GRCh38
rs950831138 Chr.15: 42273647-42273647 on Build GRCh38
rs773999033 Chr.15: 42273442-42273442 on Build GRCh38
rs1033931920 Chr.15: 42273539-42273539 on Build GRCh38
rs762737989 Chr.15: 42273546-42273547 on Build GRCh38
rs984899271 Chr.15: 42273649-42273649 on Build GRCh38
rs535472070 Chr.15: 42273337-42273337 on Build GRCh38
rs531864227 Chr.15: 42273544-42273544 on Build GRCh38
rs755496249 Chr.15: 42273312-42273312 on Build GRCh38
rs896888231 Chr.15: 42273508-42273508 on Build GRCh38
rs756203231 Chr.15: 42273327-42273327 on Build GRCh38
rs966755067 Chr.15: 42273572-42273572 on Build GRCh38
rs578220746 Chr.15: 42273410-42273410 on Build GRCh38
rs779188133 Chr.15: 42273413-42273413 on Build GRCh38
rs939457769 Chr.15: 42273206-42273206 on Build GRCh38
rs550267219 Chr.15: 42273621-42273621 on Build GRCh38
rs963765090 Chr.15: 42273237-42273237 on Build GRCh38
rs932476382 Chr.15: 42273619-42273619 on Build GRCh38
rs929303818 Chr.15: 42273526-42273526 on Build GRCh38
rs770209829 Chr.15: 42273261-42273261 on Build GRCh38
rs775232794 Chr.15: 42273225-42273225 on Build GRCh38
rs570365135 Chr.15: 42273630-42273630 on Build GRCh38
rs878875529 Chr.15: 42273205-42273205 on Build GRCh38
rs747128279 Chr.15: 42273216-42273216 on Build GRCh38
rs202174333 Chr.15: 42273307-42273307 on Build GRCh38
rs888397674 Chr.15: 42273464-42273464 on Build GRCh38
rs998049880 Chr.15: 42273547-42273547 on Build GRCh38
rs868439842 Chr.15: 42273313-42273313 on Build GRCh38
rs763811018 Chr.15: 42273210-42273210 on Build GRCh38
rs535299959 Chr.15: 42273611-42273611 on Build GRCh38
rs899673613 Chr.15: 42273412-42273412 on Build GRCh38
rs767925572 Chr.15: 42273400-42273400 on Build GRCh38
rs778038926 Chr.15: 42273405-42273405 on Build GRCh38
rs781141360 Chr.15: 42273422-42273422 on Build GRCh38
rs372647656 Chr.15: 42273324-42273324 on Build GRCh38
rs570327484 Chr.15: 42273213-42273213 on Build GRCh38
rs919364524 Chr.15: 42273523-42273523 on Build GRCh38
rs559472323 Chr.15: 42273555-42273555 on Build GRCh38
rs776059456 Chr.15: 42273282-42273282 on Build GRCh38
rs374705120 Chr.15: 42273298-42273298 on Build GRCh38
rs748127499 Chr.15: 42273423-42273423 on Build GRCh38
rs1002409205 Chr.15: 42273532-42273532 on Build GRCh38
rs771713450 Chr.15: 42273265-42273265 on Build GRCh38
rs888073236 Chr.15: 42273249-42273249 on Build GRCh38
rs960846985 Chr.15: 42273616-42273616 on Build GRCh38
rs370752042 Chr.15: 42273223-42273223 on Build GRCh38
rs958019797 Chr.15: 42273387-42273387 on Build GRCh38
rs373908111 Chr.15: 42273394-42273394 on Build GRCh38
rs768573666 Chr.15: 42273217-42273217 on Build GRCh38
rs1050848791 Chr.15: 42273566-42273566 on Build GRCh38
rs772623318 Chr.15: 42273214-42273214 on Build GRCh38
rs764065757 Chr.15: 42273270-42273270 on Build GRCh38
rs992247578 Chr.15: 42273516-42273516 on Build GRCh38
rs867813136 Chr.15: 42273578-42273578 on Build GRCh38
rs368638014 Chr.15: 42273380-42273380 on Build GRCh38
rs17525771 Chr.15: 42273549-42273552 on Build GRCh38
rs905311011 Chr.15: 42273194-42273194 on Build GRCh38
rs549698332 Chr.15: 42273645-42273645 on Build GRCh38
rs977201017 Chr.15: 42273580-42273580 on Build GRCh38
rs201431407 Chr.15: 42273368-42273368 on Build GRCh38
rs760253720 Chr.15: 42273226-42273226 on Build GRCh38
rs745668846 Chr.15: 42273264-42273264 on Build GRCh38
rs368577068 Chr.15: 42273318-42273318 on Build GRCh38
rs2277533 Chr.15: 42273390-42273390 on Build GRCh38
rs769701784 Chr.15: 42273425-42273425 on Build GRCh38
rs563845220 Chr.15: 42273444-42273444 on Build GRCh38
rs1014390854 Chr.15: 42273512-42273512 on Build GRCh38
rs529365545 Chr.15: 42273454-42273454 on Build GRCh38
rs753382605 Chr.15: 42273212-42273212 on Build GRCh38
rs376292282 Chr.15: 42273421-42273421 on Build GRCh38
rs377338968 Chr.15: 42273644-42273644 on Build GRCh38
rs140400607 Chr.15: 42273285-42273285 on Build GRCh38
rs773504077 Chr.15: 42273263-42273263 on Build GRCh38
rs772766614 Chr.15: 42273431-42273431 on Build GRCh38
rs550307753 Chr.15: 42273355-42273357 on Build GRCh38
rs780635972 Chr.15: 42273239-42273239 on Build GRCh38
rs939404076 Chr.15: 42273355-42273355 on Build GRCh38
rs759403180 Chr.15: 42273222-42273222 on Build GRCh38
rs748226584 Chr.15: 42273259-42273259 on Build GRCh38
rs939559927 Chr.15: 42273514-42273514 on Build GRCh38
rs371967261 Chr.15: 42273402-42273402 on Build GRCh38
rs752596908 Chr.15: 42273513-42273513 on Build GRCh38
rs1021346894 Chr.15: 42273561-42273561 on Build GRCh38
rs777493651 Chr.15: 42273211-42273211 on Build GRCh38
rs1021981253 Chr.15: 42273460-42273460 on Build GRCh38
rs760569422 Chr.15: 42273268-42273268 on Build GRCh38
rs776947137 Chr.15: 42273218-42273218 on Build GRCh38
rs967690141 Chr.15: 42273583-42273583 on Build GRCh38
rs555842171 Chr.15: 42273250-42273250 on Build GRCh38
rs773746117 Chr.15: 42273348-42273348 on Build GRCh38
rs1037126938 Chr.15: 42273505-42273505 on Build GRCh38
rs568200203 Chr.15: 42273473-42273473 on Build GRCh38
rs200115858 Chr.15: 42273540-42273541 on Build GRCh38
rs958300404 Chr.15: 42273540-42273540 on Build GRCh38
rs750032382 Chr.15: 42273291-42273291 on Build GRCh38
rs961456056 Chr.15: 42273304-42273304 on Build GRCh38
rs778161696 Chr.15: 42273331-42273331 on Build GRCh38
rs371647438 Chr.15: 42273276-42273276 on Build GRCh38
rs906667336 Chr.15: 42273529-42273529 on Build GRCh38
rs368047226 Chr.15: 42273245-42273245 on Build GRCh38
rs144133599 Chr.15: 42273290-42273290 on Build GRCh38
rs369094214 Chr.15: 42273434-42273434 on Build GRCh38
rs755949366 Chr.15: 42273602-42273602 on Build GRCh38
rs1002408141 Chr.15: 42273595-42273595 on Build GRCh38
rs768540778 Chr.15: 42273354-42273354 on Build GRCh38
rs992759602 Chr.15: 42273618-42273618 on Build GRCh38
rs1025591504 Chr.15: 42273332-42273332 on Build GRCh38
rs751937373 Chr.15: 42273392-42273392 on Build GRCh38
rs762566100 Chr.15: 42273384-42273384 on Build GRCh38
rs372604834 Chr.15: 42273506-42273506 on Build GRCh38
rs572735519 Chr.15: 42273374-42273374 on Build GRCh38

View Full Product Details