Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42307981-42308478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GANC
Entrez Gene ID
2595
Gene Name
glucosidase alpha, neutral C
Gene Aliases
-
UniGene
Hs.730806
Gene Chromosome Location
Chr.15: 42273658-42353666 on Build GRCh38
Transcripts
NM_001301409.1 NM_198141.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6142159 c.651C>G p.F217L Chr.15: 42308247-42308247 on Build GRCh38
2187067 c.640G>A p.G214S Chr.15: 42308236-42308236 on Build GRCh38
2187068 c.640G>T p.G214C Chr.15: 42308236-42308236 on Build GRCh38
2187071 c.721G>A p.G241S Chr.15: 42308317-42308317 on Build GRCh38
5039357 c.626-1G>A p.? Chr.15: 42308221-42308221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769285909 Chr.15: 42308112-42308112 on Build GRCh38
rs985457145 Chr.15: 42308075-42308075 on Build GRCh38
rs772185652 Chr.15: 42308183-42308183 on Build GRCh38
rs185765111 Chr.15: 42308014-42308014 on Build GRCh38
rs764756448 Chr.15: 42308225-42308227 on Build GRCh38
rs1007107946 Chr.15: 42308409-42308409 on Build GRCh38
rs978787785 Chr.15: 42308149-42308149 on Build GRCh38
rs760285749 Chr.15: 42308240-42308240 on Build GRCh38
rs756261460 Chr.15: 42308355-42308355 on Build GRCh38
rs767101665 Chr.15: 42308234-42308234 on Build GRCh38
rs754185846 Chr.15: 42308245-42308245 on Build GRCh38
rs1029906370 Chr.15: 42308042-42308042 on Build GRCh38
rs745953932 Chr.15: 42308182-42308182 on Build GRCh38
rs967258886 Chr.15: 42308020-42308020 on Build GRCh38
rs750901480 Chr.15: 42308360-42308360 on Build GRCh38
rs150417087 Chr.15: 42308261-42308261 on Build GRCh38
rs746176787 Chr.15: 42308003-42308003 on Build GRCh38
rs774955956 Chr.15: 42308319-42308319 on Build GRCh38
rs747979213 Chr.15: 42308191-42308191 on Build GRCh38
rs749829857 Chr.15: 42308173-42308173 on Build GRCh38
rs962744580 Chr.15: 42308120-42308120 on Build GRCh38
rs1016456196 Chr.15: 42308044-42308045 on Build GRCh38
rs770789348 Chr.15: 42308294-42308294 on Build GRCh38
rs145199858 Chr.15: 42308288-42308288 on Build GRCh38
rs767479709 Chr.15: 42308291-42308292 on Build GRCh38
rs767830971 Chr.15: 42308421-42308421 on Build GRCh38
rs1020775048 Chr.15: 42308375-42308375 on Build GRCh38
rs954271341 Chr.15: 42308400-42308400 on Build GRCh38
rs370747535 Chr.15: 42308316-42308316 on Build GRCh38
rs778914756 Chr.15: 42308180-42308180 on Build GRCh38
rs947357925 Chr.15: 42308140-42308141 on Build GRCh38
rs954911158 Chr.15: 42308116-42308116 on Build GRCh38
rs757769027 Chr.15: 42308250-42308250 on Build GRCh38
rs747711947 Chr.15: 42308063-42308063 on Build GRCh38
rs201715151 Chr.15: 42308220-42308220 on Build GRCh38
rs773741729 Chr.15: 42308231-42308231 on Build GRCh38
rs368072322 Chr.15: 42308362-42308362 on Build GRCh38
rs374621298 Chr.15: 42308201-42308201 on Build GRCh38
rs780454796 Chr.15: 42308255-42308255 on Build GRCh38
rs200487161 Chr.15: 42308302-42308302 on Build GRCh38
rs752890631 Chr.15: 42308354-42308355 on Build GRCh38
rs927240107 Chr.15: 42308443-42308443 on Build GRCh38
rs968762256 Chr.15: 42308379-42308379 on Build GRCh38
rs958728492 Chr.15: 42308448-42308448 on Build GRCh38
rs113066307 Chr.15: 42308047-42308047 on Build GRCh38
rs529109463 Chr.15: 42308137-42308137 on Build GRCh38
rs528338285 Chr.15: 42308441-42308441 on Build GRCh38
rs764488546 Chr.15: 42308242-42308242 on Build GRCh38
rs573383850 Chr.15: 42308064-42308064 on Build GRCh38
rs752310663 Chr.15: 42308236-42308236 on Build GRCh38
rs142337004 Chr.15: 42308308-42308308 on Build GRCh38
rs1009925327 Chr.15: 42308304-42308304 on Build GRCh38
rs760375625 Chr.15: 42308342-42308342 on Build GRCh38
rs758454765 Chr.15: 42308254-42308254 on Build GRCh38
rs937291195 Chr.15: 42308258-42308258 on Build GRCh38
rs780365052 Chr.15: 42308188-42308188 on Build GRCh38
rs754204442 Chr.15: 42308353-42308353 on Build GRCh38
rs768987114 Chr.15: 42308273-42308273 on Build GRCh38
rs778280466 Chr.15: 42308172-42308172 on Build GRCh38
rs757757459 Chr.15: 42308177-42308177 on Build GRCh38
rs1017203252 Chr.15: 42308435-42308435 on Build GRCh38
rs114866035 Chr.15: 42308029-42308029 on Build GRCh38
rs937888478 Chr.15: 42308197-42308197 on Build GRCh38
rs770900021 Chr.15: 42308224-42308224 on Build GRCh38
rs978859771 Chr.15: 42308442-42308442 on Build GRCh38
rs199517317 Chr.15: 42308352-42308352 on Build GRCh38
rs116401148 Chr.15: 42308364-42308364 on Build GRCh38
rs76587217 Chr.15: 42308232-42308232 on Build GRCh38

View Full Product Details