Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42389761-42390306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN3
Entrez Gene ID
825
Gene Name
calpain 3
Gene Aliases
CANP3, CANPL3, LGMD2, LGMD2A, nCL-1, p94
UniGene
Hs.143261
Gene Chromosome Location
Chr.15: 42359500-42412317 on Build GRCh38
Transcripts
NM_000070.2 NM_024344.1 NM_173087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6244694 c.838G>T p.G280C Chr.15: 42389989-42389989 on Build GRCh38
6244695 c.577G>T p.G193C Chr.15: 42389989-42389989 on Build GRCh38
7191422 c.915C>A p.D305E Chr.15: 42390066-42390066 on Build GRCh38
7317904 c.629C>T p.S210L Chr.15: 42390041-42390041 on Build GRCh38
7317901 c.890C>T p.S297L Chr.15: 42390041-42390041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767466229 Chr.15: 42389978-42389978 on Build GRCh38
rs566100726 Chr.15: 42390211-42390211 on Build GRCh38
rs1010282102 Chr.15: 42389925-42389925 on Build GRCh38
rs985712258 Chr.15: 42390047-42390047 on Build GRCh38
rs777076120 Chr.15: 42389961-42389961 on Build GRCh38
rs773319272 Chr.15: 42390037-42390037 on Build GRCh38
rs966010074 Chr.15: 42389807-42389807 on Build GRCh38
rs759821346 Chr.15: 42390115-42390115 on Build GRCh38
rs28364427 Chr.15: 42390152-42390152 on Build GRCh38
rs764033551 Chr.15: 42390121-42390121 on Build GRCh38
rs373936933 Chr.15: 42389965-42389965 on Build GRCh38
rs567256305 Chr.15: 42390111-42390111 on Build GRCh38
rs926310716 Chr.15: 42389892-42389892 on Build GRCh38
rs374073070 Chr.15: 42389889-42389889 on Build GRCh38
rs863224966 Chr.15: 42390034-42390037 on Build GRCh38
rs886044545 Chr.15: 42390004-42390005 on Build GRCh38
rs933393545 Chr.15: 42390265-42390265 on Build GRCh38
rs28364425 Chr.15: 42389793-42389793 on Build GRCh38
rs778476064 Chr.15: 42390003-42390003 on Build GRCh38
rs918910759 Chr.15: 42390147-42390147 on Build GRCh38
rs749173640 Chr.15: 42389992-42389992 on Build GRCh38
rs150356488 Chr.15: 42390081-42390081 on Build GRCh38
rs776185666 Chr.15: 42389984-42389986 on Build GRCh38
rs764874721 Chr.15: 42390033-42390033 on Build GRCh38
rs768952783 Chr.15: 42390023-42390023 on Build GRCh38
rs753780627 Chr.15: 42390064-42390064 on Build GRCh38
rs778330298 Chr.15: 42390079-42390079 on Build GRCh38
rs747399155 Chr.15: 42390019-42390019 on Build GRCh38
rs138914944 Chr.15: 42389972-42389972 on Build GRCh38
rs371015074 Chr.15: 42390131-42390131 on Build GRCh38
rs967799484 Chr.15: 42389876-42389876 on Build GRCh38
rs747895099 Chr.15: 42389943-42389943 on Build GRCh38
rs918629110 Chr.15: 42389822-42389822 on Build GRCh38
rs1017640529 Chr.15: 42389798-42389798 on Build GRCh38
rs942177311 Chr.15: 42390228-42390228 on Build GRCh38
rs901966668 Chr.15: 42390240-42390240 on Build GRCh38
rs1017206270 Chr.15: 42390243-42390243 on Build GRCh38
rs765292152 Chr.15: 42389964-42389964 on Build GRCh38
rs187281527 Chr.15: 42390206-42390206 on Build GRCh38
rs750085971 Chr.15: 42390112-42390113 on Build GRCh38
rs575411009 Chr.15: 42389875-42389875 on Build GRCh38
rs766540393 Chr.15: 42389896-42389896 on Build GRCh38
rs201155077 Chr.15: 42390095-42390095 on Build GRCh38
rs1006975937 Chr.15: 42390235-42390235 on Build GRCh38
rs369727850 Chr.15: 42389917-42389917 on Build GRCh38
rs752483058 Chr.15: 42390089-42390089 on Build GRCh38
rs905681564 Chr.15: 42389837-42389837 on Build GRCh38
rs757909241 Chr.15: 42390148-42390148 on Build GRCh38
rs775084606 Chr.15: 42390017-42390017 on Build GRCh38
rs398123150 Chr.15: 42390015-42390016 on Build GRCh38
rs546291253 Chr.15: 42390187-42390187 on Build GRCh38
rs376801349 Chr.15: 42389909-42389909 on Build GRCh38
rs889962606 Chr.15: 42390227-42390227 on Build GRCh38
rs765291508 Chr.15: 42390035-42390035 on Build GRCh38
rs558004763 Chr.15: 42389823-42389823 on Build GRCh38
rs886044104 Chr.15: 42389995-42389995 on Build GRCh38
rs528417986 Chr.15: 42390016-42390016 on Build GRCh38
rs757855868 Chr.15: 42390086-42390086 on Build GRCh38
rs750516376 Chr.15: 42389862-42389862 on Build GRCh38
rs761475803 Chr.15: 42390139-42390139 on Build GRCh38
rs777125401 Chr.15: 42390027-42390027 on Build GRCh38
rs748651267 Chr.15: 42390094-42390094 on Build GRCh38
rs757200858 Chr.15: 42390066-42390066 on Build GRCh38
rs781522974 Chr.15: 42390092-42390092 on Build GRCh38
rs764617439 Chr.15: 42389957-42389957 on Build GRCh38
rs775813174 Chr.15: 42389941-42389941 on Build GRCh38
rs761257703 Chr.15: 42390000-42390002 on Build GRCh38
rs561216369 Chr.15: 42389913-42389913 on Build GRCh38
rs763112832 Chr.15: 42390110-42390110 on Build GRCh38
rs751780260 Chr.15: 42390055-42390055 on Build GRCh38
rs756786521 Chr.15: 42389998-42389998 on Build GRCh38
rs574822029 Chr.15: 42389915-42389915 on Build GRCh38
rs988031430 Chr.15: 42389835-42389835 on Build GRCh38
rs147337720 Chr.15: 42389981-42389981 on Build GRCh38
rs773547765 Chr.15: 42390101-42390101 on Build GRCh38
rs1045561992 Chr.15: 42389834-42389834 on Build GRCh38
rs370313391 Chr.15: 42390048-42390048 on Build GRCh38
rs565111470 Chr.15: 42390082-42390082 on Build GRCh38
rs929980918 Chr.15: 42389833-42389860 on Build GRCh38
rs750271830 Chr.15: 42390144-42390144 on Build GRCh38
rs374702796 Chr.15: 42390063-42390063 on Build GRCh38
rs368037925 Chr.15: 42389932-42389932 on Build GRCh38
rs78369269 Chr.15: 42390090-42390090 on Build GRCh38
rs28364426 Chr.15: 42389833-42389833 on Build GRCh38
rs929547663 Chr.15: 42389918-42389919 on Build GRCh38
rs1028694587 Chr.15: 42390217-42390217 on Build GRCh38
rs377524046 Chr.15: 42390106-42390106 on Build GRCh38
rs761211705 Chr.15: 42389944-42389944 on Build GRCh38
rs755930521 Chr.15: 42389986-42389986 on Build GRCh38
rs541406793 Chr.15: 42389802-42389802 on Build GRCh38
rs777872328 Chr.15: 42389987-42389987 on Build GRCh38
rs774783950 Chr.15: 42390113-42390113 on Build GRCh38
rs929604853 Chr.15: 42390150-42390150 on Build GRCh38
rs200460321 Chr.15: 42390143-42390143 on Build GRCh38
rs1041746198 Chr.15: 42390168-42390168 on Build GRCh38

View Full Product Details