Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42396722-42397220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN3
Entrez Gene ID
825
Gene Name
calpain 3
Gene Aliases
CANP3, CANPL3, LGMD2, LGMD2A, nCL-1, p94
UniGene
Hs.143261
Gene Chromosome Location
Chr.15: 42359500-42412317 on Build GRCh38
Transcripts
NM_000070.2 NM_024344.1 NM_173087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5618098 c.1179G>T p.E393D Chr.15: 42396863-42396863 on Build GRCh38
6244698 c.1135G>T p.V379L Chr.15: 42396819-42396819 on Build GRCh38
5740303 c.1157G>A p.R386H Chr.15: 42396841-42396841 on Build GRCh38
1247323 c.926A>C p.E309A Chr.15: 42396871-42396871 on Build GRCh38
6244699 c.874G>T p.V292L Chr.15: 42396819-42396819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3803342 Chr.15: 42397040-42397040 on Build GRCh38
rs779860644 Chr.15: 42396924-42396924 on Build GRCh38
rs3803343 Chr.15: 42396752-42396752 on Build GRCh38
rs370355559 Chr.15: 42396872-42396872 on Build GRCh38
rs1041440400 Chr.15: 42396771-42396771 on Build GRCh38
rs1007130873 Chr.15: 42396903-42396903 on Build GRCh38
rs79097695 Chr.15: 42397170-42397170 on Build GRCh38
rs752046508 Chr.15: 42396871-42396871 on Build GRCh38
rs1034582062 Chr.15: 42397102-42397102 on Build GRCh38
rs776494254 Chr.15: 42396836-42396836 on Build GRCh38
rs770776609 Chr.15: 42396779-42396779 on Build GRCh38
rs879172045 Chr.15: 42396933-42396933 on Build GRCh38
rs377497251 Chr.15: 42396890-42396890 on Build GRCh38
rs1039476293 Chr.15: 42396957-42396957 on Build GRCh38
rs778663080 Chr.15: 42396786-42396786 on Build GRCh38
rs28364467 Chr.15: 42396795-42396795 on Build GRCh38
rs749348916 Chr.15: 42396905-42396910 on Build GRCh38
rs771846628 Chr.15: 42396915-42396915 on Build GRCh38
rs374740787 Chr.15: 42396887-42396887 on Build GRCh38
rs945681899 Chr.15: 42397150-42397150 on Build GRCh38
rs1051798261 Chr.15: 42396846-42396846 on Build GRCh38
rs781402273 Chr.15: 42396760-42396760 on Build GRCh38
rs765747938 Chr.15: 42396845-42396845 on Build GRCh38
rs35842469 Chr.15: 42397055-42397056 on Build GRCh38
rs933928787 Chr.15: 42396968-42396968 on Build GRCh38
rs1033775597 Chr.15: 42397130-42397130 on Build GRCh38
rs923020660 Chr.15: 42397167-42397167 on Build GRCh38
rs28364468 Chr.15: 42397000-42397000 on Build GRCh38
rs886043889 Chr.15: 42396832-42396832 on Build GRCh38
rs981268322 Chr.15: 42397046-42397046 on Build GRCh38
rs750721662 Chr.15: 42396851-42396851 on Build GRCh38
rs904564825 Chr.15: 42396863-42396863 on Build GRCh38
rs1043564246 Chr.15: 42397065-42397065 on Build GRCh38
rs914273580 Chr.15: 42397126-42397126 on Build GRCh38
rs532054168 Chr.15: 42397078-42397078 on Build GRCh38
rs780220983 Chr.15: 42396751-42396751 on Build GRCh38
rs765016398 Chr.15: 42397134-42397134 on Build GRCh38
rs370871737 Chr.15: 42396757-42396757 on Build GRCh38
rs28364470 Chr.15: 42397188-42397188 on Build GRCh38
rs934135114 Chr.15: 42396838-42396838 on Build GRCh38
rs777934714 Chr.15: 42396904-42396904 on Build GRCh38
rs748299263 Chr.15: 42396772-42396772 on Build GRCh38
rs768215038 Chr.15: 42396835-42396835 on Build GRCh38
rs566834040 Chr.15: 42396988-42396988 on Build GRCh38
rs960336658 Chr.15: 42397147-42397147 on Build GRCh38
rs919442493 Chr.15: 42396840-42396840 on Build GRCh38
rs1009388780 Chr.15: 42397050-42397050 on Build GRCh38
rs28364469 Chr.15: 42397120-42397120 on Build GRCh38
rs760121531 Chr.15: 42396814-42396814 on Build GRCh38
rs569412461 Chr.15: 42397118-42397118 on Build GRCh38
rs570748085 Chr.15: 42397152-42397152 on Build GRCh38
rs771938059 Chr.15: 42396800-42396800 on Build GRCh38
rs886042537 Chr.15: 42396877-42396877 on Build GRCh38
rs376273996 Chr.15: 42396841-42396841 on Build GRCh38
rs546144154 Chr.15: 42396962-42396962 on Build GRCh38
rs567188857 Chr.15: 42396852-42396852 on Build GRCh38
rs775453643 Chr.15: 42396801-42396801 on Build GRCh38
rs768203258 Chr.15: 42396992-42396992 on Build GRCh38
rs898493662 Chr.15: 42396986-42396986 on Build GRCh38
rs781241796 Chr.15: 42396879-42396879 on Build GRCh38
rs1016986038 Chr.15: 42396926-42396926 on Build GRCh38
rs1013845586 Chr.15: 42397169-42397169 on Build GRCh38
rs766817296 Chr.15: 42396864-42396864 on Build GRCh38
rs745675826 Chr.15: 42396913-42396913 on Build GRCh38
rs199816913 Chr.15: 42396898-42396898 on Build GRCh38

View Full Product Details