Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42723618-42724126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDAN1
Entrez Gene ID
146059
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
UniGene
Hs.599232
Gene Chromosome Location
Chr.15: 42723557-42737219 on Build GRCh38
Transcripts
NM_138477.2 XM_005254176.4 XM_011521270.2 XM_011521271.2 XM_011521274.2

SNP Detail

SNP ID Chromosome Location
rs769879392 Chr.15: 42723962-42723962 on Build GRCh38
rs1001103522 Chr.15: 42723723-42723723 on Build GRCh38
rs1056947710 Chr.15: 42723951-42723951 on Build GRCh38
rs1011188304 Chr.15: 42723738-42723738 on Build GRCh38
rs961793252 Chr.15: 42723782-42723782 on Build GRCh38
rs758214658 Chr.15: 42724077-42724077 on Build GRCh38
rs11556778 Chr.15: 42723870-42723870 on Build GRCh38
rs1032588940 Chr.15: 42724097-42724097 on Build GRCh38
rs141378204 Chr.15: 42723969-42723969 on Build GRCh38
rs886051154 Chr.15: 42723703-42723703 on Build GRCh38
rs1003283351 Chr.15: 42723759-42723759 on Build GRCh38
rs377716127 Chr.15: 42723798-42723798 on Build GRCh38
rs756388906 Chr.15: 42723905-42723905 on Build GRCh38
rs1012674382 Chr.15: 42723685-42723685 on Build GRCh38
rs530367723 Chr.15: 42723928-42723929 on Build GRCh38
rs894023212 Chr.15: 42723733-42723733 on Build GRCh38
rs765014914 Chr.15: 42723954-42723954 on Build GRCh38
rs886051155 Chr.15: 42723891-42723891 on Build GRCh38
rs1023701782 Chr.15: 42723739-42723740 on Build GRCh38
rs984036128 Chr.15: 42723860-42723860 on Build GRCh38
rs752891192 Chr.15: 42723834-42723834 on Build GRCh38
rs971867501 Chr.15: 42723813-42723813 on Build GRCh38
rs1044189990 Chr.15: 42724048-42724048 on Build GRCh38
rs942785975 Chr.15: 42723665-42723665 on Build GRCh38
rs1004476592 Chr.15: 42723647-42723647 on Build GRCh38
rs530760324 Chr.15: 42723919-42723919 on Build GRCh38
rs369460870 Chr.15: 42723687-42723687 on Build GRCh38
rs915438395 Chr.15: 42724007-42724007 on Build GRCh38
rs182979453 Chr.15: 42723890-42723890 on Build GRCh38
rs750240635 Chr.15: 42724006-42724006 on Build GRCh38
rs774242839 Chr.15: 42723648-42723648 on Build GRCh38
rs751250885 Chr.15: 42723689-42723690 on Build GRCh38
rs187557635 Chr.15: 42723914-42723914 on Build GRCh38
rs528323093 Chr.15: 42723769-42723769 on Build GRCh38
rs975474725 Chr.15: 42723830-42723830 on Build GRCh38
rs865924586 Chr.15: 42723855-42723855 on Build GRCh38
rs192603127 Chr.15: 42723737-42723737 on Build GRCh38
rs563918254 Chr.15: 42723748-42723748 on Build GRCh38
rs1022603666 Chr.15: 42723703-42723703 on Build GRCh38
rs886922339 Chr.15: 42723644-42723644 on Build GRCh38
rs1056159226 Chr.15: 42724104-42724104 on Build GRCh38
rs1033033821 Chr.15: 42723732-42723732 on Build GRCh38
rs939868888 Chr.15: 42723913-42723913 on Build GRCh38
rs931324102 Chr.15: 42723848-42723848 on Build GRCh38
rs80066191 Chr.15: 42723668-42723668 on Build GRCh38
rs531320636 Chr.15: 42723709-42723712 on Build GRCh38
rs904173420 Chr.15: 42724050-42724050 on Build GRCh38
rs111961345 Chr.15: 42724026-42724028 on Build GRCh38
rs935462910 Chr.15: 42723725-42723725 on Build GRCh38
rs573659922 Chr.15: 42723664-42723664 on Build GRCh38
rs563564146 Chr.15: 42724053-42724053 on Build GRCh38
rs557465473 Chr.15: 42723937-42723954 on Build GRCh38
rs904200394 Chr.15: 42723709-42723709 on Build GRCh38
rs572923073 Chr.15: 42723773-42723773 on Build GRCh38
rs965392126 Chr.15: 42723811-42723811 on Build GRCh38
rs895608597 Chr.15: 42723676-42723676 on Build GRCh38
rs1019652367 Chr.15: 42723780-42723780 on Build GRCh38
rs868540622 Chr.15: 42723701-42723701 on Build GRCh38

View Full Product Details