Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42726010-42726521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDAN1
Entrez Gene ID
146059
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
UniGene
Hs.599232
Gene Chromosome Location
Chr.15: 42723557-42737219 on Build GRCh38
Transcripts
NM_138477.2 XM_005254176.4 XM_011521270.2 XM_011521271.2 XM_011521274.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5430747 c.3146C>T p.S1049F Chr.15: 42726368-42726368 on Build GRCh38
1731782 c.3247G>T p.V1083L Chr.15: 42726118-42726118 on Build GRCh38
4054740 c.3210G>T p.L1070L Chr.15: 42726155-42726155 on Build GRCh38
6988624 c.3223G>A p.E1075K Chr.15: 42726142-42726142 on Build GRCh38
1372945 c.3126G>A p.R1042R Chr.15: 42726388-42726388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780892720 Chr.15: 42726281-42726282 on Build GRCh38
rs181942118 Chr.15: 42726217-42726217 on Build GRCh38
rs28661826 Chr.15: 42726361-42726361 on Build GRCh38
rs762992673 Chr.15: 42726171-42726171 on Build GRCh38
rs148832555 Chr.15: 42726415-42726415 on Build GRCh38
rs754699165 Chr.15: 42726079-42726079 on Build GRCh38
rs778218831 Chr.15: 42726098-42726098 on Build GRCh38
rs890690272 Chr.15: 42726255-42726255 on Build GRCh38
rs769679860 Chr.15: 42726368-42726369 on Build GRCh38
rs199584270 Chr.15: 42726174-42726174 on Build GRCh38
rs755256438 Chr.15: 42726353-42726353 on Build GRCh38
rs769605831 Chr.15: 42726164-42726164 on Build GRCh38
rs778622745 Chr.15: 42726439-42726439 on Build GRCh38
rs372636364 Chr.15: 42726440-42726440 on Build GRCh38
rs923254013 Chr.15: 42726277-42726277 on Build GRCh38
rs765932895 Chr.15: 42726061-42726061 on Build GRCh38
rs768551906 Chr.15: 42726273-42726274 on Build GRCh38
rs201911736 Chr.15: 42726193-42726193 on Build GRCh38
rs767955137 Chr.15: 42726196-42726197 on Build GRCh38
rs766862847 Chr.15: 42726401-42726401 on Build GRCh38
rs900718057 Chr.15: 42726040-42726040 on Build GRCh38
rs781379707 Chr.15: 42726359-42726359 on Build GRCh38
rs954699080 Chr.15: 42726410-42726410 on Build GRCh38
rs146289653 Chr.15: 42726379-42726379 on Build GRCh38
rs776165533 Chr.15: 42726389-42726389 on Build GRCh38
rs749845511 Chr.15: 42726103-42726103 on Build GRCh38
rs149704797 Chr.15: 42726362-42726362 on Build GRCh38
rs780702942 Chr.15: 42726081-42726081 on Build GRCh38
rs749031658 Chr.15: 42726280-42726280 on Build GRCh38
rs772202915 Chr.15: 42726442-42726442 on Build GRCh38
rs745816995 Chr.15: 42726372-42726372 on Build GRCh38
rs1010512890 Chr.15: 42726263-42726263 on Build GRCh38
rs371822328 Chr.15: 42726409-42726409 on Build GRCh38
rs897669129 Chr.15: 42726321-42726321 on Build GRCh38
rs372937659 Chr.15: 42726448-42726448 on Build GRCh38
rs767169772 Chr.15: 42726293-42726293 on Build GRCh38
rs754299138 Chr.15: 42726315-42726315 on Build GRCh38
rs374335881 Chr.15: 42726428-42726428 on Build GRCh38
rs760831544 Chr.15: 42726163-42726163 on Build GRCh38
rs371567068 Chr.15: 42726104-42726104 on Build GRCh38
rs758940751 Chr.15: 42726400-42726400 on Build GRCh38
rs894680890 Chr.15: 42726358-42726358 on Build GRCh38
rs1000279491 Chr.15: 42726303-42726303 on Build GRCh38
rs768840475 Chr.15: 42726269-42726269 on Build GRCh38
rs575678260 Chr.15: 42726153-42726153 on Build GRCh38
rs140304348 Chr.15: 42726325-42726325 on Build GRCh38
rs759617371 Chr.15: 42726382-42726383 on Build GRCh38
rs375739663 Chr.15: 42726056-42726056 on Build GRCh38
rs990764211 Chr.15: 42726238-42726238 on Build GRCh38
rs143857276 Chr.15: 42726326-42726326 on Build GRCh38
rs370604256 Chr.15: 42726205-42726205 on Build GRCh38
rs80338697 Chr.15: 42726390-42726390 on Build GRCh38
rs2305085 Chr.15: 42726288-42726288 on Build GRCh38
rs530246239 Chr.15: 42726218-42726218 on Build GRCh38
rs933211406 Chr.15: 42726073-42726073 on Build GRCh38
rs772939236 Chr.15: 42726165-42726165 on Build GRCh38
rs531938639 Chr.15: 42726253-42726253 on Build GRCh38
rs747252024 Chr.15: 42726267-42726267 on Build GRCh38
rs561641913 Chr.15: 42726212-42726212 on Build GRCh38
rs61746356 Chr.15: 42726320-42726320 on Build GRCh38
rs779099184 Chr.15: 42726108-42726108 on Build GRCh38
rs368678391 Chr.15: 42726432-42726432 on Build GRCh38
rs1037783864 Chr.15: 42726204-42726204 on Build GRCh38
rs772408802 Chr.15: 42726138-42726138 on Build GRCh38
rs376238524 Chr.15: 42726184-42726184 on Build GRCh38
rs963590294 Chr.15: 42726445-42726445 on Build GRCh38
rs774431944 Chr.15: 42726375-42726376 on Build GRCh38
rs774154290 Chr.15: 42726365-42726365 on Build GRCh38
rs373329905 Chr.15: 42726167-42726167 on Build GRCh38
rs753169494 Chr.15: 42726093-42726093 on Build GRCh38
rs751403518 Chr.15: 42726285-42726285 on Build GRCh38
rs2305084 Chr.15: 42726493-42726493 on Build GRCh38
rs758423857 Chr.15: 42726211-42726211 on Build GRCh38
rs765554263 Chr.15: 42726394-42726394 on Build GRCh38
rs923285215 Chr.15: 42726035-42726035 on Build GRCh38
rs750362336 Chr.15: 42726052-42726052 on Build GRCh38
rs993247706 Chr.15: 42726182-42726182 on Build GRCh38
rs962968321 Chr.15: 42726498-42726498 on Build GRCh38
rs747056209 Chr.15: 42726352-42726352 on Build GRCh38
rs143281373 Chr.15: 42726486-42726486 on Build GRCh38
rs755062475 Chr.15: 42726414-42726414 on Build GRCh38
rs778306555 Chr.15: 42726201-42726201 on Build GRCh38
rs758062781 Chr.15: 42726172-42726173 on Build GRCh38
rs775710942 Chr.15: 42726437-42726443 on Build GRCh38
rs560414741 Chr.15: 42726247-42726247 on Build GRCh38
rs967403245 Chr.15: 42726248-42726248 on Build GRCh38
rs757822823 Chr.15: 42726202-42726202 on Build GRCh38
rs775092615 Chr.15: 42726295-42726295 on Build GRCh38
rs776881666 Chr.15: 42726148-42726148 on Build GRCh38
rs756312314 Chr.15: 42726434-42726434 on Build GRCh38
rs996376012 Chr.15: 42726074-42726074 on Build GRCh38
rs906185429 Chr.15: 42726335-42726335 on Build GRCh38
rs1004460447 Chr.15: 42726489-42726489 on Build GRCh38
rs772242139 Chr.15: 42726386-42726408 on Build GRCh38
rs745908891 Chr.15: 42726125-42726125 on Build GRCh38
rs146049960 Chr.15: 42726314-42726314 on Build GRCh38
rs184244219 Chr.15: 42726236-42726236 on Build GRCh38
rs929153491 Chr.15: 42726497-42726497 on Build GRCh38
rs748114736 Chr.15: 42726340-42726341 on Build GRCh38
rs758888123 Chr.15: 42726332-42726332 on Build GRCh38
rs559053820 Chr.15: 42726097-42726097 on Build GRCh38
rs751007896 Chr.15: 42726075-42726075 on Build GRCh38
rs80338698 Chr.15: 42726386-42726386 on Build GRCh38
rs371724263 Chr.15: 42726210-42726210 on Build GRCh38
rs201125492 Chr.15: 42726305-42726305 on Build GRCh38
rs760935717 Chr.15: 42726462-42726469 on Build GRCh38
rs779888204 Chr.15: 42726173-42726174 on Build GRCh38
rs1010288682 Chr.15: 42726082-42726082 on Build GRCh38
rs1034605660 Chr.15: 42726346-42726346 on Build GRCh38
rs539031324 Chr.15: 42726449-42726449 on Build GRCh38
rs772114869 Chr.15: 42726378-42726378 on Build GRCh38
rs761419053 Chr.15: 42726468-42726468 on Build GRCh38
rs1041939992 Chr.15: 42726411-42726411 on Build GRCh38
rs370724643 Chr.15: 42726287-42726287 on Build GRCh38
rs752400883 Chr.15: 42726190-42726190 on Build GRCh38
rs61746359 Chr.15: 42726345-42726345 on Build GRCh38
rs977491664 Chr.15: 42726249-42726249 on Build GRCh38
rs958465581 Chr.15: 42726310-42726310 on Build GRCh38
rs1035043369 Chr.15: 42726223-42726223 on Build GRCh38
rs773027053 Chr.15: 42726274-42726274 on Build GRCh38

View Full Product Details