Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43189179-43189675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNDBP1
Entrez Gene ID
23582
Gene Name
cyclin D1 binding protein 1
Gene Aliases
DIP1, GCIP, HHM
UniGene
Hs.36794
Gene Chromosome Location
Chr.15: 43185118-43197177 on Build GRCh38
Transcripts
NM_012142.4 NR_027513.2 NR_027514.2 NR_045998.1 NR_045999.1 XM_006720448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6622979 c.282T>C p.A94A Chr.15: 43189231-43189231 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs72721549 Chr.15: 43189592-43189592 on Build GRCh38
rs916232671 Chr.15: 43189434-43189434 on Build GRCh38
rs374008204 Chr.15: 43189213-43189213 on Build GRCh38
rs761384847 Chr.15: 43189269-43189269 on Build GRCh38
rs988152782 Chr.15: 43189609-43189609 on Build GRCh38
rs776463825 Chr.15: 43189268-43189268 on Build GRCh38
rs1018467916 Chr.15: 43189294-43189294 on Build GRCh38
rs769905849 Chr.15: 43189231-43189231 on Build GRCh38
rs936781653 Chr.15: 43189249-43189249 on Build GRCh38
rs1013866398 Chr.15: 43189487-43189487 on Build GRCh38
rs377745084 Chr.15: 43189290-43189290 on Build GRCh38
rs555786304 Chr.15: 43189414-43189414 on Build GRCh38
rs913988922 Chr.15: 43189279-43189279 on Build GRCh38
rs12916956 Chr.15: 43189330-43189330 on Build GRCh38
rs932881278 Chr.15: 43189373-43189373 on Build GRCh38
rs573139770 Chr.15: 43189316-43189317 on Build GRCh38
rs765379197 Chr.15: 43189234-43189235 on Build GRCh38
rs1005785726 Chr.15: 43189248-43189248 on Build GRCh38
rs914004777 Chr.15: 43189616-43189616 on Build GRCh38
rs969280584 Chr.15: 43189212-43189212 on Build GRCh38
rs745841620 Chr.15: 43189238-43189238 on Build GRCh38
rs143593280 Chr.15: 43189561-43189561 on Build GRCh38
rs543170200 Chr.15: 43189598-43189598 on Build GRCh38
rs115357955 Chr.15: 43189210-43189210 on Build GRCh38
rs967954735 Chr.15: 43189423-43189423 on Build GRCh38
rs771727395 Chr.15: 43189240-43189240 on Build GRCh38
rs1002090215 Chr.15: 43189572-43189572 on Build GRCh38
rs1024792566 Chr.15: 43189543-43189543 on Build GRCh38
rs770716049 Chr.15: 43189562-43189562 on Build GRCh38
rs926464124 Chr.15: 43189522-43189522 on Build GRCh38
rs760444564 Chr.15: 43189256-43189256 on Build GRCh38
rs755369340 Chr.15: 43189307-43189307 on Build GRCh38
rs767180022 Chr.15: 43189618-43189618 on Build GRCh38
rs758945431 Chr.15: 43189284-43189284 on Build GRCh38
rs111861500 Chr.15: 43189627-43189627 on Build GRCh38
rs777864502 Chr.15: 43189332-43189332 on Build GRCh38
rs867528884 Chr.15: 43189233-43189234 on Build GRCh38
rs756137689 Chr.15: 43189326-43189326 on Build GRCh38
rs748227477 Chr.15: 43189227-43189227 on Build GRCh38
rs371159934 Chr.15: 43189296-43189296 on Build GRCh38
rs763832976 Chr.15: 43189259-43189259 on Build GRCh38
rs774251321 Chr.15: 43189234-43189234 on Build GRCh38
rs374174194 Chr.15: 43189308-43189308 on Build GRCh38
rs764859089 Chr.15: 43189271-43189271 on Build GRCh38
rs377643826 Chr.15: 43189309-43189309 on Build GRCh38
rs536019942 Chr.15: 43189280-43189280 on Build GRCh38
rs151237666 Chr.15: 43189622-43189622 on Build GRCh38
rs1036559876 Chr.15: 43189428-43189428 on Build GRCh38
rs150871238 Chr.15: 43189241-43189241 on Build GRCh38
rs762033687 Chr.15: 43189206-43189208 on Build GRCh38
rs113055981 Chr.15: 43189320-43189321 on Build GRCh38
rs774368386 Chr.15: 43189583-43189583 on Build GRCh38
rs775361787 Chr.15: 43189246-43189246 on Build GRCh38
rs980626188 Chr.15: 43189498-43189498 on Build GRCh38
rs1050066073 Chr.15: 43189392-43189392 on Build GRCh38
rs540664090 Chr.15: 43189468-43189468 on Build GRCh38
rs1005816743 Chr.15: 43189651-43189651 on Build GRCh38
rs750756341 Chr.15: 43189238-43189238 on Build GRCh38
rs114395814 Chr.15: 43189421-43189421 on Build GRCh38

View Full Product Details