Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43596205-43596754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKMT1B
Entrez Gene ID
1159
Gene Name
creatine kinase, mitochondrial 1B
Gene Aliases
CKMT, CKMT1, UMTCK
UniGene
Hs.741420
Gene Chromosome Location
Chr.15: 43592857-43599406 on Build GRCh38
Transcripts
NM_020990.4 NR_135748.1 NR_135749.1 NR_135750.1 NR_135751.1 NR_135752.1 XM_005254150.3 XM_011521194.1 XM_011521195.2 XM_011521196.1 XM_011521197.2 XM_011521198.1 XM_011521199.2 XM_017021902.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7092066 c.837G>A p.M279I Chr.15: 43596492-43596492 on Build GRCh38
140742 c.815C>T p.S272F Chr.15: 43596470-43596470 on Build GRCh38
4939754 c.866G>A p.G289D Chr.15: 43596521-43596521 on Build GRCh38
4387585 c.865G>C p.G289R Chr.15: 43596520-43596520 on Build GRCh38
4148779 c.756C>G p.H252Q Chr.15: 43596411-43596411 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs547616289 Chr.15: 43596692-43596692 on Build GRCh38
rs750358938 Chr.15: 43596498-43596498 on Build GRCh38
rs764221006 Chr.15: 43596497-43596497 on Build GRCh38
rs927408121 Chr.15: 43596364-43596364 on Build GRCh38
rs564165376 Chr.15: 43596665-43596665 on Build GRCh38
rs760943751 Chr.15: 43596582-43596582 on Build GRCh38
rs748834288 Chr.15: 43596553-43596553 on Build GRCh38
rs183186835 Chr.15: 43596709-43596709 on Build GRCh38
rs756932011 Chr.15: 43596391-43596391 on Build GRCh38
rs376120318 Chr.15: 43596545-43596545 on Build GRCh38
rs781025329 Chr.15: 43596399-43596399 on Build GRCh38
rs766182182 Chr.15: 43596613-43596613 on Build GRCh38
rs754845862 Chr.15: 43596532-43596532 on Build GRCh38
rs199826616 Chr.15: 43596491-43596491 on Build GRCh38
rs776227809 Chr.15: 43596239-43596239 on Build GRCh38
rs968688131 Chr.15: 43596624-43596624 on Build GRCh38
rs554099912 Chr.15: 43596517-43596517 on Build GRCh38
rs201489142 Chr.15: 43596574-43596574 on Build GRCh38
rs751567706 Chr.15: 43596618-43596618 on Build GRCh38
rs572277175 Chr.15: 43596652-43596652 on Build GRCh38
rs778716925 Chr.15: 43596415-43596417 on Build GRCh38
rs780970254 Chr.15: 43596544-43596544 on Build GRCh38
rs77834292 Chr.15: 43596654-43596654 on Build GRCh38
rs747668618 Chr.15: 43596261-43596261 on Build GRCh38
rs113292887 Chr.15: 43596679-43596679 on Build GRCh38
rs770387733 Chr.15: 43596264-43596264 on Build GRCh38
rs779766866 Chr.15: 43596367-43596367 on Build GRCh38
rs955092272 Chr.15: 43596272-43596272 on Build GRCh38
rs769477880 Chr.15: 43596450-43596450 on Build GRCh38
rs759618789 Chr.15: 43596304-43596304 on Build GRCh38
rs773878598 Chr.15: 43596265-43596265 on Build GRCh38
rs749828844 Chr.15: 43596460-43596460 on Build GRCh38
rs979204293 Chr.15: 43596637-43596637 on Build GRCh38
rs768203090 Chr.15: 43596236-43596236 on Build GRCh38
rs754614855 Chr.15: 43596398-43596398 on Build GRCh38
rs746837384 Chr.15: 43596376-43596376 on Build GRCh38
rs531998037 Chr.15: 43596287-43596287 on Build GRCh38
rs937182282 Chr.15: 43596416-43596416 on Build GRCh38
rs189827985 Chr.15: 43596576-43596576 on Build GRCh38
rs187911665 Chr.15: 43596717-43596717 on Build GRCh38
rs751276442 Chr.15: 43596518-43596518 on Build GRCh38
rs532632372 Chr.15: 43596666-43596666 on Build GRCh38
rs767731362 Chr.15: 43596326-43596326 on Build GRCh38
rs768877088 Chr.15: 43596585-43596585 on Build GRCh38
rs756206991 Chr.15: 43596333-43596333 on Build GRCh38
rs1057333279 Chr.15: 43596438-43596438 on Build GRCh38
rs771946559 Chr.15: 43596550-43596551 on Build GRCh38
rs746208388 Chr.15: 43596572-43596572 on Build GRCh38
rs548911205 Chr.15: 43596340-43596340 on Build GRCh38
rs149230297 Chr.15: 43596479-43596479 on Build GRCh38
rs775737368 Chr.15: 43596492-43596492 on Build GRCh38
rs1020515479 Chr.15: 43596583-43596583 on Build GRCh38
rs1047042470 Chr.15: 43596464-43596464 on Build GRCh38
rs771315137 Chr.15: 43596461-43596461 on Build GRCh38
rs555262009 Chr.15: 43596605-43596605 on Build GRCh38
rs773825047 Chr.15: 43596456-43596456 on Build GRCh38
rs766693799 Chr.15: 43596282-43596282 on Build GRCh38
rs762738855 Chr.15: 43596512-43596512 on Build GRCh38
rs568638041 Chr.15: 43596343-43596343 on Build GRCh38
rs1034360770 Chr.15: 43596639-43596639 on Build GRCh38
rs772232119 Chr.15: 43596490-43596490 on Build GRCh38
rs989039286 Chr.15: 43596314-43596314 on Build GRCh38
rs146365795 Chr.15: 43596546-43596546 on Build GRCh38
rs997950497 Chr.15: 43596507-43596507 on Build GRCh38
rs748869600 Chr.15: 43596313-43596314 on Build GRCh38
rs745624577 Chr.15: 43596461-43596461 on Build GRCh38
rs752731710 Chr.15: 43596329-43596329 on Build GRCh38
rs774511453 Chr.15: 43596465-43596465 on Build GRCh38
rs747615641 Chr.15: 43596423-43596423 on Build GRCh38
rs371987094 Chr.15: 43596356-43596356 on Build GRCh38
rs541687320 Chr.15: 43596589-43596589 on Build GRCh38
rs555737933 Chr.15: 43596563-43596563 on Build GRCh38
rs374661313 Chr.15: 43596555-43596555 on Build GRCh38
rs546319506 Chr.15: 43596670-43596670 on Build GRCh38
rs760710877 Chr.15: 43596495-43596495 on Build GRCh38
rs763355494 Chr.15: 43596281-43596281 on Build GRCh38

View Full Product Details