Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43640422-43640919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER2
Entrez Gene ID
117155
Gene Name
cation channel sperm associated 2
Gene Aliases
-
UniGene
Hs.662284
Gene Chromosome Location
Chr.15: 43630562-43648845 on Build GRCh38
Transcripts
NM_001282309.1 NM_001282310.1 NM_054020.3 NM_172095.2 NR_110319.1 XM_017021904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5637369 c.417A>G p.L139L Chr.15: 43640468-43640468 on Build GRCh38
416921 c.462G>A p.L154L Chr.15: 43640423-43640423 on Build GRCh38
962049 c.448G>T p.A150S Chr.15: 43640437-43640437 on Build GRCh38
5383373 c.410_411CC>TT p.T137I Chr.15: 43640474-43640475 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373002988 Chr.15: 43640834-43640834 on Build GRCh38
rs1039685521 Chr.15: 43640870-43640870 on Build GRCh38
rs769438098 Chr.15: 43640480-43640480 on Build GRCh38
rs768576630 Chr.15: 43640463-43640463 on Build GRCh38
rs1036742084 Chr.15: 43640792-43640792 on Build GRCh38
rs778939044 Chr.15: 43640449-43640449 on Build GRCh38
rs779172294 Chr.15: 43640535-43640535 on Build GRCh38
rs543272456 Chr.15: 43640586-43640586 on Build GRCh38
rs773102038 Chr.15: 43640488-43640488 on Build GRCh38
rs754276323 Chr.15: 43640530-43640530 on Build GRCh38
rs375184195 Chr.15: 43640501-43640501 on Build GRCh38
rs943577477 Chr.15: 43640869-43640869 on Build GRCh38
rs914378053 Chr.15: 43640649-43640649 on Build GRCh38
rs969896399 Chr.15: 43640657-43640657 on Build GRCh38
rs746003553 Chr.15: 43640462-43640462 on Build GRCh38
rs750649085 Chr.15: 43640545-43640545 on Build GRCh38
rs761273306 Chr.15: 43640527-43640527 on Build GRCh38
rs980409521 Chr.15: 43640691-43640691 on Build GRCh38
rs928600176 Chr.15: 43640744-43640744 on Build GRCh38
rs569089602 Chr.15: 43640503-43640503 on Build GRCh38
rs956067727 Chr.15: 43640539-43640539 on Build GRCh38
rs372404910 Chr.15: 43640520-43640520 on Build GRCh38
rs113658666 Chr.15: 43640806-43640806 on Build GRCh38
rs34055090 Chr.15: 43640807-43640808 on Build GRCh38
rs781361393 Chr.15: 43640522-43640522 on Build GRCh38
rs560099166 Chr.15: 43640645-43640645 on Build GRCh38
rs35333173 Chr.15: 43640818-43640818 on Build GRCh38
rs377457004 Chr.15: 43640817-43640817 on Build GRCh38
rs887924286 Chr.15: 43640857-43640857 on Build GRCh38
rs573974411 Chr.15: 43640553-43640553 on Build GRCh38
rs558570338 Chr.15: 43640882-43640882 on Build GRCh38
rs1048145320 Chr.15: 43640835-43640835 on Build GRCh38
rs902132060 Chr.15: 43640876-43640876 on Build GRCh38
rs759054585 Chr.15: 43640506-43640506 on Build GRCh38
rs552052362 Chr.15: 43640791-43640791 on Build GRCh38
rs966427638 Chr.15: 43640461-43640461 on Build GRCh38
rs767145307 Chr.15: 43640510-43640510 on Build GRCh38
rs776494497 Chr.15: 43640468-43640468 on Build GRCh38
rs748139549 Chr.15: 43640469-43640469 on Build GRCh38
rs193015077 Chr.15: 43640788-43640788 on Build GRCh38
rs138398505 Chr.15: 43640443-43640443 on Build GRCh38
rs762494160 Chr.15: 43640491-43640491 on Build GRCh38
rs550866834 Chr.15: 43640442-43640442 on Build GRCh38
rs757455081 Chr.15: 43640533-43640533 on Build GRCh38

View Full Product Details