Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 25820503-25821005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC646471
Entrez Gene ID
646471
Gene Name
uncharacterized LOC646471
Gene Aliases
-
UniGene
Hs.727271
Gene Chromosome Location
Chr.1: 25819954-25823606 on Build GRCh38
Transcripts
NR_024498.1 NM_001099625.1 NM_001099626.1 NM_001099627.1 NM_019557.5

Gene Symbol
MTFR1L
Entrez Gene ID
56181
Gene Name
mitochondrial fission regulator 1 like
Gene Aliases
FAM54B, HYST1888, MST116, MSTP116
UniGene
Hs.149305
Gene Chromosome Location
Chr.1: 25819906-25832942 on Build GRCh38
Transcripts
NR_024498.1 NM_001099625.1 NM_001099626.1 NM_001099627.1 NM_019557.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3771704 c.20C>T p.S7L Chr.1: 25820740-25820740 on Build GRCh38
3943823 c.255C>G p.R85R Chr.1: 25820505-25820505 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs116015767 Chr.1: 25820906-25820906 on Build GRCh38
rs775453215 Chr.1: 25820625-25820625 on Build GRCh38
rs920561238 Chr.1: 25820569-25820569 on Build GRCh38
rs112296613 Chr.1: 25820641-25820641 on Build GRCh38
rs1000942467 Chr.1: 25820850-25820850 on Build GRCh38
rs772028064 Chr.1: 25820727-25820727 on Build GRCh38
rs917798255 Chr.1: 25820888-25820888 on Build GRCh38
rs1038489760 Chr.1: 25820639-25820639 on Build GRCh38
rs761313124 Chr.1: 25820856-25820856 on Build GRCh38
rs544240457 Chr.1: 25820634-25820634 on Build GRCh38
rs986064593 Chr.1: 25820732-25820732 on Build GRCh38
rs551958370 Chr.1: 25820817-25820817 on Build GRCh38
rs1004312081 Chr.1: 25820590-25820590 on Build GRCh38
rs376466538 Chr.1: 25820828-25820829 on Build GRCh38
rs867788014 Chr.1: 25820963-25820963 on Build GRCh38
rs532218328 Chr.1: 25820700-25820700 on Build GRCh38
rs762416030 Chr.1: 25820705-25820705 on Build GRCh38
rs944415717 Chr.1: 25820619-25820619 on Build GRCh38
rs756957394 Chr.1: 25820787-25820787 on Build GRCh38
rs765038172 Chr.1: 25820524-25820524 on Build GRCh38
rs995543227 Chr.1: 25820714-25820714 on Build GRCh38
rs574609934 Chr.1: 25820642-25820642 on Build GRCh38
rs114425492 Chr.1: 25820568-25820568 on Build GRCh38
rs116526506 Chr.1: 25820579-25820579 on Build GRCh38
rs1038545895 Chr.1: 25820789-25820789 on Build GRCh38
rs1020946253 Chr.1: 25820841-25820841 on Build GRCh38
rs568231345 Chr.1: 25820557-25820557 on Build GRCh38
rs765045803 Chr.1: 25820784-25820784 on Build GRCh38
rs907659216 Chr.1: 25820773-25820773 on Build GRCh38
rs949050418 Chr.1: 25820759-25820759 on Build GRCh38
rs959429348 Chr.1: 25820864-25820864 on Build GRCh38
rs75503083 Chr.1: 25820588-25820588 on Build GRCh38
rs941821333 Chr.1: 25820965-25820965 on Build GRCh38
rs777388229 Chr.1: 25820646-25820654 on Build GRCh38
rs569548222 Chr.1: 25820816-25820816 on Build GRCh38
rs1004375925 Chr.1: 25820880-25820880 on Build GRCh38
rs183150119 Chr.1: 25820972-25820972 on Build GRCh38
rs1028359774 Chr.1: 25820680-25820680 on Build GRCh38
rs560336705 Chr.1: 25820692-25820692 on Build GRCh38
rs764945736 Chr.1: 25820909-25820909 on Build GRCh38
rs556682593 Chr.1: 25820973-25820973 on Build GRCh38
rs774312170 Chr.1: 25820725-25820725 on Build GRCh38
rs962749037 Chr.1: 25820566-25820566 on Build GRCh38
rs898653603 Chr.1: 25820795-25820795 on Build GRCh38
rs898608471 Chr.1: 25820898-25820898 on Build GRCh38
rs568110369 Chr.1: 25820798-25820798 on Build GRCh38
rs1027970925 Chr.1: 25820719-25820719 on Build GRCh38
rs941769024 Chr.1: 25820782-25820782 on Build GRCh38
rs111395389 Chr.1: 25820751-25820751 on Build GRCh38
rs771423467 Chr.1: 25820712-25820712 on Build GRCh38
rs575444055 Chr.1: 25820826-25820827 on Build GRCh38
rs576780321 Chr.1: 25820608-25820608 on Build GRCh38
rs1037057749 Chr.1: 25820881-25820881 on Build GRCh38
rs917594509 Chr.1: 25820753-25820753 on Build GRCh38
rs754111112 Chr.1: 25820897-25820897 on Build GRCh38
rs915577889 Chr.1: 25820818-25820818 on Build GRCh38
rs760853821 Chr.1: 25820783-25820783 on Build GRCh38
rs955516331 Chr.1: 25820686-25820686 on Build GRCh38
rs767203068 Chr.1: 25820887-25820887 on Build GRCh38
rs548212866 Chr.1: 25820761-25820761 on Build GRCh38
rs1045968913 Chr.1: 25820835-25820835 on Build GRCh38
rs1008259306 Chr.1: 25820834-25820834 on Build GRCh38
rs569095058 Chr.1: 25820525-25820525 on Build GRCh38
rs779532323 Chr.1: 25820912-25820912 on Build GRCh38
rs770332650 Chr.1: 25820606-25820606 on Build GRCh38
rs759318034 Chr.1: 25820765-25820765 on Build GRCh38
rs534066714 Chr.1: 25820804-25820804 on Build GRCh38
rs1004269808 Chr.1: 25820549-25820549 on Build GRCh38
rs556581949 Chr.1: 25820607-25820607 on Build GRCh38
rs966771390 Chr.1: 25820702-25820702 on Build GRCh38
rs1028300974 Chr.1: 25820910-25820910 on Build GRCh38
rs537852674 Chr.1: 25820532-25820532 on Build GRCh38
rs528686254 Chr.1: 25820726-25820726 on Build GRCh38
rs190554893 Chr.1: 25820659-25820659 on Build GRCh38
rs907548197 Chr.1: 25820955-25820955 on Build GRCh38
rs115285222 Chr.1: 25820842-25820842 on Build GRCh38
rs1015663550 Chr.1: 25820703-25820703 on Build GRCh38
rs550412869 Chr.1: 25820785-25820788 on Build GRCh38
rs891139733 Chr.1: 25820830-25820830 on Build GRCh38
rs778253621 Chr.1: 25820640-25820640 on Build GRCh38
rs562466186 Chr.1: 25820731-25820731 on Build GRCh38
rs750168056 Chr.1: 25820791-25820791 on Build GRCh38
rs907381272 Chr.1: 25820701-25820703 on Build GRCh38
rs974669722 Chr.1: 25820684-25820692 on Build GRCh38
rs935648903 Chr.1: 25820809-25820809 on Build GRCh38
rs554391055 Chr.1: 25820576-25820576 on Build GRCh38
rs548152478 Chr.1: 25820544-25820544 on Build GRCh38

View Full Product Details