Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 25823283-25823780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC646471
Entrez Gene ID
646471
Gene Name
uncharacterized LOC646471
Gene Aliases
-
UniGene
Hs.727271
Gene Chromosome Location
Chr.1: 25819954-25823606 on Build GRCh38
Transcripts
NR_024498.1 NM_001099625.1 NM_001099626.1 NM_001099627.1 NM_019557.5

Gene Symbol
MTFR1L
Entrez Gene ID
56181
Gene Name
mitochondrial fission regulator 1 like
Gene Aliases
FAM54B, HYST1888, MST116, MSTP116
UniGene
Hs.149305
Gene Chromosome Location
Chr.1: 25819906-25832942 on Build GRCh38
Transcripts
NR_024498.1 NM_001099625.1 NM_001099626.1 NM_001099627.1 NM_019557.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5673410 c.35T>C p.I12T Chr.1: 25823654-25823654 on Build GRCh38
1193363 c.53A>G p.H18R Chr.1: 25823672-25823672 on Build GRCh38
1341275 c.75_77delAAG p.R27delR Chr.1: 25823694-25823696 on Build GRCh38
3984926 c.25-6C>T p.? Chr.1: 25823638-25823638 on Build GRCh38
6716321 c.84T>C p.I28I Chr.1: 25823703-25823703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199996657 Chr.1: 25823669-25823669 on Build GRCh38
rs528687191 Chr.1: 25823392-25823392 on Build GRCh38
rs761288487 Chr.1: 25823716-25823716 on Build GRCh38
rs966105613 Chr.1: 25823371-25823371 on Build GRCh38
rs781109475 Chr.1: 25823379-25823379 on Build GRCh38
rs200706432 Chr.1: 25823607-25823607 on Build GRCh38
rs990095875 Chr.1: 25823717-25823717 on Build GRCh38
rs868752709 Chr.1: 25823619-25823619 on Build GRCh38
rs371673961 Chr.1: 25823738-25823738 on Build GRCh38
rs1056593350 Chr.1: 25823585-25823585 on Build GRCh38
rs746977795 Chr.1: 25823684-25823684 on Build GRCh38
rs766134653 Chr.1: 25823604-25823604 on Build GRCh38
rs753231852 Chr.1: 25823741-25823741 on Build GRCh38
rs989958162 Chr.1: 25823306-25823306 on Build GRCh38
rs34087705 Chr.1: 25823413-25823414 on Build GRCh38
rs1007663879 Chr.1: 25823347-25823347 on Build GRCh38
rs769343659 Chr.1: 25823718-25823718 on Build GRCh38
rs779989106 Chr.1: 25823428-25823429 on Build GRCh38
rs768615749 Chr.1: 25823607-25823607 on Build GRCh38
rs975938403 Chr.1: 25823383-25823383 on Build GRCh38
rs772891310 Chr.1: 25823726-25823726 on Build GRCh38
rs201755779 Chr.1: 25823727-25823727 on Build GRCh38
rs200941313 Chr.1: 25823699-25823699 on Build GRCh38
rs1021989561 Chr.1: 25823723-25823723 on Build GRCh38
rs775630751 Chr.1: 25823584-25823584 on Build GRCh38
rs565587860 Chr.1: 25823544-25823544 on Build GRCh38
rs927976351 Chr.1: 25823506-25823506 on Build GRCh38
rs180868781 Chr.1: 25823609-25823609 on Build GRCh38
rs34173733 Chr.1: 25823711-25823711 on Build GRCh38
rs777912511 Chr.1: 25823670-25823670 on Build GRCh38
rs924429734 Chr.1: 25823398-25823398 on Build GRCh38
rs1008003716 Chr.1: 25823638-25823638 on Build GRCh38
rs928410428 Chr.1: 25823535-25823535 on Build GRCh38
rs377434391 Chr.1: 25823620-25823620 on Build GRCh38
rs771975891 Chr.1: 25823581-25823581 on Build GRCh38
rs745597185 Chr.1: 25823580-25823580 on Build GRCh38
rs990431917 Chr.1: 25823447-25823447 on Build GRCh38
rs952101732 Chr.1: 25823307-25823307 on Build GRCh38
rs578027160 Chr.1: 25823735-25823735 on Build GRCh38
rs535212212 Chr.1: 25823649-25823649 on Build GRCh38
rs748423350 Chr.1: 25823403-25823403 on Build GRCh38
rs752650833 Chr.1: 25823665-25823665 on Build GRCh38
rs776683282 Chr.1: 25823648-25823656 on Build GRCh38
rs764740956 Chr.1: 25823612-25823612 on Build GRCh38
rs11247758 Chr.1: 25823532-25823532 on Build GRCh38
rs184122423 Chr.1: 25823733-25823733 on Build GRCh38
rs767852018 Chr.1: 25823739-25823739 on Build GRCh38
rs746895402 Chr.1: 25823577-25823578 on Build GRCh38
rs368084911 Chr.1: 25823672-25823672 on Build GRCh38
rs992721480 Chr.1: 25823564-25823564 on Build GRCh38
rs774197376 Chr.1: 25823579-25823579 on Build GRCh38
rs772728038 Chr.1: 25823391-25823391 on Build GRCh38
rs1044053644 Chr.1: 25823533-25823533 on Build GRCh38
rs760671204 Chr.1: 25823591-25823591 on Build GRCh38
rs34450889 Chr.1: 25823671-25823671 on Build GRCh38
rs980773287 Chr.1: 25823438-25823438 on Build GRCh38
rs755090742 Chr.1: 25823350-25823350 on Build GRCh38
rs200155933 Chr.1: 25823628-25823628 on Build GRCh38
rs751880717 Chr.1: 25823736-25823736 on Build GRCh38
rs955946766 Chr.1: 25823429-25823429 on Build GRCh38
rs929852338 Chr.1: 25823600-25823600 on Build GRCh38
rs781362747 Chr.1: 25823659-25823659 on Build GRCh38
rs769865538 Chr.1: 25823487-25823487 on Build GRCh38
rs752108412 Chr.1: 25823629-25823629 on Build GRCh38
rs761898452 Chr.1: 25823694-25823696 on Build GRCh38
rs769940018 Chr.1: 25823749-25823752 on Build GRCh38
rs914259451 Chr.1: 25823518-25823518 on Build GRCh38
rs758829247 Chr.1: 25823623-25823623 on Build GRCh38

View Full Product Details