Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 25834641-25835137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUNIP
Entrez Gene ID
79000
Gene Name
aurora kinase A and ninein interacting protein
Gene Aliases
AIBP, C1orf135
UniGene
Hs.149305
Gene Chromosome Location
Chr.1: 25831913-25859458 on Build GRCh38
Transcripts
NM_001287490.1 NM_024037.2 XM_017002300.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6608836 c.960T>C p.F320F Chr.1: 25835107-25835107 on Build GRCh38
7136534 c.1018G>T p.D340Y Chr.1: 25835049-25835049 on Build GRCh38
159535 c.1026C>G p.L342L Chr.1: 25835041-25835041 on Build GRCh38
4562821 c.944G>A p.W315* Chr.1: 25835123-25835123 on Build GRCh38
1296205 c.994G>A p.G332R Chr.1: 25835073-25835073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756183152 Chr.1: 25834854-25834854 on Build GRCh38
rs939262839 Chr.1: 25834662-25834662 on Build GRCh38
rs995677481 Chr.1: 25834666-25834666 on Build GRCh38
rs958137506 Chr.1: 25834828-25834828 on Build GRCh38
rs890041404 Chr.1: 25834755-25834755 on Build GRCh38
rs915963667 Chr.1: 25835080-25835080 on Build GRCh38
rs965449276 Chr.1: 25834774-25834774 on Build GRCh38
rs981412306 Chr.1: 25835105-25835105 on Build GRCh38
rs994994291 Chr.1: 25834814-25834814 on Build GRCh38
rs765553797 Chr.1: 25834983-25834983 on Build GRCh38
rs989318692 Chr.1: 25834830-25834830 on Build GRCh38
rs745803624 Chr.1: 25834691-25834691 on Build GRCh38
rs1007142951 Chr.1: 25834847-25834847 on Build GRCh38
rs373622719 Chr.1: 25834988-25834988 on Build GRCh38
rs571187562 Chr.1: 25834729-25834729 on Build GRCh38
rs752651934 Chr.1: 25835033-25835033 on Build GRCh38
rs552242318 Chr.1: 25834820-25834820 on Build GRCh38
rs940438820 Chr.1: 25834931-25834931 on Build GRCh38
rs368653551 Chr.1: 25835049-25835049 on Build GRCh38
rs966236670 Chr.1: 25834907-25834907 on Build GRCh38
rs745488262 Chr.1: 25835110-25835110 on Build GRCh38
rs768489838 Chr.1: 25834719-25834719 on Build GRCh38
rs1036142382 Chr.1: 25834934-25834934 on Build GRCh38
rs761217364 Chr.1: 25834957-25834957 on Build GRCh38
rs947957310 Chr.1: 25834914-25834914 on Build GRCh38
rs527379546 Chr.1: 25834920-25834920 on Build GRCh38
rs1007307500 Chr.1: 25834756-25834756 on Build GRCh38
rs78766448 Chr.1: 25835102-25835102 on Build GRCh38
rs756004856 Chr.1: 25835034-25835034 on Build GRCh38
rs182099645 Chr.1: 25834975-25834975 on Build GRCh38
rs897682493 Chr.1: 25834762-25834762 on Build GRCh38
rs768527578 Chr.1: 25835081-25835081 on Build GRCh38
rs1031025365 Chr.1: 25834793-25834793 on Build GRCh38
rs776191185 Chr.1: 25834942-25834942 on Build GRCh38
rs747641812 Chr.1: 25835112-25835112 on Build GRCh38
rs776788672 Chr.1: 25834980-25834980 on Build GRCh38
rs981780535 Chr.1: 25834917-25834917 on Build GRCh38
rs1018581680 Chr.1: 25834877-25834877 on Build GRCh38
rs761954930 Chr.1: 25834982-25834982 on Build GRCh38
rs753283648 Chr.1: 25835069-25835069 on Build GRCh38
rs767275773 Chr.1: 25835030-25835030 on Build GRCh38
rs189884388 Chr.1: 25834911-25834911 on Build GRCh38
rs1032236482 Chr.1: 25834946-25834946 on Build GRCh38
rs927584690 Chr.1: 25834927-25834927 on Build GRCh38
rs772512389 Chr.1: 25835111-25835111 on Build GRCh38
rs554672926 Chr.1: 25835058-25835058 on Build GRCh38
rs746884388 Chr.1: 25834982-25834982 on Build GRCh38
rs756706513 Chr.1: 25835088-25835088 on Build GRCh38
rs1019541500 Chr.1: 25834763-25834763 on Build GRCh38
rs759417915 Chr.1: 25835014-25835014 on Build GRCh38
rs184957289 Chr.1: 25834819-25834819 on Build GRCh38
rs978125796 Chr.1: 25834777-25834777 on Build GRCh38

View Full Product Details