Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26183985-26184475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNKSR1
Entrez Gene ID
10256
Gene Name
connector enhancer of kinase suppressor of Ras 1
Gene Aliases
CNK, CNK1
UniGene
Hs.16232
Gene Chromosome Location
Chr.1: 26177490-26189884 on Build GRCh38
Transcripts
NM_001297647.1 NM_001297648.1 NM_006314.2 XM_011540484.2 XM_011540485.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5907860 c.1060C>T p.P354S Chr.1: 26184460-26184460 on Build GRCh38
5903316 c.1039C>T p.P347S Chr.1: 26184439-26184439 on Build GRCh38
4515317 c.1037_1038CC>TT p.P346L Chr.1: 26184437-26184438 on Build GRCh38
6631198 c.979G>A p.G327R Chr.1: 26184266-26184266 on Build GRCh38
5053350 c.858G>T p.T286T Chr.1: 26184073-26184073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749955627 Chr.1: 26184059-26184059 on Build GRCh38
rs978411182 Chr.1: 26184100-26184100 on Build GRCh38
rs769043013 Chr.1: 26184074-26184074 on Build GRCh38
rs200315188 Chr.1: 26184229-26184229 on Build GRCh38
rs757826978 Chr.1: 26184237-26184237 on Build GRCh38
rs190132337 Chr.1: 26184233-26184233 on Build GRCh38
rs1019551538 Chr.1: 26184081-26184081 on Build GRCh38
rs773767026 Chr.1: 26184096-26184096 on Build GRCh38
rs938963655 Chr.1: 26184224-26184224 on Build GRCh38
rs780810191 Chr.1: 26184157-26184157 on Build GRCh38
rs770946606 Chr.1: 26184163-26184163 on Build GRCh38
rs151076692 Chr.1: 26184072-26184072 on Build GRCh38
rs760558603 Chr.1: 26184028-26184028 on Build GRCh38
rs746008112 Chr.1: 26184265-26184265 on Build GRCh38
rs537321990 Chr.1: 26184315-26184315 on Build GRCh38
rs751291867 Chr.1: 26184434-26184435 on Build GRCh38
rs772209035 Chr.1: 26184266-26184266 on Build GRCh38
rs111870124 Chr.1: 26184321-26184321 on Build GRCh38
rs557328646 Chr.1: 26184349-26184349 on Build GRCh38
rs116353873 Chr.1: 26184158-26184158 on Build GRCh38
rs779270949 Chr.1: 26184242-26184242 on Build GRCh38
rs780324234 Chr.1: 26184378-26184378 on Build GRCh38
rs999478093 Chr.1: 26184329-26184329 on Build GRCh38
rs746626702 Chr.1: 26184073-26184073 on Build GRCh38
rs759792555 Chr.1: 26184128-26184128 on Build GRCh38
rs202082294 Chr.1: 26184064-26184064 on Build GRCh38
rs766036204 Chr.1: 26184402-26184403 on Build GRCh38
rs867995657 Chr.1: 26184410-26184410 on Build GRCh38
rs146897334 Chr.1: 26184130-26184130 on Build GRCh38
rs752376171 Chr.1: 26184317-26184317 on Build GRCh38
rs751736078 Chr.1: 26184024-26184024 on Build GRCh38
rs201899944 Chr.1: 26184180-26184180 on Build GRCh38
rs184934373 Chr.1: 26184005-26184005 on Build GRCh38
rs868021183 Chr.1: 26184142-26184142 on Build GRCh38
rs1019664538 Chr.1: 26184303-26184303 on Build GRCh38
rs747067656 Chr.1: 26184384-26184384 on Build GRCh38
rs1052318474 Chr.1: 26184014-26184014 on Build GRCh38
rs751128178 Chr.1: 26184203-26184203 on Build GRCh38
rs375678852 Chr.1: 26184162-26184162 on Build GRCh38
rs537608274 Chr.1: 26184135-26184135 on Build GRCh38
rs765877057 Chr.1: 26184147-26184147 on Build GRCh38
rs772477115 Chr.1: 26184175-26184175 on Build GRCh38
rs534718562 Chr.1: 26184181-26184181 on Build GRCh38
rs763869609 Chr.1: 26184454-26184454 on Build GRCh38
rs763931405 Chr.1: 26184042-26184042 on Build GRCh38
rs372355864 Chr.1: 26184052-26184052 on Build GRCh38
rs773056896 Chr.1: 26184185-26184185 on Build GRCh38
rs199807460 Chr.1: 26184230-26184230 on Build GRCh38
rs773112396 Chr.1: 26184285-26184285 on Build GRCh38
rs755720967 Chr.1: 26184393-26184393 on Build GRCh38
rs57880419 Chr.1: 26184370-26184370 on Build GRCh38
rs2297710 Chr.1: 26184449-26184449 on Build GRCh38
rs754674938 Chr.1: 26184156-26184156 on Build GRCh38
rs762990730 Chr.1: 26184188-26184188 on Build GRCh38
rs147047319 Chr.1: 26184283-26184283 on Build GRCh38
rs149576633 Chr.1: 26184440-26184440 on Build GRCh38
rs768985826 Chr.1: 26184414-26184414 on Build GRCh38
rs776613345 Chr.1: 26184131-26184131 on Build GRCh38
rs770345109 Chr.1: 26184093-26184093 on Build GRCh38
rs754282674 Chr.1: 26184350-26184350 on Build GRCh38
rs1031304351 Chr.1: 26184333-26184333 on Build GRCh38
rs1048478305 Chr.1: 26184264-26184264 on Build GRCh38
rs370866944 Chr.1: 26184371-26184371 on Build GRCh38
rs754480508 Chr.1: 26184208-26184208 on Build GRCh38
rs866662054 Chr.1: 26184248-26184248 on Build GRCh38
rs760415083 Chr.1: 26184320-26184320 on Build GRCh38
rs759061601 Chr.1: 26184314-26184314 on Build GRCh38
rs748648581 Chr.1: 26184299-26184299 on Build GRCh38
rs781267315 Chr.1: 26184417-26184417 on Build GRCh38
rs749115087 Chr.1: 26184425-26184425 on Build GRCh38
rs878875205 Chr.1: 26184187-26184187 on Build GRCh38
rs369925886 Chr.1: 26184290-26184290 on Build GRCh38
rs200925514 Chr.1: 26184160-26184160 on Build GRCh38
rs148237566 Chr.1: 26184441-26184441 on Build GRCh38
rs771965978 Chr.1: 26184446-26184446 on Build GRCh38
rs762582924 Chr.1: 26184248-26184248 on Build GRCh38
rs767163195 Chr.1: 26184214-26184214 on Build GRCh38
rs763076370 Chr.1: 26184017-26184017 on Build GRCh38
rs964069349 Chr.1: 26184200-26184200 on Build GRCh38
rs778968795 Chr.1: 26184055-26184055 on Build GRCh38
rs369638842 Chr.1: 26184027-26184027 on Build GRCh38
rs757871462 Chr.1: 26184145-26184145 on Build GRCh38
rs763632608 Chr.1: 26184337-26184337 on Build GRCh38
rs892390188 Chr.1: 26184015-26184015 on Build GRCh38
rs748489573 Chr.1: 26184084-26184084 on Build GRCh38
rs752286237 Chr.1: 26184215-26184215 on Build GRCh38
rs139381171 Chr.1: 26184274-26184274 on Build GRCh38
rs781580321 Chr.1: 26184083-26184083 on Build GRCh38
rs373382597 Chr.1: 26184326-26184326 on Build GRCh38
rs745916101 Chr.1: 26184172-26184172 on Build GRCh38
rs774457556 Chr.1: 26184115-26184115 on Build GRCh38
rs753857414 Chr.1: 26184048-26184048 on Build GRCh38
rs866031504 Chr.1: 26184216-26184216 on Build GRCh38
rs770969546 Chr.1: 26184304-26184304 on Build GRCh38
rs774529421 Chr.1: 26184170-26184170 on Build GRCh38
rs779564702 Chr.1: 26184361-26184361 on Build GRCh38
rs757554517 Chr.1: 26184356-26184356 on Build GRCh38
rs747196702 Chr.1: 26184276-26184276 on Build GRCh38
rs769323103 Chr.1: 26184091-26184092 on Build GRCh38
rs530262613 Chr.1: 26184097-26184097 on Build GRCh38
rs869198070 Chr.1: 26184215-26184215 on Build GRCh38
rs565609715 Chr.1: 26184179-26184179 on Build GRCh38
rs751006186 Chr.1: 26184151-26184151 on Build GRCh38
rs375281651 Chr.1: 26184101-26184101 on Build GRCh38
rs765152512 Chr.1: 26184140-26184140 on Build GRCh38
rs759427379 Chr.1: 26184444-26184444 on Build GRCh38
rs773214132 Chr.1: 26184013-26184013 on Build GRCh38
rs868393948 Chr.1: 26184355-26184355 on Build GRCh38
rs774208555 Chr.1: 26184312-26184312 on Build GRCh38
rs772635938 Chr.1: 26184139-26184145 on Build GRCh38
rs1023347759 Chr.1: 26184357-26184357 on Build GRCh38
rs750320145 Chr.1: 26184144-26184144 on Build GRCh38
rs766208799 Chr.1: 26184189-26184189 on Build GRCh38
rs766579166 Chr.1: 26184020-26184020 on Build GRCh38

View Full Product Details