Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26191206-26191707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER4
Entrez Gene ID
378807
Gene Name
cation channel sperm associated 4
Gene Aliases
-
UniGene
Hs.123532
Gene Chromosome Location
Chr.1: 26189962-26202542 on Build GRCh38
Transcripts
NM_198137.1 XM_011541432.2 XM_011541433.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
425987 c.312C>T p.I104I Chr.1: 26191385-26191385 on Build GRCh38
7248919 c.214-5T>C p.? Chr.1: 26191282-26191282 on Build GRCh38
1199702 c.328G>A p.A110T Chr.1: 26191401-26191401 on Build GRCh38
6321521 c.217G>T p.A73S Chr.1: 26191290-26191290 on Build GRCh38
5772751 c.269G>T p.R90L Chr.1: 26191342-26191342 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879209516 Chr.1: 26191528-26191528 on Build GRCh38
rs375645560 Chr.1: 26191247-26191247 on Build GRCh38
rs1019852625 Chr.1: 26191655-26191655 on Build GRCh38
rs557773497 Chr.1: 26191475-26191475 on Build GRCh38
rs779125947 Chr.1: 26191384-26191384 on Build GRCh38
rs200854480 Chr.1: 26191349-26191349 on Build GRCh38
rs778169515 Chr.1: 26191290-26191290 on Build GRCh38
rs200425327 Chr.1: 26191417-26191417 on Build GRCh38
rs781022634 Chr.1: 26191473-26191473 on Build GRCh38
rs1008846538 Chr.1: 26191650-26191650 on Build GRCh38
rs755501499 Chr.1: 26191278-26191278 on Build GRCh38
rs544019345 Chr.1: 26191346-26191346 on Build GRCh38
rs147002798 Chr.1: 26191311-26191311 on Build GRCh38
rs112294330 Chr.1: 26191480-26191480 on Build GRCh38
rs548427276 Chr.1: 26191495-26191495 on Build GRCh38
rs925016821 Chr.1: 26191481-26191481 on Build GRCh38
rs755450072 Chr.1: 26191458-26191458 on Build GRCh38
rs764323538 Chr.1: 26191358-26191358 on Build GRCh38
rs765400036 Chr.1: 26191468-26191468 on Build GRCh38
rs781584800 Chr.1: 26191466-26191467 on Build GRCh38
rs571115045 Chr.1: 26191569-26191569 on Build GRCh38
rs112088682 Chr.1: 26191468-26191469 on Build GRCh38
rs540174441 Chr.1: 26191600-26191600 on Build GRCh38
rs750619611 Chr.1: 26191470-26191470 on Build GRCh38
rs761896845 Chr.1: 26191467-26191467 on Build GRCh38
rs764883192 Chr.1: 26191240-26191240 on Build GRCh38
rs774003285 Chr.1: 26191419-26191419 on Build GRCh38
rs186603174 Chr.1: 26191344-26191344 on Build GRCh38
rs748751122 Chr.1: 26191469-26191470 on Build GRCh38
rs568555822 Chr.1: 26191496-26191496 on Build GRCh38
rs987036190 Chr.1: 26191470-26191470 on Build GRCh38
rs1046306287 Chr.1: 26191505-26191507 on Build GRCh38
rs142583631 Chr.1: 26191320-26191320 on Build GRCh38
rs547813971 Chr.1: 26191556-26191556 on Build GRCh38
rs779774662 Chr.1: 26191297-26191297 on Build GRCh38
rs748764690 Chr.1: 26191412-26191412 on Build GRCh38
rs867111001 Chr.1: 26191284-26191284 on Build GRCh38
rs531829467 Chr.1: 26191482-26191482 on Build GRCh38
rs138873095 Chr.1: 26191425-26191425 on Build GRCh38
rs747912994 Chr.1: 26191617-26191617 on Build GRCh38
rs1028833518 Chr.1: 26191636-26191636 on Build GRCh38
rs764695924 Chr.1: 26191455-26191455 on Build GRCh38
rs762569758 Chr.1: 26191341-26191341 on Build GRCh38
rs751938347 Chr.1: 26191269-26191269 on Build GRCh38
rs757490747 Chr.1: 26191374-26191374 on Build GRCh38
rs552254324 Chr.1: 26191439-26191439 on Build GRCh38
rs376375618 Chr.1: 26191342-26191342 on Build GRCh38
rs11247866 Chr.1: 26191303-26191303 on Build GRCh38
rs761235880 Chr.1: 26191353-26191353 on Build GRCh38
rs370488506 Chr.1: 26191434-26191434 on Build GRCh38
rs149799938 Chr.1: 26191302-26191302 on Build GRCh38
rs751327394 Chr.1: 26191391-26191391 on Build GRCh38
rs779127944 Chr.1: 26191408-26191408 on Build GRCh38
rs925069190 Chr.1: 26191280-26191280 on Build GRCh38
rs752834576 Chr.1: 26191289-26191289 on Build GRCh38
rs769706025 Chr.1: 26191407-26191407 on Build GRCh38
rs766702056 Chr.1: 26191260-26191260 on Build GRCh38
rs377237727 Chr.1: 26191421-26191421 on Build GRCh38
rs146575130 Chr.1: 26191291-26191291 on Build GRCh38
rs111438863 Chr.1: 26191350-26191350 on Build GRCh38
rs113478680 Chr.1: 26191486-26191486 on Build GRCh38
rs267598523 Chr.1: 26191314-26191314 on Build GRCh38
rs11247867 Chr.1: 26191400-26191400 on Build GRCh38
rs527657916 Chr.1: 26191525-26191525 on Build GRCh38
rs903864098 Chr.1: 26191598-26191598 on Build GRCh38
rs754719888 Chr.1: 26191397-26191397 on Build GRCh38
rs929068306 Chr.1: 26191398-26191398 on Build GRCh38
rs1004201454 Chr.1: 26191538-26191538 on Build GRCh38
rs11247865 Chr.1: 26191282-26191282 on Build GRCh38
rs780460020 Chr.1: 26191255-26191256 on Build GRCh38
rs1002652489 Chr.1: 26191647-26191647 on Build GRCh38
rs775978312 Chr.1: 26191422-26191422 on Build GRCh38
rs370751508 Chr.1: 26191401-26191401 on Build GRCh38
rs758595560 Chr.1: 26191471-26191471 on Build GRCh38
rs374451102 Chr.1: 26191459-26191459 on Build GRCh38
rs886388886 Chr.1: 26191509-26191509 on Build GRCh38
rs945046902 Chr.1: 26191229-26191229 on Build GRCh38
rs762634415 Chr.1: 26191250-26191250 on Build GRCh38
rs752482451 Chr.1: 26191476-26191476 on Build GRCh38
rs753943049 Chr.1: 26191369-26191369 on Build GRCh38
rs1033983401 Chr.1: 26191307-26191307 on Build GRCh38

View Full Product Details