Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26201060-26201561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER4
Entrez Gene ID
378807
Gene Name
cation channel sperm associated 4
Gene Aliases
-
UniGene
Hs.123532
Gene Chromosome Location
Chr.1: 26189962-26202542 on Build GRCh38
Transcripts
NM_198137.1 XM_011541432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6540232 c.1234C>T p.Q412* Chr.1: 26201388-26201388 on Build GRCh38
3804793 c.1249G>C p.E417Q Chr.1: 26201403-26201403 on Build GRCh38
6233319 c.1283C>T p.S428L Chr.1: 26201437-26201437 on Build GRCh38
309872 c.1278C>A p.S426R Chr.1: 26201432-26201432 on Build GRCh38
4882382 c.1245G>T p.E415D Chr.1: 26201399-26201399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41310785 Chr.1: 26201393-26201393 on Build GRCh38
rs762188913 Chr.1: 26201448-26201448 on Build GRCh38
rs760297915 Chr.1: 26201365-26201365 on Build GRCh38
rs755261725 Chr.1: 26201463-26201463 on Build GRCh38
rs201122868 Chr.1: 26201379-26201379 on Build GRCh38
rs776310427 Chr.1: 26201537-26201537 on Build GRCh38
rs371719915 Chr.1: 26201446-26201446 on Build GRCh38
rs1054290051 Chr.1: 26201227-26201227 on Build GRCh38
rs959618107 Chr.1: 26201272-26201272 on Build GRCh38
rs140820355 Chr.1: 26201389-26201389 on Build GRCh38
rs1039301780 Chr.1: 26201163-26201163 on Build GRCh38
rs766627192 Chr.1: 26201458-26201458 on Build GRCh38
rs760388086 Chr.1: 26201445-26201445 on Build GRCh38
rs57072055 Chr.1: 26201149-26201149 on Build GRCh38
rs1022751577 Chr.1: 26201255-26201255 on Build GRCh38
rs145608675 Chr.1: 26201373-26201373 on Build GRCh38
rs869139 Chr.1: 26201208-26201208 on Build GRCh38
rs149417932 Chr.1: 26201434-26201434 on Build GRCh38
rs149949236 Chr.1: 26201507-26201507 on Build GRCh38
rs1001393617 Chr.1: 26201429-26201429 on Build GRCh38
rs749248364 Chr.1: 26201332-26201332 on Build GRCh38
rs952250070 Chr.1: 26201331-26201331 on Build GRCh38
rs150759853 Chr.1: 26201486-26201486 on Build GRCh38
rs758562800 Chr.1: 26201311-26201311 on Build GRCh38
rs1022552942 Chr.1: 26201465-26201465 on Build GRCh38
rs751881999 Chr.1: 26201385-26201385 on Build GRCh38
rs386629719 Chr.1: 26201318-26201319 on Build GRCh38
rs200146849 Chr.1: 26201529-26201529 on Build GRCh38
rs765697791 Chr.1: 26201449-26201449 on Build GRCh38
rs553129973 Chr.1: 26201148-26201148 on Build GRCh38
rs781511663 Chr.1: 26201467-26201467 on Build GRCh38
rs1040762833 Chr.1: 26201213-26201213 on Build GRCh38
rs377636399 Chr.1: 26201273-26201273 on Build GRCh38
rs930733315 Chr.1: 26201161-26201161 on Build GRCh38
rs893055595 Chr.1: 26201409-26201409 on Build GRCh38
rs928261680 Chr.1: 26201093-26201093 on Build GRCh38
rs758328338 Chr.1: 26201500-26201500 on Build GRCh38
rs745874566 Chr.1: 26201497-26201497 on Build GRCh38
rs536204179 Chr.1: 26201095-26201095 on Build GRCh38
rs141342547 Chr.1: 26201405-26201405 on Build GRCh38
rs778382951 Chr.1: 26201421-26201423 on Build GRCh38
rs1030428064 Chr.1: 26201196-26201196 on Build GRCh38
rs904342592 Chr.1: 26201420-26201420 on Build GRCh38
rs775162393 Chr.1: 26201356-26201356 on Build GRCh38
rs574754918 Chr.1: 26201508-26201516 on Build GRCh38
rs778928926 Chr.1: 26201310-26201310 on Build GRCh38
rs900161427 Chr.1: 26201164-26201164 on Build GRCh38
rs760254567 Chr.1: 26201351-26201351 on Build GRCh38
rs753605966 Chr.1: 26201470-26201470 on Build GRCh38
rs201932747 Chr.1: 26201422-26201422 on Build GRCh38
rs6657616 Chr.1: 26201460-26201460 on Build GRCh38
rs114575246 Chr.1: 26201450-26201450 on Build GRCh38
rs6703730 Chr.1: 26201292-26201292 on Build GRCh38
rs757115096 Chr.1: 26201474-26201474 on Build GRCh38
rs920740666 Chr.1: 26201160-26201160 on Build GRCh38
rs374256521 Chr.1: 26201329-26201329 on Build GRCh38
rs200619838 Chr.1: 26201447-26201447 on Build GRCh38
rs889193226 Chr.1: 26201198-26201198 on Build GRCh38
rs114809277 Chr.1: 26201167-26201167 on Build GRCh38
rs1021429149 Chr.1: 26201222-26201222 on Build GRCh38
rs747729033 Chr.1: 26201538-26201538 on Build GRCh38
rs1889755 Chr.1: 26201318-26201318 on Build GRCh38
rs1013156892 Chr.1: 26201248-26201248 on Build GRCh38
rs185992221 Chr.1: 26201158-26201158 on Build GRCh38
rs34945330 Chr.1: 26201320-26201320 on Build GRCh38
rs17163674 Chr.1: 26201372-26201372 on Build GRCh38
rs544844334 Chr.1: 26201218-26201218 on Build GRCh38
rs548393206 Chr.1: 26201225-26201225 on Build GRCh38
rs368692279 Chr.1: 26201090-26201090 on Build GRCh38
rs971535104 Chr.1: 26201264-26201264 on Build GRCh38
rs367781080 Chr.1: 26201436-26201436 on Build GRCh38
rs917924209 Chr.1: 26201302-26201302 on Build GRCh38
rs1015647886 Chr.1: 26201294-26201294 on Build GRCh38
rs545011390 Chr.1: 26201380-26201380 on Build GRCh38
rs142696900 Chr.1: 26201381-26201381 on Build GRCh38
rs778868474 Chr.1: 26201406-26201406 on Build GRCh38
rs892940963 Chr.1: 26201235-26201235 on Build GRCh38
rs558959484 Chr.1: 26201455-26201455 on Build GRCh38
rs780379222 Chr.1: 26201312-26201312 on Build GRCh38
rs34351620 Chr.1: 26201317-26201318 on Build GRCh38
rs918033257 Chr.1: 26201105-26201105 on Build GRCh38
rs746465555 Chr.1: 26201512-26201512 on Build GRCh38
rs749050731 Chr.1: 26201396-26201396 on Build GRCh38
rs773742174 Chr.1: 26201341-26201341 on Build GRCh38
rs980006667 Chr.1: 26201256-26201256 on Build GRCh38
rs1889756 Chr.1: 26201319-26201319 on Build GRCh38
rs929400422 Chr.1: 26201120-26201120 on Build GRCh38

View Full Product Details