Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26202407-26202917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER4
Entrez Gene ID
378807
Gene Name
cation channel sperm associated 4
Gene Aliases
-
UniGene
Hs.123532
Gene Chromosome Location
Chr.1: 26189962-26202542 on Build GRCh38
Transcripts
NM_198137.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5352613 c.1397A>C p.K466T Chr.1: 26202520-26202520 on Build GRCh38
6620772 c.1393C>A p.L465I Chr.1: 26202516-26202516 on Build GRCh38
1341318 c.1396delA p.K467fs*>6 Chr.1: 26202519-26202519 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762977249 Chr.1: 26202571-26202572 on Build GRCh38
rs908322070 Chr.1: 26202825-26202825 on Build GRCh38
rs769940190 Chr.1: 26202592-26202592 on Build GRCh38
rs779408180 Chr.1: 26202459-26202459 on Build GRCh38
rs776604961 Chr.1: 26202446-26202447 on Build GRCh38
rs780387890 Chr.1: 26202586-26202586 on Build GRCh38
rs374054496 Chr.1: 26202489-26202489 on Build GRCh38
rs41284335 Chr.1: 26202760-26202760 on Build GRCh38
rs531617024 Chr.1: 26202755-26202755 on Build GRCh38
rs752281338 Chr.1: 26202533-26202533 on Build GRCh38
rs905958645 Chr.1: 26202816-26202816 on Build GRCh38
rs773833007 Chr.1: 26202666-26202666 on Build GRCh38
rs777246534 Chr.1: 26202451-26202451 on Build GRCh38
rs751712402 Chr.1: 26202577-26202578 on Build GRCh38
rs370651018 Chr.1: 26202448-26202448 on Build GRCh38
rs972588495 Chr.1: 26202653-26202653 on Build GRCh38
rs114100693 Chr.1: 26202570-26202570 on Build GRCh38
rs759526243 Chr.1: 26202514-26202514 on Build GRCh38
rs746442256 Chr.1: 26202460-26202460 on Build GRCh38
rs779423858 Chr.1: 26202580-26202580 on Build GRCh38
rs1043220354 Chr.1: 26202810-26202810 on Build GRCh38
rs890507997 Chr.1: 26202456-26202456 on Build GRCh38
rs759842760 Chr.1: 26202580-26202581 on Build GRCh38
rs908373585 Chr.1: 26202643-26202643 on Build GRCh38
rs1036106144 Chr.1: 26202890-26202890 on Build GRCh38
rs767305882 Chr.1: 26202532-26202532 on Build GRCh38
rs762093417 Chr.1: 26202591-26202591 on Build GRCh38
rs532029648 Chr.1: 26202588-26202588 on Build GRCh38
rs376750882 Chr.1: 26202446-26202446 on Build GRCh38
rs973848543 Chr.1: 26202845-26202845 on Build GRCh38
rs762004612 Chr.1: 26202497-26202497 on Build GRCh38
rs762697250 Chr.1: 26202440-26202440 on Build GRCh38
rs761834681 Chr.1: 26202467-26202469 on Build GRCh38
rs751354813 Chr.1: 26202511-26202511 on Build GRCh38
rs776681427 Chr.1: 26202590-26202590 on Build GRCh38
rs772391412 Chr.1: 26202475-26202475 on Build GRCh38
rs771470968 Chr.1: 26202455-26202455 on Build GRCh38
rs755810568 Chr.1: 26202450-26202450 on Build GRCh38
rs949796705 Chr.1: 26202625-26202625 on Build GRCh38
rs1013169004 Chr.1: 26202774-26202774 on Build GRCh38
rs756965785 Chr.1: 26202578-26202578 on Build GRCh38
rs189991470 Chr.1: 26202799-26202799 on Build GRCh38
rs769908264 Chr.1: 26202519-26202519 on Build GRCh38
rs959948051 Chr.1: 26202632-26202632 on Build GRCh38
rs910970427 Chr.1: 26202438-26202438 on Build GRCh38
rs760299739 Chr.1: 26202781-26202781 on Build GRCh38
rs561885735 Chr.1: 26202795-26202795 on Build GRCh38
rs1033292622 Chr.1: 26202582-26202582 on Build GRCh38
rs753600570 Chr.1: 26202577-26202577 on Build GRCh38
rs796793492 Chr.1: 26202570-26202571 on Build GRCh38
rs766134334 Chr.1: 26202445-26202445 on Build GRCh38
rs538754810 Chr.1: 26202717-26202717 on Build GRCh38
rs548868450 Chr.1: 26202593-26202593 on Build GRCh38
rs776879913 Chr.1: 26202495-26202495 on Build GRCh38
rs868672021 Chr.1: 26202550-26202550 on Build GRCh38
rs146769194 Chr.1: 26202757-26202757 on Build GRCh38
rs370382169 Chr.1: 26202587-26202587 on Build GRCh38
rs952584863 Chr.1: 26202672-26202672 on Build GRCh38
rs534090505 Chr.1: 26202701-26202701 on Build GRCh38
rs890464839 Chr.1: 26202688-26202688 on Build GRCh38
rs922462140 Chr.1: 26202619-26202619 on Build GRCh38
rs922253096 Chr.1: 26202714-26202714 on Build GRCh38
rs755615424 Chr.1: 26202543-26202543 on Build GRCh38
rs1038230794 Chr.1: 26202656-26202656 on Build GRCh38
rs775654140 Chr.1: 26202485-26202485 on Build GRCh38
rs10902720 Chr.1: 26202431-26202431 on Build GRCh38
rs772657385 Chr.1: 26202581-26202581 on Build GRCh38
rs748915336 Chr.1: 26202453-26202453 on Build GRCh38
rs41284337 Chr.1: 26202841-26202841 on Build GRCh38
rs932598477 Chr.1: 26202667-26202667 on Build GRCh38
rs186478083 Chr.1: 26202472-26202472 on Build GRCh38
rs371864812 Chr.1: 26202723-26202723 on Build GRCh38
rs540046061 Chr.1: 26202863-26202863 on Build GRCh38
rs6691747 Chr.1: 26202630-26202630 on Build GRCh38
rs766224537 Chr.1: 26202498-26202498 on Build GRCh38
rs773361308 Chr.1: 26202570-26202571 on Build GRCh38
rs780824102 Chr.1: 26202447-26202447 on Build GRCh38
rs942436237 Chr.1: 26202710-26202710 on Build GRCh38
rs777392436 Chr.1: 26202566-26202566 on Build GRCh38
rs75399806 Chr.1: 26202713-26202713 on Build GRCh38
rs142388907 Chr.1: 26202702-26202702 on Build GRCh38
rs941225918 Chr.1: 26202649-26202649 on Build GRCh38
rs556463096 Chr.1: 26202740-26202740 on Build GRCh38

View Full Product Details