Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27379875-27380366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD164L2
Entrez Gene ID
388611
Gene Name
CD164 molecule like 2
Gene Aliases
EAPG6122, UNQ6122
UniGene
Hs.664836
Gene Chromosome Location
Chr.1: 27379100-27383333 on Build GRCh38
Transcripts
NM_207397.3 XM_005245868.3 XM_011541441.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3487693 c.429C>T p.I143I Chr.1: 27380140-27380140 on Build GRCh38
7299441 c.487C>T p.L163F Chr.1: 27380082-27380082 on Build GRCh38
1667305 c.400A>G p.S134G Chr.1: 27380169-27380169 on Build GRCh38
3804851 c.452G>A p.S151N Chr.1: 27380117-27380117 on Build GRCh38
1296247 c.415G>A p.G139R Chr.1: 27380154-27380154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763286270 Chr.1: 27380040-27380041 on Build GRCh38
rs768373343 Chr.1: 27380091-27380091 on Build GRCh38
rs775345862 Chr.1: 27380073-27380073 on Build GRCh38
rs757459339 Chr.1: 27380184-27380184 on Build GRCh38
rs935939658 Chr.1: 27379974-27379974 on Build GRCh38
rs534756456 Chr.1: 27380054-27380054 on Build GRCh38
rs2504777 Chr.1: 27380011-27380011 on Build GRCh38
rs992932439 Chr.1: 27380154-27380154 on Build GRCh38
rs564064762 Chr.1: 27379905-27379905 on Build GRCh38
rs977247292 Chr.1: 27380262-27380262 on Build GRCh38
rs891744896 Chr.1: 27379990-27379990 on Build GRCh38
rs755349105 Chr.1: 27380028-27380028 on Build GRCh38
rs144456933 Chr.1: 27380146-27380146 on Build GRCh38
rs202152114 Chr.1: 27380225-27380225 on Build GRCh38
rs376059290 Chr.1: 27380199-27380199 on Build GRCh38
rs942162630 Chr.1: 27379945-27379945 on Build GRCh38
rs533015030 Chr.1: 27379926-27379926 on Build GRCh38
rs755393856 Chr.1: 27380185-27380186 on Build GRCh38
rs950982387 Chr.1: 27380164-27380164 on Build GRCh38
rs370414726 Chr.1: 27380219-27380219 on Build GRCh38
rs748006968 Chr.1: 27380204-27380204 on Build GRCh38
rs948376657 Chr.1: 27380334-27380334 on Build GRCh38
rs749382625 Chr.1: 27380055-27380055 on Build GRCh38
rs572293698 Chr.1: 27380194-27380194 on Build GRCh38
rs147548703 Chr.1: 27379928-27379928 on Build GRCh38
rs748279412 Chr.1: 27380044-27380044 on Build GRCh38
rs977278667 Chr.1: 27379968-27379968 on Build GRCh38
rs376916990 Chr.1: 27380150-27380150 on Build GRCh38
rs752006574 Chr.1: 27380165-27380171 on Build GRCh38
rs563847570 Chr.1: 27379935-27379935 on Build GRCh38
rs924521315 Chr.1: 27379969-27379969 on Build GRCh38
rs1045396316 Chr.1: 27380342-27380342 on Build GRCh38
rs191291802 Chr.1: 27379988-27379988 on Build GRCh38
rs769426863 Chr.1: 27380206-27380206 on Build GRCh38
rs772129402 Chr.1: 27380072-27380072 on Build GRCh38
rs199850149 Chr.1: 27380114-27380114 on Build GRCh38
rs776227227 Chr.1: 27380203-27380203 on Build GRCh38
rs942070389 Chr.1: 27380185-27380185 on Build GRCh38
rs772709028 Chr.1: 27380110-27380110 on Build GRCh38
rs147343428 Chr.1: 27380155-27380155 on Build GRCh38
rs1004318961 Chr.1: 27380126-27380126 on Build GRCh38
rs199994753 Chr.1: 27380240-27380240 on Build GRCh38
rs1047947578 Chr.1: 27379932-27379932 on Build GRCh38
rs145796710 Chr.1: 27380140-27380140 on Build GRCh38
rs891713677 Chr.1: 27380248-27380248 on Build GRCh38
rs753126507 Chr.1: 27380142-27380142 on Build GRCh38
rs374222075 Chr.1: 27380189-27380189 on Build GRCh38
rs184338267 Chr.1: 27380107-27380107 on Build GRCh38
rs746794490 Chr.1: 27380088-27380088 on Build GRCh38
rs915841096 Chr.1: 27380326-27380326 on Build GRCh38
rs1039413656 Chr.1: 27379950-27379950 on Build GRCh38
rs557037446 Chr.1: 27379989-27379989 on Build GRCh38
rs945972908 Chr.1: 27380291-27380291 on Build GRCh38
rs983612102 Chr.1: 27380182-27380182 on Build GRCh38
rs187568809 Chr.1: 27379936-27379936 on Build GRCh38
rs944512251 Chr.1: 27379940-27379940 on Build GRCh38
rs762420401 Chr.1: 27380214-27380214 on Build GRCh38
rs77539957 Chr.1: 27380308-27380308 on Build GRCh38
rs781446752 Chr.1: 27380037-27380037 on Build GRCh38
rs900503279 Chr.1: 27379965-27379965 on Build GRCh38
rs138512492 Chr.1: 27380130-27380130 on Build GRCh38
rs571660185 Chr.1: 27380108-27380108 on Build GRCh38
rs1051739897 Chr.1: 27380233-27380233 on Build GRCh38
rs772902870 Chr.1: 27380213-27380213 on Build GRCh38
rs755541317 Chr.1: 27380132-27380132 on Build GRCh38
rs924404923 Chr.1: 27380280-27380280 on Build GRCh38
rs928384276 Chr.1: 27380344-27380344 on Build GRCh38
rs767703107 Chr.1: 27380089-27380124 on Build GRCh38
rs367980682 Chr.1: 27380290-27380290 on Build GRCh38
rs142703194 Chr.1: 27380131-27380131 on Build GRCh38
rs766786758 Chr.1: 27380129-27380131 on Build GRCh38
rs904371888 Chr.1: 27380325-27380325 on Build GRCh38

View Full Product Details