Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27614772-27615266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGR
Entrez Gene ID
2268
Gene Name
FGR proto-oncogene, Src family tyrosine kinase
Gene Aliases
SRC2, c-fgr, c-src2, p55-Fgr, p55c-fgr, p58-Fgr, p58c-fgr
UniGene
Hs.1422
Gene Chromosome Location
Chr.1: 27612289-27635561 on Build GRCh38
Transcripts
NM_001042729.1 NM_001042747.1 NM_005248.2 XM_006710452.2 XM_011541010.1 XM_011541011.2 XM_011541012.1 XM_011541013.2 XM_011541014.2 XM_017000673.1 XM_017000674.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3930736 c.1062G>C p.L354F Chr.1: 27614883-27614883 on Build GRCh38
4894891 c.1024T>C p.L342L Chr.1: 27614921-27614921 on Build GRCh38
7417439 c.1019G>A p.G340D Chr.1: 27614926-27614926 on Build GRCh38
4941011 c.1080G>T p.V360V Chr.1: 27614865-27614865 on Build GRCh38
5682340 c.1095+2_1095+7delTAACTG p.? Chr.1: 27614843-27614848 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs553963840 Chr.1: 27614843-27614843 on Build GRCh38
rs546506290 Chr.1: 27615062-27615062 on Build GRCh38
rs1041486327 Chr.1: 27615009-27615009 on Build GRCh38
rs1007209934 Chr.1: 27615145-27615145 on Build GRCh38
rs533612613 Chr.1: 27614808-27614808 on Build GRCh38
rs1055122621 Chr.1: 27614879-27614879 on Build GRCh38
rs752070517 Chr.1: 27614950-27614950 on Build GRCh38
rs576689059 Chr.1: 27615234-27615235 on Build GRCh38
rs771975393 Chr.1: 27614833-27614833 on Build GRCh38
rs1028003445 Chr.1: 27615056-27615056 on Build GRCh38
rs749406970 Chr.1: 27614811-27614811 on Build GRCh38
rs994651397 Chr.1: 27615048-27615048 on Build GRCh38
rs757459529 Chr.1: 27615065-27615065 on Build GRCh38
rs1012491783 Chr.1: 27615107-27615107 on Build GRCh38
rs766898502 Chr.1: 27614935-27614935 on Build GRCh38
rs761409623 Chr.1: 27614871-27614871 on Build GRCh38
rs889020627 Chr.1: 27615104-27615104 on Build GRCh38
rs749566844 Chr.1: 27615160-27615160 on Build GRCh38
rs577010848 Chr.1: 27615053-27615053 on Build GRCh38
rs931917459 Chr.1: 27614798-27614798 on Build GRCh38
rs113042902 Chr.1: 27615148-27615148 on Build GRCh38
rs897655283 Chr.1: 27615047-27615047 on Build GRCh38
rs1050721631 Chr.1: 27614811-27614812 on Build GRCh38
rs948716952 Chr.1: 27614895-27614895 on Build GRCh38
rs894574731 Chr.1: 27615005-27615005 on Build GRCh38
rs890284687 Chr.1: 27614832-27614832 on Build GRCh38
rs375607314 Chr.1: 27614907-27614907 on Build GRCh38
rs967338741 Chr.1: 27615023-27615023 on Build GRCh38
rs541868995 Chr.1: 27615155-27615155 on Build GRCh38
rs1036235980 Chr.1: 27614973-27614973 on Build GRCh38
rs185123115 Chr.1: 27615183-27615183 on Build GRCh38
rs760502992 Chr.1: 27614845-27614845 on Build GRCh38
rs957783423 Chr.1: 27615230-27615230 on Build GRCh38
rs763837829 Chr.1: 27614847-27614847 on Build GRCh38
rs755337653 Chr.1: 27614951-27614951 on Build GRCh38
rs748419059 Chr.1: 27614803-27614803 on Build GRCh38
rs763758174 Chr.1: 27614977-27615007 on Build GRCh38
rs563038840 Chr.1: 27615082-27615082 on Build GRCh38
rs1024251135 Chr.1: 27615128-27615128 on Build GRCh38
rs894099450 Chr.1: 27615091-27615091 on Build GRCh38
rs1004291358 Chr.1: 27614965-27614965 on Build GRCh38
rs369648234 Chr.1: 27614919-27614919 on Build GRCh38
rs868013931 Chr.1: 27614857-27614857 on Build GRCh38
rs1032919287 Chr.1: 27615028-27615058 on Build GRCh38
rs1031386981 Chr.1: 27615198-27615198 on Build GRCh38
rs973403182 Chr.1: 27615207-27615207 on Build GRCh38
rs779001229 Chr.1: 27614929-27614929 on Build GRCh38
rs776328458 Chr.1: 27614870-27614870 on Build GRCh38
rs990308392 Chr.1: 27615236-27615236 on Build GRCh38
rs373209754 Chr.1: 27614892-27614892 on Build GRCh38
rs920588170 Chr.1: 27615233-27615233 on Build GRCh38
rs1011659054 Chr.1: 27615013-27615013 on Build GRCh38
rs1021669512 Chr.1: 27615017-27615017 on Build GRCh38
rs1032602253 Chr.1: 27615084-27615084 on Build GRCh38

View Full Product Details