Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40140121-40140617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC3
Entrez Gene ID
22794
Gene Name
cancer susceptibility candidate 3
Gene Aliases
BTZ, MLN51
UniGene
Hs.743287
Gene Chromosome Location
Chr.17: 40140254-40172178 on Build GRCh38
Transcripts
NM_007359.4 XM_005257163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3516963 c.14G>C p.R5P Chr.17: 40140562-40140562 on Build GRCh38
7108535 c.62C>T p.S21L Chr.17: 40140610-40140610 on Build GRCh38
560919 c.29C>T p.S10L Chr.17: 40140577-40140577 on Build GRCh38
1610176 c.27T>A p.A9A Chr.17: 40140575-40140575 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745438579 Chr.17: 40140535-40140535 on Build GRCh38
rs60785254 Chr.17: 40140389-40140390 on Build GRCh38
rs1024009608 Chr.17: 40140267-40140267 on Build GRCh38
rs930218378 Chr.17: 40140218-40140218 on Build GRCh38
rs886600175 Chr.17: 40140517-40140517 on Build GRCh38
rs375234919 Chr.17: 40140587-40140587 on Build GRCh38
rs768305986 Chr.17: 40140566-40140566 on Build GRCh38
rs911276777 Chr.17: 40140185-40140185 on Build GRCh38
rs371404204 Chr.17: 40140526-40140526 on Build GRCh38
rs747095074 Chr.17: 40140182-40140182 on Build GRCh38
rs904926741 Chr.17: 40140427-40140427 on Build GRCh38
rs753023837 Chr.17: 40140529-40140529 on Build GRCh38
rs386419093 Chr.17: 40140354-40140355 on Build GRCh38
rs943324198 Chr.17: 40140465-40140465 on Build GRCh38
rs892168811 Chr.17: 40140475-40140475 on Build GRCh38
rs757937019 Chr.17: 40140562-40140563 on Build GRCh38
rs777353592 Chr.17: 40140374-40140389 on Build GRCh38
rs559985140 Chr.17: 40140374-40140387 on Build GRCh38
rs1033151782 Chr.17: 40140403-40140403 on Build GRCh38
rs892911228 Chr.17: 40140594-40140594 on Build GRCh38
rs556469120 Chr.17: 40140513-40140513 on Build GRCh38
rs1047967945 Chr.17: 40140515-40140515 on Build GRCh38
rs776240931 Chr.17: 40140502-40140502 on Build GRCh38
rs61340736 Chr.17: 40140351-40140351 on Build GRCh38
rs905492928 Chr.17: 40140275-40140275 on Build GRCh38
rs147866677 Chr.17: 40140261-40140261 on Build GRCh38
rs116535311 Chr.17: 40140533-40140533 on Build GRCh38
rs780831290 Chr.17: 40140543-40140543 on Build GRCh38
rs538168058 Chr.17: 40140305-40140305 on Build GRCh38
rs753976978 Chr.17: 40140524-40140524 on Build GRCh38
rs759646109 Chr.17: 40140591-40140591 on Build GRCh38
rs376795075 Chr.17: 40140507-40140507 on Build GRCh38
rs60339782 Chr.17: 40140350-40140350 on Build GRCh38
rs10644850 Chr.17: 40140339-40140340 on Build GRCh38
rs911878432 Chr.17: 40140184-40140184 on Build GRCh38
rs1001579303 Chr.17: 40140551-40140551 on Build GRCh38
rs900614362 Chr.17: 40140390-40140390 on Build GRCh38
rs370453796 Chr.17: 40140568-40140568 on Build GRCh38
rs573660532 Chr.17: 40140262-40140262 on Build GRCh38
rs955329891 Chr.17: 40140431-40140431 on Build GRCh38
rs35659601 Chr.17: 40140464-40140465 on Build GRCh38
rs899331432 Chr.17: 40140239-40140239 on Build GRCh38
rs778813984 Chr.17: 40140514-40140518 on Build GRCh38
rs911903591 Chr.17: 40140455-40140455 on Build GRCh38
rs1054036678 Chr.17: 40140558-40140558 on Build GRCh38
rs61449096 Chr.17: 40140427-40140430 on Build GRCh38
rs199655625 Chr.17: 40140536-40140536 on Build GRCh38
rs755139876 Chr.17: 40140525-40140525 on Build GRCh38
rs988707748 Chr.17: 40140154-40140154 on Build GRCh38
rs1045278503 Chr.17: 40140304-40140304 on Build GRCh38
rs988633852 Chr.17: 40140425-40140425 on Build GRCh38
rs930773479 Chr.17: 40140247-40140247 on Build GRCh38
rs757845458 Chr.17: 40140540-40140540 on Build GRCh38
rs957215700 Chr.17: 40140419-40140419 on Build GRCh38
rs10644852 Chr.17: 40140385-40140386 on Build GRCh38
rs982794113 Chr.17: 40140570-40140570 on Build GRCh38
rs761851988 Chr.17: 40140521-40140521 on Build GRCh38
rs758715323 Chr.17: 40140530-40140530 on Build GRCh38
rs974461238 Chr.17: 40140204-40140204 on Build GRCh38
rs919973951 Chr.17: 40140206-40140206 on Build GRCh38
rs957163382 Chr.17: 40140147-40140147 on Build GRCh38
rs933896090 Chr.17: 40140171-40140171 on Build GRCh38
rs1054604670 Chr.17: 40140393-40140393 on Build GRCh38
rs1039525189 Chr.17: 40140348-40140348 on Build GRCh38
rs755006932 Chr.17: 40140306-40140306 on Build GRCh38
rs1039492969 Chr.17: 40140236-40140236 on Build GRCh38
rs748999365 Chr.17: 40140563-40140563 on Build GRCh38
rs71152646 Chr.17: 40140374-40140375 on Build GRCh38
rs200244530 Chr.17: 40140520-40140520 on Build GRCh38
rs745384918 Chr.17: 40140548-40140548 on Build GRCh38
rs541790941 Chr.17: 40140336-40140353 on Build GRCh38
rs964114687 Chr.17: 40140190-40140190 on Build GRCh38
rs71152645 Chr.17: 40140349-40140350 on Build GRCh38
rs936098042 Chr.17: 40140392-40140392 on Build GRCh38
rs766366139 Chr.17: 40140523-40140523 on Build GRCh38
rs1020056369 Chr.17: 40140429-40140429 on Build GRCh38
rs913122295 Chr.17: 40140162-40140162 on Build GRCh38
rs1014919984 Chr.17: 40140266-40140266 on Build GRCh38
rs769627462 Chr.17: 40140505-40140505 on Build GRCh38
rs572495306 Chr.17: 40140593-40140593 on Build GRCh38
rs943498815 Chr.17: 40140212-40140212 on Build GRCh38
rs73983012 Chr.17: 40140260-40140260 on Build GRCh38
rs769504102 Chr.17: 40140554-40140554 on Build GRCh38
rs759253952 Chr.17: 40140503-40140503 on Build GRCh38
rs775311641 Chr.17: 40140559-40140559 on Build GRCh38
rs987114129 Chr.17: 40140447-40140447 on Build GRCh38
rs368657427 Chr.17: 40140350-40140353 on Build GRCh38
rs1007324316 Chr.17: 40140527-40140527 on Build GRCh38

View Full Product Details