Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40140513-40141022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC3
Entrez Gene ID
22794
Gene Name
cancer susceptibility candidate 3
Gene Aliases
BTZ, MLN51
UniGene
Hs.743287
Gene Chromosome Location
Chr.17: 40140254-40172178 on Build GRCh38
Transcripts
NM_007359.4 XM_005257163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3516963 c.14G>C p.R5P Chr.17: 40140562-40140562 on Build GRCh38
7108535 c.62C>T p.S21L Chr.17: 40140610-40140610 on Build GRCh38
436516 c.134G>C p.S45T Chr.17: 40140682-40140682 on Build GRCh38
560919 c.29C>T p.S10L Chr.17: 40140577-40140577 on Build GRCh38
5706368 c.83C>G p.S28C Chr.17: 40140631-40140631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745438579 Chr.17: 40140535-40140535 on Build GRCh38
rs1008446856 Chr.17: 40140980-40140980 on Build GRCh38
rs375234919 Chr.17: 40140587-40140587 on Build GRCh38
rs1051363672 Chr.17: 40140808-40140808 on Build GRCh38
rs959123078 Chr.17: 40140812-40140812 on Build GRCh38
rs757937019 Chr.17: 40140562-40140563 on Build GRCh38
rs1025011193 Chr.17: 40140729-40140729 on Build GRCh38
rs778663672 Chr.17: 40140723-40140723 on Build GRCh38
rs964347107 Chr.17: 40140996-40140996 on Build GRCh38
rs531575982 Chr.17: 40140754-40140754 on Build GRCh38
rs116535311 Chr.17: 40140533-40140533 on Build GRCh38
rs780831290 Chr.17: 40140543-40140543 on Build GRCh38
rs771835371 Chr.17: 40140727-40140727 on Build GRCh38
rs543731181 Chr.17: 40140768-40140768 on Build GRCh38
rs778338353 Chr.17: 40140787-40140787 on Build GRCh38
rs775610775 Chr.17: 40140668-40140668 on Build GRCh38
rs1001579303 Chr.17: 40140551-40140551 on Build GRCh38
rs779362937 Chr.17: 40140673-40140681 on Build GRCh38
rs367657738 Chr.17: 40140874-40140874 on Build GRCh38
rs1027042678 Chr.17: 40140721-40140721 on Build GRCh38
rs762268612 Chr.17: 40140689-40140689 on Build GRCh38
rs773759957 Chr.17: 40140809-40140834 on Build GRCh38
rs560982223 Chr.17: 40140623-40140623 on Build GRCh38
rs908044757 Chr.17: 40140718-40140718 on Build GRCh38
rs1009948970 Chr.17: 40140929-40140929 on Build GRCh38
rs1038893730 Chr.17: 40140849-40140849 on Build GRCh38
rs920594638 Chr.17: 40140671-40140671 on Build GRCh38
rs567290073 Chr.17: 40140906-40140906 on Build GRCh38
rs1009204314 Chr.17: 40140641-40140641 on Build GRCh38
rs199655625 Chr.17: 40140536-40140536 on Build GRCh38
rs756753448 Chr.17: 40140700-40140700 on Build GRCh38
rs763067390 Chr.17: 40140669-40140669 on Build GRCh38
rs757845458 Chr.17: 40140540-40140540 on Build GRCh38
rs982794113 Chr.17: 40140570-40140570 on Build GRCh38
rs1045996173 Chr.17: 40140869-40140869 on Build GRCh38
rs756660140 Chr.17: 40140951-40140953 on Build GRCh38
rs770909452 Chr.17: 40140742-40140742 on Build GRCh38
rs768413648 Chr.17: 40140621-40140621 on Build GRCh38
rs892796616 Chr.17: 40140914-40140914 on Build GRCh38
rs747851575 Chr.17: 40140726-40140726 on Build GRCh38
rs753403484 Chr.17: 40140712-40140712 on Build GRCh38
rs769855586 Chr.17: 40140991-40140991 on Build GRCh38
rs565408472 Chr.17: 40140788-40140788 on Build GRCh38
rs766998291 Chr.17: 40140769-40140769 on Build GRCh38
rs890153310 Chr.17: 40140827-40140827 on Build GRCh38
rs200212576 Chr.17: 40140603-40140603 on Build GRCh38
rs1004335609 Chr.17: 40140903-40140903 on Build GRCh38
rs763098180 Chr.17: 40140749-40140749 on Build GRCh38
rs754524245 Chr.17: 40140720-40140720 on Build GRCh38
rs566252028 Chr.17: 40140861-40140861 on Build GRCh38
rs752015716 Chr.17: 40140606-40140606 on Build GRCh38
rs751804046 Chr.17: 40140682-40140682 on Build GRCh38
rs572495306 Chr.17: 40140593-40140593 on Build GRCh38
rs1054086902 Chr.17: 40140886-40140886 on Build GRCh38
rs1027404174 Chr.17: 40140839-40140839 on Build GRCh38
rs943041997 Chr.17: 40140848-40140848 on Build GRCh38
rs750877718 Chr.17: 40140764-40140764 on Build GRCh38
rs769504102 Chr.17: 40140554-40140554 on Build GRCh38
rs775370619 Chr.17: 40140747-40140747 on Build GRCh38
rs746942151 Chr.17: 40140739-40140739 on Build GRCh38
rs1003235427 Chr.17: 40140805-40140805 on Build GRCh38
rs768305986 Chr.17: 40140566-40140566 on Build GRCh38
rs558372528 Chr.17: 40140875-40140875 on Build GRCh38
rs769830307 Chr.17: 40140635-40140635 on Build GRCh38
rs760657758 Chr.17: 40140632-40140632 on Build GRCh38
rs892911228 Chr.17: 40140594-40140594 on Build GRCh38
rs1041407667 Chr.17: 40140937-40140937 on Build GRCh38
rs757646540 Chr.17: 40140609-40140609 on Build GRCh38
rs927566160 Chr.17: 40140872-40140872 on Build GRCh38
rs779698182 Chr.17: 40140708-40140708 on Build GRCh38
rs759646109 Chr.17: 40140591-40140591 on Build GRCh38
rs537807425 Chr.17: 40140938-40140938 on Build GRCh38
rs781765686 Chr.17: 40140612-40140612 on Build GRCh38
rs578093163 Chr.17: 40140901-40140901 on Build GRCh38
rs773157954 Chr.17: 40140627-40140627 on Build GRCh38
rs750930900 Chr.17: 40140698-40140698 on Build GRCh38
rs777582439 Chr.17: 40140728-40140728 on Build GRCh38
rs945143379 Chr.17: 40140793-40140793 on Build GRCh38
rs769668604 Chr.17: 40140921-40140921 on Build GRCh38
rs532751417 Chr.17: 40140789-40140789 on Build GRCh38
rs768144459 Chr.17: 40140805-40140805 on Build GRCh38
rs370453796 Chr.17: 40140568-40140568 on Build GRCh38
rs758684260 Chr.17: 40140601-40140601 on Build GRCh38
rs748293528 Chr.17: 40140829-40140829 on Build GRCh38
rs767990289 Chr.17: 40140762-40140762 on Build GRCh38
rs1054036678 Chr.17: 40140558-40140558 on Build GRCh38
rs898959999 Chr.17: 40140716-40140716 on Build GRCh38
rs906190516 Chr.17: 40140800-40140800 on Build GRCh38
rs913627525 Chr.17: 40140792-40140793 on Build GRCh38
rs768086249 Chr.17: 40140690-40140690 on Build GRCh38
rs747983753 Chr.17: 40140624-40140624 on Build GRCh38
rs571403877 Chr.17: 40140990-40140990 on Build GRCh38
rs774654065 Chr.17: 40140755-40140755 on Build GRCh38
rs556430091 Chr.17: 40140951-40140951 on Build GRCh38
rs764387381 Chr.17: 40140675-40140675 on Build GRCh38
rs1024980060 Chr.17: 40140907-40140907 on Build GRCh38
rs973143783 Chr.17: 40140816-40140816 on Build GRCh38
rs371476223 Chr.17: 40140912-40140912 on Build GRCh38
rs748999365 Chr.17: 40140563-40140563 on Build GRCh38
rs887188563 Chr.17: 40140887-40140887 on Build GRCh38
rs890978201 Chr.17: 40140945-40140945 on Build GRCh38
rs1033831586 Chr.17: 40140810-40140810 on Build GRCh38
rs745384918 Chr.17: 40140548-40140548 on Build GRCh38
rs547629691 Chr.17: 40140801-40140801 on Build GRCh38
rs1009937642 Chr.17: 40140607-40140607 on Build GRCh38
rs746704281 Chr.17: 40140765-40140766 on Build GRCh38
rs184119888 Chr.17: 40140613-40140613 on Build GRCh38
rs891972517 Chr.17: 40140644-40140644 on Build GRCh38
rs899091919 Chr.17: 40140856-40140856 on Build GRCh38
rs748890511 Chr.17: 40140614-40140614 on Build GRCh38
rs775311641 Chr.17: 40140559-40140559 on Build GRCh38
rs761989441 Chr.17: 40140756-40140756 on Build GRCh38
rs970675644 Chr.17: 40140936-40140936 on Build GRCh38

View Full Product Details