Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40360477-40361017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJD3
Entrez Gene ID
125111
Gene Name
gap junction protein delta 3
Gene Aliases
CX31.9, Cx30.2, GJA11, GJC1
UniGene
Hs.444663
Gene Chromosome Location
Chr.17: 40360652-40364693 on Build GRCh38
Transcripts
NM_152219.3

SNP Detail

SNP ID Chromosome Location
rs528185342 Chr.17: 40360655-40360655 on Build GRCh38
rs574061081 Chr.17: 40360972-40360972 on Build GRCh38
rs1012832063 Chr.17: 40360987-40360987 on Build GRCh38
rs941780587 Chr.17: 40360706-40360706 on Build GRCh38
rs555834407 Chr.17: 40360968-40360968 on Build GRCh38
rs942199199 Chr.17: 40360842-40360842 on Build GRCh38
rs546146500 Chr.17: 40360885-40360885 on Build GRCh38
rs988371419 Chr.17: 40360505-40360505 on Build GRCh38
rs1037414672 Chr.17: 40360877-40360877 on Build GRCh38
rs923545345 Chr.17: 40360536-40360536 on Build GRCh38
rs1050541676 Chr.17: 40360679-40360679 on Build GRCh38
rs1037888329 Chr.17: 40360715-40360715 on Build GRCh38
rs190215142 Chr.17: 40360725-40360725 on Build GRCh38
rs906535154 Chr.17: 40360942-40360942 on Build GRCh38
rs15685 Chr.17: 40360719-40360719 on Build GRCh38
rs956710087 Chr.17: 40360798-40360798 on Build GRCh38
rs779462487 Chr.17: 40360538-40360538 on Build GRCh38
rs986383974 Chr.17: 40360824-40360824 on Build GRCh38
rs1057188528 Chr.17: 40360965-40360965 on Build GRCh38
rs1048569968 Chr.17: 40360879-40360879 on Build GRCh38
rs540702547 Chr.17: 40360768-40360770 on Build GRCh38
rs753632288 Chr.17: 40360986-40360986 on Build GRCh38
rs746215561 Chr.17: 40360684-40360684 on Build GRCh38
rs933464800 Chr.17: 40360637-40360637 on Build GRCh38
rs928956607 Chr.17: 40360932-40360932 on Build GRCh38
rs908752366 Chr.17: 40360891-40360891 on Build GRCh38
rs796969688 Chr.17: 40360735-40360735 on Build GRCh38
rs35530628 Chr.17: 40360757-40360757 on Build GRCh38
rs567701064 Chr.17: 40360723-40360723 on Build GRCh38
rs954942577 Chr.17: 40360819-40360819 on Build GRCh38
rs879157798 Chr.17: 40360677-40360677 on Build GRCh38
rs887386162 Chr.17: 40360524-40360524 on Build GRCh38
rs573306496 Chr.17: 40360905-40360905 on Build GRCh38
rs1004591280 Chr.17: 40360540-40360540 on Build GRCh38
rs570455416 Chr.17: 40360528-40360528 on Build GRCh38
rs554266718 Chr.17: 40360630-40360642 on Build GRCh38
rs1047126140 Chr.17: 40360913-40360913 on Build GRCh38
rs1000923898 Chr.17: 40360768-40360768 on Build GRCh38
rs920367324 Chr.17: 40360833-40360833 on Build GRCh38
rs1057024502 Chr.17: 40360730-40360730 on Build GRCh38
rs988562871 Chr.17: 40360803-40360803 on Build GRCh38
rs540713680 Chr.17: 40360926-40360926 on Build GRCh38
rs940287292 Chr.17: 40360912-40360912 on Build GRCh38
rs1012866523 Chr.17: 40360740-40360740 on Build GRCh38
rs1024981630 Chr.17: 40360591-40360591 on Build GRCh38
rs1032844322 Chr.17: 40360788-40360788 on Build GRCh38
rs910710484 Chr.17: 40360829-40360829 on Build GRCh38
rs34247423 Chr.17: 40360829-40360830 on Build GRCh38
rs1002241443 Chr.17: 40360636-40360636 on Build GRCh38
rs968574750 Chr.17: 40360765-40360765 on Build GRCh38
rs182843306 Chr.17: 40360816-40360816 on Build GRCh38
rs779818411 Chr.17: 40360949-40360949 on Build GRCh38

View Full Product Details