Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40362081-40362581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJD3
Entrez Gene ID
125111
Gene Name
gap junction protein delta 3
Gene Aliases
CX31.9, Cx30.2, GJA11, GJC1
UniGene
Hs.444663
Gene Chromosome Location
Chr.17: 40360652-40364693 on Build GRCh38
Transcripts
NM_152219.3

SNP Detail

SNP ID Chromosome Location
rs899204081 Chr.17: 40362140-40362140 on Build GRCh38
rs1018003487 Chr.17: 40362129-40362129 on Build GRCh38
rs1039152531 Chr.17: 40362476-40362476 on Build GRCh38
rs562497551 Chr.17: 40362339-40362339 on Build GRCh38
rs989092595 Chr.17: 40362381-40362381 on Build GRCh38
rs751040394 Chr.17: 40362530-40362533 on Build GRCh38
rs1034092107 Chr.17: 40362167-40362170 on Build GRCh38
rs976151439 Chr.17: 40362302-40362302 on Build GRCh38
rs780341429 Chr.17: 40362443-40362443 on Build GRCh38
rs113876371 Chr.17: 40362477-40362477 on Build GRCh38
rs940604470 Chr.17: 40362496-40362496 on Build GRCh38
rs923933623 Chr.17: 40362264-40362273 on Build GRCh38
rs943454152 Chr.17: 40362459-40362459 on Build GRCh38
rs1022517207 Chr.17: 40362219-40362219 on Build GRCh38
rs911076917 Chr.17: 40362457-40362457 on Build GRCh38
rs528911672 Chr.17: 40362321-40362321 on Build GRCh38
rs913164656 Chr.17: 40362406-40362406 on Build GRCh38
rs533110960 Chr.17: 40362405-40362405 on Build GRCh38
rs899122746 Chr.17: 40362480-40362480 on Build GRCh38
rs966105000 Chr.17: 40362269-40362269 on Build GRCh38
rs994866390 Chr.17: 40362517-40362517 on Build GRCh38
rs1002571863 Chr.17: 40362167-40362168 on Build GRCh38
rs935685833 Chr.17: 40362379-40362379 on Build GRCh38
rs532772535 Chr.17: 40362265-40362265 on Build GRCh38
rs1024508752 Chr.17: 40362186-40362186 on Build GRCh38
rs552476516 Chr.17: 40362409-40362409 on Build GRCh38
rs950924694 Chr.17: 40362155-40362155 on Build GRCh38
rs561458368 Chr.17: 40362274-40362274 on Build GRCh38
rs969778729 Chr.17: 40362189-40362189 on Build GRCh38
rs1051772939 Chr.17: 40362107-40362107 on Build GRCh38
rs747155274 Chr.17: 40362533-40362533 on Build GRCh38
rs549152261 Chr.17: 40362530-40362530 on Build GRCh38
rs968713798 Chr.17: 40362251-40362251 on Build GRCh38
rs979890653 Chr.17: 40362193-40362193 on Build GRCh38
rs550209221 Chr.17: 40362331-40362331 on Build GRCh38
rs995489554 Chr.17: 40362141-40362141 on Build GRCh38
rs993010552 Chr.17: 40362162-40362162 on Build GRCh38
rs570212474 Chr.17: 40362253-40362253 on Build GRCh38
rs547132886 Chr.17: 40362126-40362126 on Build GRCh38
rs1004521322 Chr.17: 40362138-40362138 on Build GRCh38
rs560549224 Chr.17: 40362108-40362108 on Build GRCh38
rs193258767 Chr.17: 40362516-40362516 on Build GRCh38
rs1050871126 Chr.17: 40362429-40362429 on Build GRCh38
rs367751199 Chr.17: 40362147-40362147 on Build GRCh38
rs922035217 Chr.17: 40362328-40362328 on Build GRCh38
rs182077639 Chr.17: 40362531-40362531 on Build GRCh38
rs115475812 Chr.17: 40362258-40362258 on Build GRCh38

View Full Product Details