Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56956673-56957211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8B
Entrez Gene ID
732
Gene Name
complement component 8, beta polypeptide
Gene Aliases
C82
UniGene
Hs.391835
Gene Chromosome Location
Chr.1: 56929210-56974383 on Build GRCh38
Transcripts
NM_000066.3 NM_001278543.1 NM_001278544.1 XM_017002235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5478970 c.391+6A>C p.? Chr.1: 56956763-56956763 on Build GRCh38
3491227 c.292G>A p.E98K Chr.1: 56956868-56956868 on Build GRCh38
3491228 c.291G>A p.G97G Chr.1: 56956869-56956869 on Build GRCh38
1685390 c.304T>C p.F102L Chr.1: 56956856-56956856 on Build GRCh38
1685415 c.306C>T p.F102F Chr.1: 56956854-56956854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753002884 Chr.1: 56956812-56956812 on Build GRCh38
rs747647469 Chr.1: 56956798-56956798 on Build GRCh38
rs1000787260 Chr.1: 56957146-56957146 on Build GRCh38
rs146187042 Chr.1: 56956889-56956889 on Build GRCh38
rs1031822747 Chr.1: 56957157-56957157 on Build GRCh38
rs777968221 Chr.1: 56956743-56956743 on Build GRCh38
rs757827839 Chr.1: 56956749-56956749 on Build GRCh38
rs937811818 Chr.1: 56957114-56957114 on Build GRCh38
rs981880625 Chr.1: 56957107-56957107 on Build GRCh38
rs897926779 Chr.1: 56956964-56956964 on Build GRCh38
rs531949198 Chr.1: 56957057-56957057 on Build GRCh38
rs751686321 Chr.1: 56956834-56956834 on Build GRCh38
rs530181040 Chr.1: 56956905-56956905 on Build GRCh38
rs750982520 Chr.1: 56956936-56956936 on Build GRCh38
rs1042370531 Chr.1: 56956873-56956873 on Build GRCh38
rs746455376 Chr.1: 56956886-56956886 on Build GRCh38
rs12067507 Chr.1: 56956838-56956838 on Build GRCh38
rs548731487 Chr.1: 56957063-56957063 on Build GRCh38
rs867837249 Chr.1: 56956868-56956868 on Build GRCh38
rs547100051 Chr.1: 56957147-56957147 on Build GRCh38
rs752741094 Chr.1: 56956737-56956737 on Build GRCh38
rs904741068 Chr.1: 56957137-56957137 on Build GRCh38
rs952374036 Chr.1: 56956744-56956744 on Build GRCh38
rs767944212 Chr.1: 56956933-56956933 on Build GRCh38
rs758039686 Chr.1: 56956870-56956870 on Build GRCh38
rs114982349 Chr.1: 56956930-56956930 on Build GRCh38
rs1035626416 Chr.1: 56956817-56956817 on Build GRCh38
rs545739274 Chr.1: 56956830-56956830 on Build GRCh38
rs201509703 Chr.1: 56956903-56956903 on Build GRCh38
rs374795894 Chr.1: 56956899-56956899 on Build GRCh38
rs533596476 Chr.1: 56957123-56957123 on Build GRCh38
rs934988680 Chr.1: 56956712-56956712 on Build GRCh38
rs1023160619 Chr.1: 56956846-56956846 on Build GRCh38
rs372968576 Chr.1: 56956824-56956824 on Build GRCh38
rs778583924 Chr.1: 56956783-56956783 on Build GRCh38
rs1015225501 Chr.1: 56957064-56957064 on Build GRCh38
rs150022116 Chr.1: 56956799-56956799 on Build GRCh38
rs759678226 Chr.1: 56956728-56956728 on Build GRCh38
rs747849141 Chr.1: 56956864-56956864 on Build GRCh38
rs969282586 Chr.1: 56956850-56956850 on Build GRCh38
rs552338985 Chr.1: 56957005-56957005 on Build GRCh38
rs895806242 Chr.1: 56957100-56957100 on Build GRCh38
rs373375237 Chr.1: 56956842-56956842 on Build GRCh38
rs970886088 Chr.1: 56957066-56957066 on Build GRCh38
rs760669928 Chr.1: 56956805-56956805 on Build GRCh38
rs888935609 Chr.1: 56956789-56956789 on Build GRCh38
rs758511199 Chr.1: 56956741-56956741 on Build GRCh38
rs767296454 Chr.1: 56956800-56956800 on Build GRCh38
rs55749099 Chr.1: 56956779-56956779 on Build GRCh38
rs567801377 Chr.1: 56956729-56956729 on Build GRCh38
rs761526316 Chr.1: 56957055-56957055 on Build GRCh38
rs149147808 Chr.1: 56956807-56956807 on Build GRCh38
rs766218381 Chr.1: 56956722-56956722 on Build GRCh38
rs764047095 Chr.1: 56957021-56957021 on Build GRCh38
rs987826628 Chr.1: 56956897-56956897 on Build GRCh38
rs768010940 Chr.1: 56956839-56956839 on Build GRCh38
rs759864328 Chr.1: 56956755-56956756 on Build GRCh38
rs568338938 Chr.1: 56957068-56957068 on Build GRCh38
rs766831248 Chr.1: 56956948-56956948 on Build GRCh38
rs1052069348 Chr.1: 56956733-56956733 on Build GRCh38
rs753659239 Chr.1: 56956724-56956724 on Build GRCh38
rs1057081665 Chr.1: 56957062-56957062 on Build GRCh38
rs781478248 Chr.1: 56956753-56956753 on Build GRCh38
rs199745832 Chr.1: 56956808-56956808 on Build GRCh38
rs746177242 Chr.1: 56956757-56956757 on Build GRCh38
rs368854622 Chr.1: 56956745-56956745 on Build GRCh38
rs138368595 Chr.1: 56956884-56956884 on Build GRCh38
rs144804643 Chr.1: 56957148-56957148 on Build GRCh38
rs371013393 Chr.1: 56956755-56956755 on Build GRCh38
rs925004421 Chr.1: 56956708-56956708 on Build GRCh38
rs55655103 Chr.1: 56956700-56956700 on Build GRCh38
rs532544319 Chr.1: 56956966-56956966 on Build GRCh38
rs765027143 Chr.1: 56957111-56957111 on Build GRCh38
rs868322115 Chr.1: 56957132-56957132 on Build GRCh38
rs563177630 Chr.1: 56957049-56957049 on Build GRCh38
rs868422544 Chr.1: 56956764-56956764 on Build GRCh38
rs956466753 Chr.1: 56957181-56957181 on Build GRCh38
rs199751348 Chr.1: 56956922-56956922 on Build GRCh38
rs151032934 Chr.1: 56956863-56956863 on Build GRCh38
rs753802566 Chr.1: 56956941-56956941 on Build GRCh38
rs774905937 Chr.1: 56956858-56956858 on Build GRCh38
rs776590472 Chr.1: 56956892-56956892 on Build GRCh38
rs768211721 Chr.1: 56956774-56956774 on Build GRCh38
rs1013579 Chr.1: 56956811-56956811 on Build GRCh38
rs952404894 Chr.1: 56957016-56957016 on Build GRCh38
rs987876849 Chr.1: 56957187-56957187 on Build GRCh38
rs756465335 Chr.1: 56956861-56956861 on Build GRCh38
rs763243467 Chr.1: 56956823-56956823 on Build GRCh38
rs770784749 Chr.1: 56956806-56956806 on Build GRCh38
rs767381902 Chr.1: 56956880-56956881 on Build GRCh38
rs774090414 Chr.1: 56956746-56956746 on Build GRCh38
rs113900156 Chr.1: 56956902-56956902 on Build GRCh38

View Full Product Details