Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62454950-62455464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK7
Entrez Gene ID
85440
Gene Name
dedicator of cytokinesis 7
Gene Aliases
EIEE23, ZIR2
UniGene
Hs.744927
Gene Chromosome Location
Chr.1: 62454726-62688368 on Build GRCh38
Transcripts
NM_001271999.1 NM_001272000.1 NM_001272001.1 NM_033407.3 XM_005271292.2 XM_011542326.2 XM_011542327.2 XM_011542328.2 XM_017002639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6648359 c.6288-6delT p.? Chr.1: 62455462-62455462 on Build GRCh38
6648360 c.6348-6delT p.? Chr.1: 62455462-62455462 on Build GRCh38
4810184 c.6357C>T p.F2119F Chr.1: 62455447-62455447 on Build GRCh38
5034261 c.6294C>T p.S2098S Chr.1: 62455450-62455450 on Build GRCh38
5034262 c.6354C>T p.S2118S Chr.1: 62455450-62455450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777107297 Chr.1: 62455313-62455313 on Build GRCh38
rs971877567 Chr.1: 62455042-62455042 on Build GRCh38
rs879178909 Chr.1: 62455153-62455153 on Build GRCh38
rs768300646 Chr.1: 62455004-62455004 on Build GRCh38
rs547645113 Chr.1: 62455280-62455280 on Build GRCh38
rs201452223 Chr.1: 62455430-62455430 on Build GRCh38
rs896634193 Chr.1: 62455224-62455224 on Build GRCh38
rs565541256 Chr.1: 62455334-62455336 on Build GRCh38
rs186911256 Chr.1: 62455060-62455060 on Build GRCh38
rs866803433 Chr.1: 62455116-62455116 on Build GRCh38
rs765897636 Chr.1: 62455351-62455352 on Build GRCh38
rs944929053 Chr.1: 62455380-62455380 on Build GRCh38
rs1019024296 Chr.1: 62454977-62454979 on Build GRCh38
rs13455 Chr.1: 62455332-62455332 on Build GRCh38
rs956640129 Chr.1: 62455268-62455268 on Build GRCh38
rs764960952 Chr.1: 62455394-62455394 on Build GRCh38
rs750610768 Chr.1: 62455378-62455378 on Build GRCh38
rs962062028 Chr.1: 62454997-62454997 on Build GRCh38
rs776419206 Chr.1: 62455324-62455324 on Build GRCh38
rs538808802 Chr.1: 62455056-62455056 on Build GRCh38
rs889360870 Chr.1: 62455169-62455169 on Build GRCh38
rs375171813 Chr.1: 62455374-62455374 on Build GRCh38
rs968459790 Chr.1: 62455254-62455254 on Build GRCh38
rs751519461 Chr.1: 62455436-62455436 on Build GRCh38
rs530096458 Chr.1: 62455334-62455334 on Build GRCh38
rs763567676 Chr.1: 62455184-62455184 on Build GRCh38
rs145548218 Chr.1: 62455425-62455425 on Build GRCh38
rs767647635 Chr.1: 62455376-62455376 on Build GRCh38
rs941673770 Chr.1: 62455182-62455182 on Build GRCh38
rs1052634851 Chr.1: 62455120-62455122 on Build GRCh38
rs11207971 Chr.1: 62455093-62455093 on Build GRCh38
rs748011581 Chr.1: 62455293-62455293 on Build GRCh38
rs1023343479 Chr.1: 62455246-62455246 on Build GRCh38
rs745556451 Chr.1: 62455328-62455328 on Build GRCh38
rs577395735 Chr.1: 62455304-62455304 on Build GRCh38
rs908891220 Chr.1: 62455173-62455173 on Build GRCh38
rs1056591995 Chr.1: 62455215-62455215 on Build GRCh38
rs775948328 Chr.1: 62455344-62455344 on Build GRCh38
rs747120406 Chr.1: 62455312-62455315 on Build GRCh38
rs1051150739 Chr.1: 62455147-62455147 on Build GRCh38
rs1038597975 Chr.1: 62455190-62455190 on Build GRCh38
rs770622934 Chr.1: 62455321-62455321 on Build GRCh38
rs994376451 Chr.1: 62455203-62455203 on Build GRCh38
rs576665826 Chr.1: 62455003-62455003 on Build GRCh38
rs975985784 Chr.1: 62455276-62455276 on Build GRCh38
rs764746776 Chr.1: 62455307-62455309 on Build GRCh38
rs778718024 Chr.1: 62455271-62455271 on Build GRCh38
rs754760811 Chr.1: 62455266-62455266 on Build GRCh38
rs576810730 Chr.1: 62455225-62455225 on Build GRCh38
rs763331531 Chr.1: 62455346-62455346 on Build GRCh38
rs761558847 Chr.1: 62455241-62455241 on Build GRCh38
rs138319770 Chr.1: 62455229-62455229 on Build GRCh38
rs923818143 Chr.1: 62455058-62455058 on Build GRCh38
rs971743642 Chr.1: 62455392-62455392 on Build GRCh38
rs993967581 Chr.1: 62455219-62455219 on Build GRCh38
rs150100104 Chr.1: 62455117-62455117 on Build GRCh38
rs79913813 Chr.1: 62455259-62455259 on Build GRCh38
rs760933841 Chr.1: 62455385-62455385 on Build GRCh38
rs189241213 Chr.1: 62455199-62455199 on Build GRCh38
rs968051630 Chr.1: 62455027-62455027 on Build GRCh38
rs962113150 Chr.1: 62455209-62455209 on Build GRCh38
rs893988993 Chr.1: 62455128-62455128 on Build GRCh38
rs949734543 Chr.1: 62455096-62455096 on Build GRCh38
rs752567493 Chr.1: 62455403-62455403 on Build GRCh38
rs1030606241 Chr.1: 62455285-62455285 on Build GRCh38
rs114747052 Chr.1: 62455318-62455318 on Build GRCh38
rs1050165824 Chr.1: 62455133-62455133 on Build GRCh38
rs541826520 Chr.1: 62455363-62455363 on Build GRCh38
rs1045423646 Chr.1: 62455108-62455108 on Build GRCh38
rs559461705 Chr.1: 62455333-62455333 on Build GRCh38

View Full Product Details