Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62821048-62821547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG4C
Entrez Gene ID
84938
Gene Name
autophagy related 4C cysteine peptidase
Gene Aliases
APG4-C, APG4C, AUTL1, AUTL3
UniGene
Hs.7353
Gene Chromosome Location
Chr.1: 62784106-62865270 on Build GRCh38
Transcripts
NM_032852.3 NM_178221.2 XM_005271288.2 XM_011542313.2 XM_017002600.1 XM_017002601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7327789 c.726-10A>G p.? Chr.1: 62821129-62821129 on Build GRCh38
5511520 c.771_774delTATT p.I258fs*13 Chr.1: 62821184-62821187 on Build GRCh38
5511521 c.2_5delTATT p.F2fs*13 Chr.1: 62821184-62821187 on Build GRCh38
7327788 c.726-10A>G p.? Chr.1: 62821129-62821129 on Build GRCh38
6482633 c.764G>A p.G255E Chr.1: 62821177-62821177 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751030760 Chr.1: 62821116-62821116 on Build GRCh38
rs1013839815 Chr.1: 62821502-62821502 on Build GRCh38
rs777866917 Chr.1: 62821297-62821297 on Build GRCh38
rs769027073 Chr.1: 62821223-62821223 on Build GRCh38
rs532493005 Chr.1: 62821079-62821079 on Build GRCh38
rs896707161 Chr.1: 62821497-62821497 on Build GRCh38
rs796538435 Chr.1: 62821280-62821280 on Build GRCh38
rs754210252 Chr.1: 62821148-62821148 on Build GRCh38
rs777963015 Chr.1: 62821218-62821218 on Build GRCh38
rs775383471 Chr.1: 62821251-62821251 on Build GRCh38
rs150523110 Chr.1: 62821091-62821091 on Build GRCh38
rs764158005 Chr.1: 62821108-62821108 on Build GRCh38
rs1026928605 Chr.1: 62821514-62821514 on Build GRCh38
rs1002455504 Chr.1: 62821080-62821080 on Build GRCh38
rs372248687 Chr.1: 62821128-62821128 on Build GRCh38
rs894127585 Chr.1: 62821122-62821122 on Build GRCh38
rs938206376 Chr.1: 62821424-62821424 on Build GRCh38
rs747512962 Chr.1: 62821158-62821158 on Build GRCh38
rs761020296 Chr.1: 62821090-62821090 on Build GRCh38
rs140019854 Chr.1: 62821187-62821187 on Build GRCh38
rs199941221 Chr.1: 62821092-62821092 on Build GRCh38
rs977177161 Chr.1: 62821200-62821200 on Build GRCh38
rs758399411 Chr.1: 62821197-62821197 on Build GRCh38
rs1055310338 Chr.1: 62821431-62821431 on Build GRCh38
rs750041221 Chr.1: 62821142-62821142 on Build GRCh38
rs1023995407 Chr.1: 62821137-62821137 on Build GRCh38
rs776972479 Chr.1: 62821184-62821187 on Build GRCh38
rs755442365 Chr.1: 62821177-62821177 on Build GRCh38
rs570370435 Chr.1: 62821258-62821258 on Build GRCh38
rs558841965 Chr.1: 62821420-62821423 on Build GRCh38
rs557229180 Chr.1: 62821455-62821455 on Build GRCh38
rs755824271 Chr.1: 62821143-62821143 on Build GRCh38
rs769253661 Chr.1: 62821100-62821100 on Build GRCh38
rs984880106 Chr.1: 62821254-62821254 on Build GRCh38
rs536838871 Chr.1: 62821299-62821299 on Build GRCh38
rs570466673 Chr.1: 62821364-62821364 on Build GRCh38
rs779315690 Chr.1: 62821182-62821182 on Build GRCh38
rs190127657 Chr.1: 62821321-62821321 on Build GRCh38
rs1011769066 Chr.1: 62821401-62821401 on Build GRCh38
rs938606832 Chr.1: 62821350-62821350 on Build GRCh38
rs150686434 Chr.1: 62821147-62821147 on Build GRCh38
rs1042797849 Chr.1: 62821427-62821427 on Build GRCh38
rs534403339 Chr.1: 62821508-62821508 on Build GRCh38
rs747779205 Chr.1: 62821519-62821519 on Build GRCh38
rs940098361 Chr.1: 62821324-62821324 on Build GRCh38
rs867449557 Chr.1: 62821156-62821156 on Build GRCh38
rs1038454837 Chr.1: 62821348-62821348 on Build GRCh38
rs966239063 Chr.1: 62821162-62821162 on Build GRCh38
rs371292038 Chr.1: 62821254-62821254 on Build GRCh38
rs767095073 Chr.1: 62821133-62821133 on Build GRCh38
rs966729496 Chr.1: 62821489-62821489 on Build GRCh38
rs193285646 Chr.1: 62821123-62821123 on Build GRCh38
rs1032838796 Chr.1: 62821216-62821216 on Build GRCh38
rs747167914 Chr.1: 62821222-62821222 on Build GRCh38
rs1006522298 Chr.1: 62821444-62821444 on Build GRCh38
rs370787693 Chr.1: 62821234-62821234 on Build GRCh38
rs920086696 Chr.1: 62821400-62821400 on Build GRCh38
rs975339577 Chr.1: 62821248-62821248 on Build GRCh38
rs185072784 Chr.1: 62821233-62821233 on Build GRCh38
rs889472785 Chr.1: 62821430-62821430 on Build GRCh38

View Full Product Details