Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 63323290-63323829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXD3
Entrez Gene ID
27022
Gene Name
forkhead box D3
Gene Aliases
AIS1, Genesis, HFH2, VAMAS2
UniGene
Hs.546573
Gene Chromosome Location
Chr.1: 63323059-63325126 on Build GRCh38
Transcripts
NM_012183.2 NR_121636.1 NR_121637.1

Gene Symbol
FOXD3-AS1
Entrez Gene ID
100996301
Gene Name
FOXD3 antisense RNA 1 (head to head)
Gene Aliases
pasFOXD3
UniGene
Gene Chromosome Location
Chr.1: 63320884-63324441 on Build GRCh38
Transcripts
NM_012183.2 NR_121636.1 NR_121637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1343818 c.568C>T p.R190C Chr.1: 63323626-63323626 on Build GRCh38
6434719 c.721C>A p.Q241K Chr.1: 63323779-63323779 on Build GRCh38
4990580 c.624delC p.P209fs*44 Chr.1: 63323682-63323682 on Build GRCh38
123219 c.434C>G p.S145W Chr.1: 63323492-63323492 on Build GRCh38
6384270 c.650T>A p.L217Q Chr.1: 63323708-63323708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943682073 Chr.1: 63323456-63323456 on Build GRCh38
rs760636951 Chr.1: 63323676-63323676 on Build GRCh38
rs141605755 Chr.1: 63323712-63323712 on Build GRCh38
rs751333747 Chr.1: 63323700-63323700 on Build GRCh38
rs759762578 Chr.1: 63323691-63323691 on Build GRCh38
rs745577800 Chr.1: 63323413-63323413 on Build GRCh38
rs370359586 Chr.1: 63323604-63323604 on Build GRCh38
rs1012272339 Chr.1: 63323312-63323312 on Build GRCh38
rs868207373 Chr.1: 63323555-63323555 on Build GRCh38
rs374869492 Chr.1: 63323802-63323802 on Build GRCh38
rs897045765 Chr.1: 63323539-63323539 on Build GRCh38
rs769354523 Chr.1: 63323487-63323487 on Build GRCh38
rs114453593 Chr.1: 63323697-63323697 on Build GRCh38
rs775039432 Chr.1: 63323430-63323430 on Build GRCh38
rs943515219 Chr.1: 63323586-63323586 on Build GRCh38
rs755691977 Chr.1: 63323580-63323580 on Build GRCh38
rs912263473 Chr.1: 63323451-63323451 on Build GRCh38
rs546862550 Chr.1: 63323365-63323365 on Build GRCh38
rs747622091 Chr.1: 63323799-63323799 on Build GRCh38
rs758342193 Chr.1: 63323377-63323377 on Build GRCh38
rs2274188 Chr.1: 63323344-63323344 on Build GRCh38
rs769191390 Chr.1: 63323509-63323509 on Build GRCh38
rs906546657 Chr.1: 63323609-63323609 on Build GRCh38
rs778117598 Chr.1: 63323796-63323796 on Build GRCh38
rs755670470 Chr.1: 63323733-63323733 on Build GRCh38
rs951092624 Chr.1: 63323416-63323416 on Build GRCh38
rs764730613 Chr.1: 63323424-63323424 on Build GRCh38
rs775959596 Chr.1: 63323502-63323502 on Build GRCh38
rs373332012 Chr.1: 63323724-63323724 on Build GRCh38
rs1043858064 Chr.1: 63323313-63323313 on Build GRCh38
rs752508967 Chr.1: 63323445-63323445 on Build GRCh38
rs145695569 Chr.1: 63323437-63323437 on Build GRCh38
rs765020342 Chr.1: 63323442-63323442 on Build GRCh38
rs778638857 Chr.1: 63323351-63323351 on Build GRCh38
rs760898422 Chr.1: 63323525-63323525 on Build GRCh38
rs147290223 Chr.1: 63323808-63323808 on Build GRCh38
rs772524720 Chr.1: 63323792-63323792 on Build GRCh38
rs750660492 Chr.1: 63323338-63323338 on Build GRCh38
rs773299639 Chr.1: 63323656-63323656 on Build GRCh38
rs754690288 Chr.1: 63323773-63323773 on Build GRCh38
rs1012319757 Chr.1: 63323572-63323572 on Build GRCh38
rs780576074 Chr.1: 63323459-63323459 on Build GRCh38
rs780705299 Chr.1: 63323570-63323570 on Build GRCh38
rs758178965 Chr.1: 63323532-63323532 on Build GRCh38
rs757158520 Chr.1: 63323562-63323562 on Build GRCh38
rs772474698 Chr.1: 63323655-63323655 on Build GRCh38
rs756791555 Chr.1: 63323454-63323454 on Build GRCh38
rs748488056 Chr.1: 63323791-63323791 on Build GRCh38
rs756292104 Chr.1: 63323438-63323438 on Build GRCh38
rs763946436 Chr.1: 63323547-63323547 on Build GRCh38
rs776336618 Chr.1: 63323410-63323410 on Build GRCh38
rs770399172 Chr.1: 63323405-63323405 on Build GRCh38
rs201881536 Chr.1: 63323419-63323419 on Build GRCh38
rs771058920 Chr.1: 63323683-63323683 on Build GRCh38
rs761770214 Chr.1: 63323705-63323705 on Build GRCh38
rs144838490 Chr.1: 63323752-63323752 on Build GRCh38
rs747009146 Chr.1: 63323453-63323453 on Build GRCh38
rs761929539 Chr.1: 63323431-63323431 on Build GRCh38
rs569969907 Chr.1: 63323596-63323596 on Build GRCh38
rs751451513 Chr.1: 63323550-63323550 on Build GRCh38
rs978759068 Chr.1: 63323329-63323329 on Build GRCh38
rs1008242321 Chr.1: 63323334-63323334 on Build GRCh38
rs200810432 Chr.1: 63323807-63323807 on Build GRCh38
rs762293981 Chr.1: 63323513-63323513 on Build GRCh38
rs779367590 Chr.1: 63323784-63323784 on Build GRCh38
rs746549793 Chr.1: 63323401-63323401 on Build GRCh38
rs779775701 Chr.1: 63323751-63323751 on Build GRCh38
rs763505896 Chr.1: 63323421-63323421 on Build GRCh38
rs151021417 Chr.1: 63323575-63323575 on Build GRCh38
rs557172178 Chr.1: 63323524-63323524 on Build GRCh38
rs770933954 Chr.1: 63323804-63323804 on Build GRCh38
rs202186939 Chr.1: 63323417-63323417 on Build GRCh38
rs754096073 Chr.1: 63323529-63323529 on Build GRCh38
rs28553389 Chr.1: 63323593-63323593 on Build GRCh38
rs774984139 Chr.1: 63323512-63323512 on Build GRCh38
rs931151703 Chr.1: 63323544-63323544 on Build GRCh38
rs776832333 Chr.1: 63323685-63323685 on Build GRCh38
rs758289291 Chr.1: 63323449-63323449 on Build GRCh38
rs777213266 Chr.1: 63323394-63323394 on Build GRCh38
rs779423935 Chr.1: 63323608-63323608 on Build GRCh38
rs779831246 Chr.1: 63323583-63323583 on Build GRCh38
rs749641121 Chr.1: 63323505-63323505 on Build GRCh38
rs767566861 Chr.1: 63323523-63323523 on Build GRCh38
rs781444610 Chr.1: 63323323-63323323 on Build GRCh38
rs766617360 Chr.1: 63323526-63323526 on Build GRCh38
rs148930260 Chr.1: 63323465-63323465 on Build GRCh38
rs958468241 Chr.1: 63323354-63323354 on Build GRCh38
rs989637519 Chr.1: 63323355-63323355 on Build GRCh38
rs777668537 Chr.1: 63323452-63323452 on Build GRCh38
rs1024971175 Chr.1: 63323321-63323321 on Build GRCh38
rs999510880 Chr.1: 63323669-63323669 on Build GRCh38
rs151026788 Chr.1: 63323313-63323324 on Build GRCh38
rs748629940 Chr.1: 63323652-63323652 on Build GRCh38

View Full Product Details